Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93901495-93901994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCLM
Entrez Gene ID
2730
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
GLCLR
UniGene
Hs.315562
Gene Chromosome Location
Chr.1: 93885199-93909598 on Build GRCh38
Transcripts
NM_001308253.1 NM_002061.3 XM_011541261.2 XM_017001056.1 XM_017001057.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381739 c.199C>T p.P67S Chr.1: 93901663-93901663 on Build GRCh38
6670027 c.229G>A p.A77T Chr.1: 93901633-93901633 on Build GRCh38
5506746 c.210G>A p.L70L Chr.1: 93901652-93901652 on Build GRCh38
4686645 c.199C>A p.P67T Chr.1: 93901663-93901663 on Build GRCh38
123332 c.235G>C p.E79Q Chr.1: 93901627-93901627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188405780 Chr.1: 93901595-93901595 on Build GRCh38
rs775509179 Chr.1: 93901614-93901614 on Build GRCh38
rs895175955 Chr.1: 93901867-93901867 on Build GRCh38
rs369348308 Chr.1: 93901540-93901540 on Build GRCh38
rs918301580 Chr.1: 93901520-93901522 on Build GRCh38
rs368663137 Chr.1: 93901647-93901647 on Build GRCh38
rs796078494 Chr.1: 93901901-93901901 on Build GRCh38
rs933751443 Chr.1: 93901530-93901530 on Build GRCh38
rs886795848 Chr.1: 93901536-93901536 on Build GRCh38
rs371519356 Chr.1: 93901640-93901640 on Build GRCh38
rs754217749 Chr.1: 93901655-93901655 on Build GRCh38
rs17884538 Chr.1: 93901866-93901867 on Build GRCh38
rs370656484 Chr.1: 93901865-93901866 on Build GRCh38
rs200917594 Chr.1: 93901546-93901546 on Build GRCh38
rs769677808 Chr.1: 93901608-93901608 on Build GRCh38
rs112794990 Chr.1: 93901718-93901718 on Build GRCh38
rs1013129302 Chr.1: 93901702-93901702 on Build GRCh38
rs1024556234 Chr.1: 93901764-93901764 on Build GRCh38
rs779781205 Chr.1: 93901719-93901719 on Build GRCh38
rs971669682 Chr.1: 93901796-93901796 on Build GRCh38
rs754992214 Chr.1: 93901579-93901580 on Build GRCh38
rs745669935 Chr.1: 93901591-93901591 on Build GRCh38
rs1041944477 Chr.1: 93901804-93901804 on Build GRCh38
rs570537075 Chr.1: 93901848-93901848 on Build GRCh38
rs765454174 Chr.1: 93901664-93901664 on Build GRCh38
rs770214680 Chr.1: 93901933-93901933 on Build GRCh38
rs933886074 Chr.1: 93901966-93901966 on Build GRCh38
rs957407056 Chr.1: 93901822-93901822 on Build GRCh38
rs202172059 Chr.1: 93901683-93901683 on Build GRCh38
rs749242807 Chr.1: 93901720-93901720 on Build GRCh38
rs990605525 Chr.1: 93901843-93901843 on Build GRCh38
rs770595920 Chr.1: 93901565-93901565 on Build GRCh38
rs776329807 Chr.1: 93901587-93901587 on Build GRCh38
rs17886352 Chr.1: 93901925-93901925 on Build GRCh38
rs1004050407 Chr.1: 93901850-93901851 on Build GRCh38
rs1039540841 Chr.1: 93901902-93901902 on Build GRCh38
rs745453033 Chr.1: 93901715-93901715 on Build GRCh38
rs774813774 Chr.1: 93901765-93901765 on Build GRCh38
rs553549055 Chr.1: 93901869-93901869 on Build GRCh38
rs1803635 Chr.1: 93901646-93901646 on Build GRCh38
rs541564052 Chr.1: 93901799-93901799 on Build GRCh38
rs942574251 Chr.1: 93901619-93901619 on Build GRCh38
rs762806260 Chr.1: 93901618-93901618 on Build GRCh38
rs199971061 Chr.1: 93901569-93901569 on Build GRCh38
rs1035016641 Chr.1: 93901852-93901865 on Build GRCh38
rs910641432 Chr.1: 93901853-93901853 on Build GRCh38
rs1054412266 Chr.1: 93901701-93901701 on Build GRCh38
rs780884972 Chr.1: 93901700-93901700 on Build GRCh38
rs756925373 Chr.1: 93901699-93901699 on Build GRCh38
rs753288142 Chr.1: 93901676-93901676 on Build GRCh38
rs570044954 Chr.1: 93901638-93901638 on Build GRCh38
rs577373058 Chr.1: 93901748-93901748 on Build GRCh38
rs770480293 Chr.1: 93901661-93901661 on Build GRCh38
rs575742627 Chr.1: 93901823-93901823 on Build GRCh38
rs111457925 Chr.1: 93901865-93901865 on Build GRCh38
rs746485264 Chr.1: 93901691-93901691 on Build GRCh38
rs559168642 Chr.1: 93901687-93901687 on Build GRCh38
rs1046739032 Chr.1: 93901528-93901528 on Build GRCh38
rs924585885 Chr.1: 93901820-93901820 on Build GRCh38
rs368561212 Chr.1: 93901551-93901551 on Build GRCh38
rs748611057 Chr.1: 93901538-93901538 on Build GRCh38
rs7515191 Chr.1: 93901541-93901541 on Build GRCh38
rs942526278 Chr.1: 93901899-93901899 on Build GRCh38

View Full Product Details