Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29329620-29330117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
UniGene
Hs.368794
Gene Chromosome Location
Chr.22: 29327680-29388583 on Build GRCh38
Transcripts
NM_001127.3 NM_001166019.1 NM_145730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601463 c.2767-10C>T p.? Chr.22: 29329730-29329730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374891079 Chr.22: 29329729-29329729 on Build GRCh38
rs73884714 Chr.22: 29329926-29329926 on Build GRCh38
rs775515095 Chr.22: 29329734-29329734 on Build GRCh38
rs769123315 Chr.22: 29329731-29329731 on Build GRCh38
rs576997057 Chr.22: 29329932-29329932 on Build GRCh38
rs900444338 Chr.22: 29329976-29329976 on Build GRCh38
rs575958834 Chr.22: 29329816-29329816 on Build GRCh38
rs1007268096 Chr.22: 29329800-29329800 on Build GRCh38
rs924105665 Chr.22: 29329912-29329912 on Build GRCh38
rs983195003 Chr.22: 29329882-29329882 on Build GRCh38
rs748010657 Chr.22: 29329987-29329987 on Build GRCh38
rs554279393 Chr.22: 29329723-29329723 on Build GRCh38
rs1046527326 Chr.22: 29329704-29329704 on Build GRCh38
rs868633109 Chr.22: 29330012-29330012 on Build GRCh38
rs763049734 Chr.22: 29329988-29329988 on Build GRCh38
rs980877373 Chr.22: 29329916-29329916 on Build GRCh38
rs766554162 Chr.22: 29329681-29329681 on Build GRCh38
rs775286524 Chr.22: 29329675-29329675 on Build GRCh38
rs963086589 Chr.22: 29329889-29329889 on Build GRCh38
rs764590905 Chr.22: 29330046-29330046 on Build GRCh38
rs999554125 Chr.22: 29329772-29329772 on Build GRCh38
rs182325230 Chr.22: 29330081-29330081 on Build GRCh38
rs983142521 Chr.22: 29329646-29329646 on Build GRCh38
rs756292667 Chr.22: 29329680-29329680 on Build GRCh38
rs762814781 Chr.22: 29329676-29329676 on Build GRCh38
rs772053428 Chr.22: 29329777-29329777 on Build GRCh38
rs925061797 Chr.22: 29329669-29329669 on Build GRCh38
rs780801041 Chr.22: 29329917-29329917 on Build GRCh38
rs980446838 Chr.22: 29329656-29329656 on Build GRCh38
rs375872616 Chr.22: 29329673-29329673 on Build GRCh38
rs982275335 Chr.22: 29329907-29329907 on Build GRCh38
rs900392672 Chr.22: 29329813-29329813 on Build GRCh38
rs878925712 Chr.22: 29329801-29329801 on Build GRCh38
rs1007325520 Chr.22: 29329833-29329833 on Build GRCh38
rs990525743 Chr.22: 29330093-29330093 on Build GRCh38
rs892301056 Chr.22: 29329720-29329720 on Build GRCh38
rs1030154684 Chr.22: 29329835-29329835 on Build GRCh38
rs898457562 Chr.22: 29329874-29329874 on Build GRCh38
rs776410742 Chr.22: 29329726-29329726 on Build GRCh38
rs774167790 Chr.22: 29329764-29329764 on Build GRCh38
rs1025712103 Chr.22: 29329885-29329885 on Build GRCh38
rs545656640 Chr.22: 29329940-29329940 on Build GRCh38
rs111588517 Chr.22: 29329650-29329650 on Build GRCh38
rs969662860 Chr.22: 29329913-29329913 on Build GRCh38
rs778950718 Chr.22: 29329682-29329682 on Build GRCh38
rs963435984 Chr.22: 29330085-29330085 on Build GRCh38
rs189336409 Chr.22: 29330035-29330035 on Build GRCh38
rs773139954 Chr.22: 29329793-29329793 on Build GRCh38
rs770039679 Chr.22: 29329876-29329876 on Build GRCh38
rs1051643813 Chr.22: 29329741-29329741 on Build GRCh38
rs992317314 Chr.22: 29329902-29329902 on Build GRCh38
rs2071578 Chr.22: 29329904-29329904 on Build GRCh38
rs755033611 Chr.22: 29329683-29329683 on Build GRCh38
rs748536823 Chr.22: 29329686-29329686 on Build GRCh38
rs886274291 Chr.22: 29330044-29330044 on Build GRCh38
rs747347896 Chr.22: 29329701-29329701 on Build GRCh38
rs777949551 Chr.22: 29329700-29329700 on Build GRCh38
rs758802600 Chr.22: 29329692-29329692 on Build GRCh38
rs987867706 Chr.22: 29329931-29329931 on Build GRCh38
rs1051528371 Chr.22: 29329982-29329982 on Build GRCh38
rs1040090506 Chr.22: 29329963-29329963 on Build GRCh38
rs57406458 Chr.22: 29329925-29329925 on Build GRCh38
rs180689769 Chr.22: 29329685-29329685 on Build GRCh38
rs542746422 Chr.22: 29329852-29329854 on Build GRCh38
rs933131036 Chr.22: 29329738-29329738 on Build GRCh38
rs796889611 Chr.22: 29329926-29329926 on Build GRCh38
rs1018268522 Chr.22: 29329860-29329860 on Build GRCh38
rs765019826 Chr.22: 29329756-29329756 on Build GRCh38
rs1047208918 Chr.22: 29330068-29330068 on Build GRCh38
rs367690797 Chr.22: 29329817-29329817 on Build GRCh38
rs951540250 Chr.22: 29329897-29329897 on Build GRCh38
rs368833345 Chr.22: 29329678-29329678 on Build GRCh38
rs549652077 Chr.22: 29330067-29330067 on Build GRCh38
rs933183465 Chr.22: 29329973-29329973 on Build GRCh38
rs950479784 Chr.22: 29329867-29329867 on Build GRCh38
rs763983880 Chr.22: 29329757-29329757 on Build GRCh38
rs375989593 Chr.22: 29329792-29329792 on Build GRCh38

View Full Product Details