Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30554510-30555040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL3ST1
Entrez Gene ID
9514
Gene Name
galactose-3-O-sulfotransferase 1
Gene Aliases
CST
UniGene
Hs.17958
Gene Chromosome Location
Chr.22: 30554635-30574588 on Build GRCh38
Transcripts
NM_001318103.1 NM_001318104.1 NM_001318105.1 NM_001318106.1 NM_001318107.1 NM_001318108.1 NM_001318109.1 NM_001318110.1 NM_001318111.1 NM_001318112.1 NM_001318113.1 NM_001318114.1 NM_001318115.1 NM_001318116.1 NM_004861.2 XM_011530518.2 XM_011530522.2 XM_011530524.1 XM_011530528.1 XM_017029096.1 XM_017029097.1 XM_017029098.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
373091 c.1185G>C p.T395T Chr.22: 30555040-30555040 on Build GRCh38
6668849 c.1265G>A p.R422Q Chr.22: 30554960-30554960 on Build GRCh38
2150651 c.1193T>G p.I398S Chr.22: 30555032-30555032 on Build GRCh38
3553351 c.1260C>T p.F420F Chr.22: 30554965-30554965 on Build GRCh38
3553352 c.1255G>A p.D419N Chr.22: 30554970-30554970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897315417 Chr.22: 30554624-30554626 on Build GRCh38
rs904311316 Chr.22: 30554550-30554550 on Build GRCh38
rs1048558351 Chr.22: 30554871-30554871 on Build GRCh38
rs111476418 Chr.22: 30554820-30554820 on Build GRCh38
rs773819077 Chr.22: 30554912-30554912 on Build GRCh38
rs1034332223 Chr.22: 30554725-30554725 on Build GRCh38
rs560226917 Chr.22: 30554919-30554919 on Build GRCh38
rs756209851 Chr.22: 30554944-30554947 on Build GRCh38
rs762616205 Chr.22: 30554944-30554944 on Build GRCh38
rs897425126 Chr.22: 30554828-30554828 on Build GRCh38
rs749005721 Chr.22: 30554999-30554999 on Build GRCh38
rs11912600 Chr.22: 30554874-30554874 on Build GRCh38
rs1018471352 Chr.22: 30554595-30554595 on Build GRCh38
rs751363027 Chr.22: 30554636-30554636 on Build GRCh38
rs952828046 Chr.22: 30554715-30554715 on Build GRCh38
rs779510522 Chr.22: 30554980-30554980 on Build GRCh38
rs891756870 Chr.22: 30554537-30554537 on Build GRCh38
rs533394859 Chr.22: 30554823-30554823 on Build GRCh38
rs755659182 Chr.22: 30554976-30554976 on Build GRCh38
rs967233492 Chr.22: 30554750-30554753 on Build GRCh38
rs759730123 Chr.22: 30554928-30554928 on Build GRCh38
rs758790086 Chr.22: 30555005-30555005 on Build GRCh38
rs570468406 Chr.22: 30554559-30554559 on Build GRCh38
rs372198377 Chr.22: 30554603-30554603 on Build GRCh38
rs1025226937 Chr.22: 30554698-30554698 on Build GRCh38
rs370603459 Chr.22: 30554949-30554949 on Build GRCh38
rs1018106269 Chr.22: 30554824-30554824 on Build GRCh38
rs553067448 Chr.22: 30554717-30554717 on Build GRCh38
rs1010123772 Chr.22: 30554540-30554540 on Build GRCh38
rs765365089 Chr.22: 30554933-30554933 on Build GRCh38
rs984077944 Chr.22: 30554726-30554726 on Build GRCh38
rs1034729331 Chr.22: 30554895-30554895 on Build GRCh38
rs112220953 Chr.22: 30554855-30554855 on Build GRCh38
rs781170292 Chr.22: 30554970-30554970 on Build GRCh38
rs1050062276 Chr.22: 30554815-30554816 on Build GRCh38
rs960060727 Chr.22: 30554945-30554945 on Build GRCh38
rs75344364 Chr.22: 30554788-30554788 on Build GRCh38
rs544892534 Chr.22: 30554568-30554568 on Build GRCh38
rs950286197 Chr.22: 30554742-30554742 on Build GRCh38
rs529416934 Chr.22: 30554943-30554943 on Build GRCh38
rs879260721 Chr.22: 30554601-30554601 on Build GRCh38
rs76876427 Chr.22: 30554767-30554767 on Build GRCh38
rs994336939 Chr.22: 30554631-30554631 on Build GRCh38
rs776544661 Chr.22: 30554920-30554920 on Build GRCh38
rs1029511065 Chr.22: 30554554-30554554 on Build GRCh38
rs1025005275 Chr.22: 30555013-30555013 on Build GRCh38
rs573041237 Chr.22: 30554745-30554745 on Build GRCh38
rs115774030 Chr.22: 30554814-30554814 on Build GRCh38
rs917441344 Chr.22: 30554730-30554730 on Build GRCh38
rs1008485470 Chr.22: 30554588-30554588 on Build GRCh38
rs757152961 Chr.22: 30554961-30554961 on Build GRCh38
rs891773381 Chr.22: 30554794-30554794 on Build GRCh38
rs117382470 Chr.22: 30554562-30554562 on Build GRCh38
rs778263669 Chr.22: 30555018-30555018 on Build GRCh38
rs761289018 Chr.22: 30554916-30554916 on Build GRCh38
rs1041299941 Chr.22: 30554827-30554827 on Build GRCh38
rs973750541 Chr.22: 30554652-30554652 on Build GRCh38
rs751403921 Chr.22: 30554960-30554960 on Build GRCh38
rs749883972 Chr.22: 30554972-30554972 on Build GRCh38
rs995181507 Chr.22: 30554893-30554893 on Build GRCh38
rs936067365 Chr.22: 30554773-30554773 on Build GRCh38
rs867028859 Chr.22: 30554718-30554718 on Build GRCh38
rs994284583 Chr.22: 30554870-30554870 on Build GRCh38
rs891033620 Chr.22: 30554821-30554821 on Build GRCh38

View Full Product Details