Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30924339-30924880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MORC2-AS1
Entrez Gene ID
150291
Gene Name
MORC2 antisense RNA 1
Gene Aliases
C22orf27, NCRNA00325
UniGene
Hs.592202
Gene Chromosome Location
Chr.22: 30922308-30926654 on Build GRCh38
Transcripts
NR_026920.1

SNP Detail

SNP ID Chromosome Location
rs773948272 Chr.22: 30924791-30924792 on Build GRCh38
rs936838455 Chr.22: 30924447-30924447 on Build GRCh38
rs1009255102 Chr.22: 30924444-30924444 on Build GRCh38
rs368818364 Chr.22: 30924601-30924601 on Build GRCh38
rs75445079 Chr.22: 30924848-30924848 on Build GRCh38
rs967729621 Chr.22: 30924477-30924477 on Build GRCh38
rs958469234 Chr.22: 30924541-30924541 on Build GRCh38
rs28451096 Chr.22: 30924670-30924670 on Build GRCh38
rs1053371424 Chr.22: 30924425-30924425 on Build GRCh38
rs865953788 Chr.22: 30924705-30924705 on Build GRCh38
rs79093879 Chr.22: 30924849-30924849 on Build GRCh38
rs73156731 Chr.22: 30924521-30924521 on Build GRCh38
rs940618736 Chr.22: 30924372-30924372 on Build GRCh38
rs572898364 Chr.22: 30924386-30924386 on Build GRCh38
rs73156730 Chr.22: 30924507-30924507 on Build GRCh38
rs950663641 Chr.22: 30924508-30924508 on Build GRCh38
rs767430505 Chr.22: 30924682-30924683 on Build GRCh38
rs1034047046 Chr.22: 30924759-30924759 on Build GRCh38
rs770589124 Chr.22: 30924518-30924518 on Build GRCh38
rs748619929 Chr.22: 30924674-30924674 on Build GRCh38
rs534181250 Chr.22: 30924851-30924851 on Build GRCh38
rs182665207 Chr.22: 30924824-30924824 on Build GRCh38
rs762438418 Chr.22: 30924784-30924784 on Build GRCh38
rs892652627 Chr.22: 30924679-30924679 on Build GRCh38
rs777832409 Chr.22: 30924479-30924479 on Build GRCh38
rs1009833650 Chr.22: 30924494-30924494 on Build GRCh38
rs191399033 Chr.22: 30924788-30924788 on Build GRCh38
rs928667824 Chr.22: 30924576-30924576 on Build GRCh38
rs138949196 Chr.22: 30924608-30924608 on Build GRCh38
rs1007173901 Chr.22: 30924636-30924636 on Build GRCh38
rs78349997 Chr.22: 30924847-30924847 on Build GRCh38
rs1050541278 Chr.22: 30924429-30924429 on Build GRCh38
rs753582129 Chr.22: 30924832-30924832 on Build GRCh38
rs73884450 Chr.22: 30924800-30924800 on Build GRCh38
rs549245108 Chr.22: 30924637-30924637 on Build GRCh38
rs892004997 Chr.22: 30924433-30924433 on Build GRCh38
rs538462927 Chr.22: 30924669-30924669 on Build GRCh38
rs775047848 Chr.22: 30924684-30924684 on Build GRCh38
rs772055190 Chr.22: 30924739-30924739 on Build GRCh38
rs768609411 Chr.22: 30924510-30924513 on Build GRCh38
rs1016119512 Chr.22: 30924557-30924557 on Build GRCh38
rs965885222 Chr.22: 30924500-30924500 on Build GRCh38
rs970469849 Chr.22: 30924792-30924792 on Build GRCh38
rs975847881 Chr.22: 30924505-30924505 on Build GRCh38
rs41282555 Chr.22: 30924607-30924607 on Build GRCh38
rs541609829 Chr.22: 30924414-30924414 on Build GRCh38
rs903556731 Chr.22: 30924628-30924628 on Build GRCh38
rs529278846 Chr.22: 30924629-30924629 on Build GRCh38
rs561442104 Chr.22: 30924503-30924503 on Build GRCh38
rs759291020 Chr.22: 30924840-30924840 on Build GRCh38
rs765922209 Chr.22: 30924821-30924821 on Build GRCh38
rs984711724 Chr.22: 30924511-30924511 on Build GRCh38
rs1002527396 Chr.22: 30924733-30924733 on Build GRCh38
rs76971857 Chr.22: 30924850-30924850 on Build GRCh38
rs564053439 Chr.22: 30924546-30924546 on Build GRCh38
rs1003032818 Chr.22: 30924423-30924423 on Build GRCh38
rs901576703 Chr.22: 30924656-30924656 on Build GRCh38
rs950561678 Chr.22: 30924765-30924765 on Build GRCh38
rs972009141 Chr.22: 30924589-30924589 on Build GRCh38
rs772588961 Chr.22: 30924785-30924785 on Build GRCh38
rs1043295662 Chr.22: 30924621-30924621 on Build GRCh38
rs149521373 Chr.22: 30924577-30924577 on Build GRCh38
rs760286438 Chr.22: 30924814-30924814 on Build GRCh38
rs907407110 Chr.22: 30924411-30924411 on Build GRCh38
rs773225198 Chr.22: 30924686-30924686 on Build GRCh38
rs761134435 Chr.22: 30924843-30924843 on Build GRCh38
rs1022352637 Chr.22: 30924469-30924469 on Build GRCh38
rs60266623 Chr.22: 30924641-30924641 on Build GRCh38
rs750531720 Chr.22: 30924694-30924695 on Build GRCh38
rs28444814 Chr.22: 30924728-30924728 on Build GRCh38
rs1017306833 Chr.22: 30924483-30924483 on Build GRCh38

View Full Product Details