Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32418944-32419465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
UniGene
Hs.372939
Gene Chromosome Location
Chr.22: 32413845-32464465 on Build GRCh38
Transcripts
NM_174932.2 XM_011530088.2 XM_011530089.1 XM_011530090.1 XM_011530091.2 XM_017028740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500806 c.1223G>A p.R408H Chr.22: 32419399-32419399 on Build GRCh38
5829207 c.1218-3_1218-2insT p.? Chr.22: 32419406-32419407 on Build GRCh38
7277733 c.1250G>A p.S417N Chr.22: 32419372-32419372 on Build GRCh38
7128049 c.1258G>C p.E420Q Chr.22: 32419364-32419364 on Build GRCh38
77346 c.1222C>T p.R408C Chr.22: 32419400-32419400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942496167 Chr.22: 32419026-32419026 on Build GRCh38
rs775252947 Chr.22: 32419136-32419136 on Build GRCh38
rs376981555 Chr.22: 32419400-32419400 on Build GRCh38
rs760530176 Chr.22: 32419320-32419320 on Build GRCh38
rs977042956 Chr.22: 32419023-32419023 on Build GRCh38
rs1018185785 Chr.22: 32419224-32419224 on Build GRCh38
rs997860149 Chr.22: 32419319-32419319 on Build GRCh38
rs528073387 Chr.22: 32418994-32418994 on Build GRCh38
rs1041784169 Chr.22: 32419091-32419091 on Build GRCh38
rs577823695 Chr.22: 32419219-32419219 on Build GRCh38
rs763597920 Chr.22: 32419379-32419379 on Build GRCh38
rs768892131 Chr.22: 32419073-32419073 on Build GRCh38
rs926660239 Chr.22: 32419126-32419126 on Build GRCh38
rs764829563 Chr.22: 32419352-32419352 on Build GRCh38
rs986710861 Chr.22: 32418989-32418991 on Build GRCh38
rs750847402 Chr.22: 32419382-32419382 on Build GRCh38
rs1000388931 Chr.22: 32419195-32419195 on Build GRCh38
rs550672263 Chr.22: 32419137-32419137 on Build GRCh38
rs906830570 Chr.22: 32419092-32419092 on Build GRCh38
rs530488284 Chr.22: 32419114-32419114 on Build GRCh38
rs544938731 Chr.22: 32419398-32419398 on Build GRCh38
rs541453281 Chr.22: 32419034-32419034 on Build GRCh38
rs977175251 Chr.22: 32419293-32419293 on Build GRCh38
rs939351732 Chr.22: 32419130-32419130 on Build GRCh38
rs772117876 Chr.22: 32419311-32419311 on Build GRCh38
rs774970761 Chr.22: 32419416-32419416 on Build GRCh38
rs768276552 Chr.22: 32419230-32419230 on Build GRCh38
rs1018024191 Chr.22: 32419203-32419203 on Build GRCh38
rs547301539 Chr.22: 32419182-32419182 on Build GRCh38
rs201693028 Chr.22: 32419314-32419314 on Build GRCh38
rs55807703 Chr.22: 32419411-32419411 on Build GRCh38
rs533572974 Chr.22: 32419150-32419150 on Build GRCh38
rs750865237 Chr.22: 32419132-32419132 on Build GRCh38
rs748115890 Chr.22: 32419425-32419425 on Build GRCh38
rs189017258 Chr.22: 32419038-32419038 on Build GRCh38
rs773069555 Chr.22: 32419316-32419316 on Build GRCh38
rs781078569 Chr.22: 32419009-32419009 on Build GRCh38
rs779385905 Chr.22: 32419020-32419020 on Build GRCh38
rs375343312 Chr.22: 32419341-32419341 on Build GRCh38
rs1010078262 Chr.22: 32419144-32419144 on Build GRCh38
rs945766635 Chr.22: 32419089-32419089 on Build GRCh38
rs755398044 Chr.22: 32419401-32419401 on Build GRCh38
rs182416250 Chr.22: 32419141-32419141 on Build GRCh38
rs752111285 Chr.22: 32419371-32419371 on Build GRCh38
rs563087410 Chr.22: 32419303-32419303 on Build GRCh38
rs768036168 Chr.22: 32419336-32419337 on Build GRCh38
rs745683065 Chr.22: 32419033-32419033 on Build GRCh38
rs372526929 Chr.22: 32419399-32419399 on Build GRCh38
rs748730898 Chr.22: 32419204-32419204 on Build GRCh38
rs910028085 Chr.22: 32419071-32419071 on Build GRCh38
rs1045140700 Chr.22: 32419124-32419124 on Build GRCh38
rs1006328752 Chr.22: 32419145-32419145 on Build GRCh38
rs759169350 Chr.22: 32419348-32419348 on Build GRCh38
rs367601696 Chr.22: 32419183-32419183 on Build GRCh38
rs914377395 Chr.22: 32419235-32419235 on Build GRCh38
rs543560715 Chr.22: 32419292-32419292 on Build GRCh38
rs371751585 Chr.22: 32419328-32419328 on Build GRCh38
rs945790051 Chr.22: 32419245-32419245 on Build GRCh38
rs986742951 Chr.22: 32419205-32419205 on Build GRCh38
rs928307558 Chr.22: 32419307-32419307 on Build GRCh38
rs200223063 Chr.22: 32419051-32419051 on Build GRCh38
rs185456915 Chr.22: 32419253-32419253 on Build GRCh38
rs367976450 Chr.22: 32419376-32419376 on Build GRCh38

View Full Product Details