Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38480972-38481483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDELR3
Entrez Gene ID
11015
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Gene Aliases
ERD2L3
UniGene
Hs.745072
Gene Chromosome Location
Chr.22: 38468062-38483447 on Build GRCh38
Transcripts
NM_006855.3 NM_016657.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694844 c.475C>T p.R159W Chr.22: 38481335-38481335 on Build GRCh38
75100 c.581T>A p.F194Y Chr.22: 38481441-38481441 on Build GRCh38
391546 c.365_368delTCTC p.F122fs*15 Chr.22: 38481225-38481228 on Build GRCh38
4186920 c.401A>G p.Q134R Chr.22: 38481261-38481261 on Build GRCh38
4432210 c.370A>G p.I124V Chr.22: 38481230-38481230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778059482 Chr.22: 38481364-38481364 on Build GRCh38
rs780748239 Chr.22: 38481444-38481445 on Build GRCh38
rs1023317658 Chr.22: 38481153-38481153 on Build GRCh38
rs928506256 Chr.22: 38481366-38481366 on Build GRCh38
rs137890906 Chr.22: 38481445-38481445 on Build GRCh38
rs772536996 Chr.22: 38481389-38481389 on Build GRCh38
rs766103461 Chr.22: 38481336-38481336 on Build GRCh38
rs754884067 Chr.22: 38481230-38481230 on Build GRCh38
rs367613491 Chr.22: 38481280-38481280 on Build GRCh38
rs113665532 Chr.22: 38481103-38481103 on Build GRCh38
rs199829844 Chr.22: 38481246-38481246 on Build GRCh38
rs750126990 Chr.22: 38481221-38481221 on Build GRCh38
rs895211587 Chr.22: 38481132-38481132 on Build GRCh38
rs779385767 Chr.22: 38481379-38481379 on Build GRCh38
rs748203741 Chr.22: 38481235-38481235 on Build GRCh38
rs999691537 Chr.22: 38481101-38481103 on Build GRCh38
rs996290488 Chr.22: 38481074-38481076 on Build GRCh38
rs771411685 Chr.22: 38481270-38481270 on Build GRCh38
rs772491104 Chr.22: 38481297-38481297 on Build GRCh38
rs751089860 Chr.22: 38481398-38481399 on Build GRCh38
rs573844625 Chr.22: 38481061-38481061 on Build GRCh38
rs375263903 Chr.22: 38481212-38481212 on Build GRCh38
rs938645849 Chr.22: 38481397-38481397 on Build GRCh38
rs779740209 Chr.22: 38481135-38481135 on Build GRCh38
rs756894009 Chr.22: 38481200-38481200 on Build GRCh38
rs866042278 Chr.22: 38481223-38481223 on Build GRCh38
rs142169266 Chr.22: 38481365-38481365 on Build GRCh38
rs769911655 Chr.22: 38481170-38481170 on Build GRCh38
rs187315002 Chr.22: 38481174-38481174 on Build GRCh38
rs772721417 Chr.22: 38481404-38481404 on Build GRCh38
rs890536760 Chr.22: 38481363-38481363 on Build GRCh38
rs759226419 Chr.22: 38481427-38481427 on Build GRCh38
rs776630707 Chr.22: 38481166-38481166 on Build GRCh38
rs774523036 Chr.22: 38481182-38481183 on Build GRCh38
rs571401019 Chr.22: 38481031-38481031 on Build GRCh38
rs375606041 Chr.22: 38481185-38481185 on Build GRCh38
rs766896088 Chr.22: 38481182-38481182 on Build GRCh38
rs989981952 Chr.22: 38481047-38481047 on Build GRCh38
rs1046988586 Chr.22: 38481023-38481025 on Build GRCh38
rs756771962 Chr.22: 38481439-38481441 on Build GRCh38
rs370748041 Chr.22: 38481184-38481184 on Build GRCh38
rs749558596 Chr.22: 38481258-38481258 on Build GRCh38
rs941287806 Chr.22: 38481282-38481282 on Build GRCh38
rs145304147 Chr.22: 38481334-38481334 on Build GRCh38
rs558258642 Chr.22: 38481196-38481196 on Build GRCh38
rs781049581 Chr.22: 38481204-38481204 on Build GRCh38
rs767383010 Chr.22: 38481433-38481433 on Build GRCh38
rs769172472 Chr.22: 38481403-38481403 on Build GRCh38
rs1052271183 Chr.22: 38481089-38481089 on Build GRCh38
rs989929760 Chr.22: 38481275-38481275 on Build GRCh38
rs774871984 Chr.22: 38481271-38481271 on Build GRCh38
rs758160492 Chr.22: 38481224-38481224 on Build GRCh38
rs777221809 Chr.22: 38481317-38481320 on Build GRCh38
rs138432 Chr.22: 38481101-38481101 on Build GRCh38
rs1034763906 Chr.22: 38481430-38481430 on Build GRCh38
rs753780926 Chr.22: 38481225-38481228 on Build GRCh38
rs573089298 Chr.22: 38481227-38481230 on Build GRCh38
rs781009492 Chr.22: 38481351-38481351 on Build GRCh38
rs966075839 Chr.22: 38481023-38481023 on Build GRCh38
rs975777444 Chr.22: 38481029-38481029 on Build GRCh38
rs752521551 Chr.22: 38481272-38481272 on Build GRCh38
rs190091888 Chr.22: 38481109-38481109 on Build GRCh38
rs746241413 Chr.22: 38481384-38481384 on Build GRCh38
rs929797440 Chr.22: 38481012-38481012 on Build GRCh38
rs775830578 Chr.22: 38481391-38481391 on Build GRCh38
rs890653863 Chr.22: 38481095-38481095 on Build GRCh38
rs147633229 Chr.22: 38481349-38481349 on Build GRCh38
rs191001974 Chr.22: 38481179-38481179 on Build GRCh38
rs142887079 Chr.22: 38481226-38481226 on Build GRCh38
rs967178209 Chr.22: 38481310-38481310 on Build GRCh38
rs148490231 Chr.22: 38481307-38481307 on Build GRCh38
rs764097708 Chr.22: 38481030-38481030 on Build GRCh38
rs752571011 Chr.22: 38481440-38481440 on Build GRCh38
rs139737165 Chr.22: 38481229-38481229 on Build GRCh38
rs761314164 Chr.22: 38481402-38481402 on Build GRCh38
rs149783182 Chr.22: 38481239-38481239 on Build GRCh38
rs758686997 Chr.22: 38481459-38481459 on Build GRCh38
rs751302050 Chr.22: 38481419-38481419 on Build GRCh38
rs144678116 Chr.22: 38481299-38481299 on Build GRCh38
rs763604969 Chr.22: 38481186-38481186 on Build GRCh38
rs762650065 Chr.22: 38481335-38481335 on Build GRCh38
rs766354740 Chr.22: 38481224-38481226 on Build GRCh38
rs756093412 Chr.22: 38481441-38481441 on Build GRCh38
rs752725188 Chr.22: 38481355-38481355 on Build GRCh38
rs376950482 Chr.22: 38481189-38481189 on Build GRCh38
rs1037713405 Chr.22: 38481044-38481044 on Build GRCh38
rs765165940 Chr.22: 38481227-38481228 on Build GRCh38
rs1052608189 Chr.22: 38481396-38481396 on Build GRCh38
rs1049200436 Chr.22: 38481138-38481138 on Build GRCh38
rs928585449 Chr.22: 38481112-38481112 on Build GRCh38
rs762524712 Chr.22: 38481406-38481406 on Build GRCh38
rs1056474151 Chr.22: 38481121-38481121 on Build GRCh38
rs569016068 Chr.22: 38481115-38481115 on Build GRCh38
rs537216751 Chr.22: 38481022-38481023 on Build GRCh38
rs751359160 Chr.22: 38481348-38481348 on Build GRCh38
rs748343869 Chr.22: 38481168-38481168 on Build GRCh38
rs140765967 Chr.22: 38481451-38481451 on Build GRCh38
rs758152725 Chr.22: 38481300-38481301 on Build GRCh38
rs945936306 Chr.22: 38481064-38481064 on Build GRCh38
rs761734090 Chr.22: 38481202-38481203 on Build GRCh38
rs12004 Chr.22: 38481456-38481456 on Build GRCh38
rs1014529926 Chr.22: 38481019-38481019 on Build GRCh38
rs144367618 Chr.22: 38481411-38481411 on Build GRCh38
rs183028088 Chr.22: 38481148-38481148 on Build GRCh38
rs931959050 Chr.22: 38481350-38481350 on Build GRCh38
rs1024293701 Chr.22: 38481407-38481407 on Build GRCh38

View Full Product Details