Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39384346-39384838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNGR1
Entrez Gene ID
9145
Gene Name
synaptogyrin 1
Gene Aliases
-
UniGene
Hs.216226
Gene Chromosome Location
Chr.22: 39349949-39385588 on Build GRCh38
Transcripts
NM_004711.4

SNP Detail

SNP ID Chromosome Location
rs968876124 Chr.22: 39384756-39384756 on Build GRCh38
rs927275913 Chr.22: 39384452-39384452 on Build GRCh38
rs1019852438 Chr.22: 39384682-39384682 on Build GRCh38
rs1020499725 Chr.22: 39384711-39384711 on Build GRCh38
rs73414378 Chr.22: 39384382-39384382 on Build GRCh38
rs76437831 Chr.22: 39384802-39384802 on Build GRCh38
rs1044251226 Chr.22: 39384786-39384786 on Build GRCh38
rs1057438220 Chr.22: 39384752-39384752 on Build GRCh38
rs563297917 Chr.22: 39384511-39384511 on Build GRCh38
rs1008995362 Chr.22: 39384616-39384616 on Build GRCh38
rs373473073 Chr.22: 39384443-39384443 on Build GRCh38
rs757940416 Chr.22: 39384554-39384554 on Build GRCh38
rs545176527 Chr.22: 39384500-39384500 on Build GRCh38
rs924855637 Chr.22: 39384726-39384726 on Build GRCh38
rs530728094 Chr.22: 39384768-39384768 on Build GRCh38
rs1050603255 Chr.22: 39384587-39384587 on Build GRCh38
rs1010407286 Chr.22: 39384697-39384697 on Build GRCh38
rs542714908 Chr.22: 39384785-39384785 on Build GRCh38
rs1012780268 Chr.22: 39384404-39384404 on Build GRCh38
rs549547227 Chr.22: 39384710-39384710 on Build GRCh38
rs953904087 Chr.22: 39384609-39384609 on Build GRCh38
rs888244757 Chr.22: 39384787-39384787 on Build GRCh38
rs554071019 Chr.22: 39384793-39384793 on Build GRCh38
rs947641357 Chr.22: 39384672-39384672 on Build GRCh38
rs910684546 Chr.22: 39384670-39384670 on Build GRCh38
rs968829570 Chr.22: 39384423-39384423 on Build GRCh38
rs888766775 Chr.22: 39384588-39384588 on Build GRCh38
rs545513775 Chr.22: 39384389-39384389 on Build GRCh38
rs779522178 Chr.22: 39384513-39384513 on Build GRCh38
rs917180225 Chr.22: 39384489-39384489 on Build GRCh38
rs745337198 Chr.22: 39384528-39384528 on Build GRCh38
rs976809101 Chr.22: 39384529-39384529 on Build GRCh38
rs376731439 Chr.22: 39384526-39384526 on Build GRCh38
rs937356220 Chr.22: 39384467-39384467 on Build GRCh38
rs1005440550 Chr.22: 39384492-39384492 on Build GRCh38
rs989007807 Chr.22: 39384396-39384396 on Build GRCh38
rs895913560 Chr.22: 39384763-39384763 on Build GRCh38
rs934701842 Chr.22: 39384738-39384738 on Build GRCh38
rs5995725 Chr.22: 39384806-39384806 on Build GRCh38
rs903144156 Chr.22: 39384373-39384373 on Build GRCh38
rs553208529 Chr.22: 39384403-39384403 on Build GRCh38
rs1057019447 Chr.22: 39384470-39384470 on Build GRCh38
rs41274557 Chr.22: 39384814-39384814 on Build GRCh38
rs993258782 Chr.22: 39384807-39384807 on Build GRCh38
rs986274415 Chr.22: 39384634-39384634 on Build GRCh38
rs1022781924 Chr.22: 39384424-39384424 on Build GRCh38
rs186938340 Chr.22: 39384425-39384425 on Build GRCh38
rs745487765 Chr.22: 39384713-39384713 on Build GRCh38
rs965643144 Chr.22: 39384695-39384695 on Build GRCh38
rs183310929 Chr.22: 39384402-39384402 on Build GRCh38
rs979021724 Chr.22: 39384680-39384680 on Build GRCh38

View Full Product Details