Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41435459-41435958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOB2
Entrez Gene ID
10766
Gene Name
transducer of ERBB2, 2
Gene Aliases
APRO5, TOB4, TOBL, TROB2
UniGene
Hs.474978
Gene Chromosome Location
Chr.22: 41433488-41448230 on Build GRCh38
Transcripts
NM_016272.3 XM_005261315.2 XM_006724105.3 XM_017028539.1

SNP Detail

SNP ID Chromosome Location
rs935934101 Chr.22: 41435484-41435484 on Build GRCh38
rs1055515282 Chr.22: 41435819-41435819 on Build GRCh38
rs951613999 Chr.22: 41435642-41435642 on Build GRCh38
rs962341371 Chr.22: 41435626-41435626 on Build GRCh38
rs964114992 Chr.22: 41435741-41435741 on Build GRCh38
rs995776771 Chr.22: 41435628-41435628 on Build GRCh38
rs145523137 Chr.22: 41435600-41435600 on Build GRCh38
rs907009562 Chr.22: 41435778-41435778 on Build GRCh38
rs1054744895 Chr.22: 41435505-41435505 on Build GRCh38
rs987364030 Chr.22: 41435695-41435695 on Build GRCh38
rs889761189 Chr.22: 41435818-41435818 on Build GRCh38
rs750333663 Chr.22: 41435849-41435849 on Build GRCh38
rs989108394 Chr.22: 41435603-41435603 on Build GRCh38
rs891628897 Chr.22: 41435522-41435522 on Build GRCh38
rs1001297081 Chr.22: 41435791-41435791 on Build GRCh38
rs760315013 Chr.22: 41435547-41435547 on Build GRCh38
rs753414740 Chr.22: 41435744-41435744 on Build GRCh38
rs1045954428 Chr.22: 41435777-41435777 on Build GRCh38
rs936635375 Chr.22: 41435750-41435750 on Build GRCh38
rs752427630 Chr.22: 41435569-41435569 on Build GRCh38
rs182015675 Chr.22: 41435876-41435876 on Build GRCh38
rs761299825 Chr.22: 41435756-41435756 on Build GRCh38
rs1056876776 Chr.22: 41435737-41435737 on Build GRCh38
rs926910821 Chr.22: 41435719-41435719 on Build GRCh38
rs1010481715 Chr.22: 41435555-41435555 on Build GRCh38
rs765209041 Chr.22: 41435769-41435769 on Build GRCh38
rs945927816 Chr.22: 41435772-41435772 on Build GRCh38
rs1024091963 Chr.22: 41435557-41435557 on Build GRCh38
rs1026315365 Chr.22: 41435638-41435638 on Build GRCh38
rs148068050 Chr.22: 41435686-41435686 on Build GRCh38
rs1054418022 Chr.22: 41435758-41435758 on Build GRCh38
rs1019647141 Chr.22: 41435835-41435836 on Build GRCh38
rs899070837 Chr.22: 41435753-41435753 on Build GRCh38
rs867180806 Chr.22: 41435493-41435493 on Build GRCh38
rs906936012 Chr.22: 41435559-41435559 on Build GRCh38
rs189462855 Chr.22: 41435704-41435704 on Build GRCh38
rs995574829 Chr.22: 41435850-41435850 on Build GRCh38
rs914792637 Chr.22: 41435664-41435664 on Build GRCh38
rs1001223324 Chr.22: 41435585-41435585 on Build GRCh38
rs925295856 Chr.22: 41435746-41435746 on Build GRCh38
rs1025926716 Chr.22: 41435882-41435882 on Build GRCh38
rs547603303 Chr.22: 41435925-41435925 on Build GRCh38
rs951699904 Chr.22: 41435899-41435899 on Build GRCh38
rs912670696 Chr.22: 41435734-41435734 on Build GRCh38
rs537358102 Chr.22: 41435693-41435693 on Build GRCh38
rs980167125 Chr.22: 41435715-41435715 on Build GRCh38
rs954137062 Chr.22: 41435495-41435495 on Build GRCh38
rs969382795 Chr.22: 41435877-41435877 on Build GRCh38

View Full Product Details