Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41705119-41705621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
UniGene
Hs.116419
Gene Chromosome Location
Chr.22: 41699503-41799456 on Build GRCh38
Transcripts
NM_152513.3 XM_011529935.1 XM_011529936.1 XM_011529937.1 XM_011529938.1 XM_011529939.1 XM_011529940.1 XM_011529941.1 XM_011529942.2 XM_011529943.1 XM_011529944.1 XM_011529945.2 XM_011529946.1 XM_011529947.1 XM_011529950.1 XM_011529955.1 XM_011529956.2 XM_017028633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4542367 c.316G>A p.E106K Chr.22: 41705521-41705521 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753574669 Chr.22: 41705512-41705512 on Build GRCh38
rs751225314 Chr.22: 41705312-41705312 on Build GRCh38
rs752191292 Chr.22: 41705423-41705423 on Build GRCh38
rs900234550 Chr.22: 41705565-41705565 on Build GRCh38
rs773751490 Chr.22: 41705585-41705585 on Build GRCh38
rs546967698 Chr.22: 41705552-41705552 on Build GRCh38
rs878880092 Chr.22: 41705358-41705358 on Build GRCh38
rs149161286 Chr.22: 41705590-41705590 on Build GRCh38
rs74469910 Chr.22: 41705190-41705190 on Build GRCh38
rs55942231 Chr.22: 41705430-41705430 on Build GRCh38
rs56201088 Chr.22: 41705461-41705461 on Build GRCh38
rs763453165 Chr.22: 41705281-41705281 on Build GRCh38
rs944328229 Chr.22: 41705467-41705467 on Build GRCh38
rs1038740489 Chr.22: 41705537-41705537 on Build GRCh38
rs780567915 Chr.22: 41705374-41705374 on Build GRCh38
rs112451181 Chr.22: 41705369-41705369 on Build GRCh38
rs777287967 Chr.22: 41705575-41705575 on Build GRCh38
rs539012930 Chr.22: 41705597-41705597 on Build GRCh38
rs777128372 Chr.22: 41705462-41705462 on Build GRCh38
rs927414776 Chr.22: 41705193-41705193 on Build GRCh38
rs1007202587 Chr.22: 41705571-41705571 on Build GRCh38
rs959502470 Chr.22: 41705422-41705422 on Build GRCh38
rs567759468 Chr.22: 41705478-41705478 on Build GRCh38
rs571501140 Chr.22: 41705245-41705245 on Build GRCh38
rs796100945 Chr.22: 41705199-41705199 on Build GRCh38
rs1048804522 Chr.22: 41705521-41705521 on Build GRCh38
rs532004218 Chr.22: 41705360-41705360 on Build GRCh38
rs774059842 Chr.22: 41705575-41705576 on Build GRCh38
rs900268759 Chr.22: 41705176-41705176 on Build GRCh38
rs145210732 Chr.22: 41705333-41705333 on Build GRCh38
rs370522835 Chr.22: 41705460-41705460 on Build GRCh38
rs1032869426 Chr.22: 41705243-41705243 on Build GRCh38
rs1000415313 Chr.22: 41705192-41705192 on Build GRCh38
rs770312110 Chr.22: 41705579-41705579 on Build GRCh38
rs759045672 Chr.22: 41705491-41705491 on Build GRCh38
rs759088845 Chr.22: 41705587-41705587 on Build GRCh38
rs757068997 Chr.22: 41705523-41705523 on Build GRCh38
rs984832690 Chr.22: 41705397-41705397 on Build GRCh38
rs751278717 Chr.22: 41705488-41705488 on Build GRCh38
rs929538209 Chr.22: 41705452-41705452 on Build GRCh38
rs779981083 Chr.22: 41705572-41705572 on Build GRCh38
rs368574546 Chr.22: 41705176-41705177 on Build GRCh38
rs777693156 Chr.22: 41705508-41705508 on Build GRCh38
rs756001668 Chr.22: 41705501-41705501 on Build GRCh38
rs768169126 Chr.22: 41705568-41705573 on Build GRCh38
rs372442616 Chr.22: 41705576-41705576 on Build GRCh38
rs79889709 Chr.22: 41705177-41705177 on Build GRCh38
rs749008133 Chr.22: 41705506-41705507 on Build GRCh38
rs368304910 Chr.22: 41705494-41705494 on Build GRCh38
rs779069793 Chr.22: 41705532-41705532 on Build GRCh38
rs1018512362 Chr.22: 41705297-41705297 on Build GRCh38
rs529763403 Chr.22: 41705459-41705459 on Build GRCh38
rs970661927 Chr.22: 41705337-41705337 on Build GRCh38
rs888401088 Chr.22: 41705534-41705534 on Build GRCh38
rs535265530 Chr.22: 41705480-41705480 on Build GRCh38
rs540161473 Chr.22: 41705311-41705311 on Build GRCh38
rs371427254 Chr.22: 41705192-41705193 on Build GRCh38
rs553915337 Chr.22: 41705350-41705350 on Build GRCh38
rs981249838 Chr.22: 41705172-41705172 on Build GRCh38
rs75482385 Chr.22: 41705191-41705191 on Build GRCh38
rs772390537 Chr.22: 41705553-41705553 on Build GRCh38
rs958460207 Chr.22: 41705448-41705448 on Build GRCh38
rs973959556 Chr.22: 41705429-41705429 on Build GRCh38
rs774797482 Chr.22: 41705595-41705595 on Build GRCh38
rs189106712 Chr.22: 41705574-41705574 on Build GRCh38

View Full Product Details