Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42693155-42693649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A4GALT
Entrez Gene ID
53947
Gene Name
alpha 1,4-galactosyltransferase
Gene Aliases
A14GALT, A4GALT1, Gb3S, P(k), P1, P1PK, PK
UniGene
Hs.105956
Gene Chromosome Location
Chr.22: 42692115-42721301 on Build GRCh38
Transcripts
NM_001318038.1 NM_017436.5 XM_005261644.2 XM_005261646.4 XM_005261647.2 XM_005261648.3 XM_006724265.3 XM_006724266.3 XM_011530233.2 XM_017028831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1535383 c.353C>T p.P118L Chr.22: 42693599-42693599 on Build GRCh38
5440198 c.452G>A p.R151Q Chr.22: 42693500-42693500 on Build GRCh38
7196099 c.492G>C p.G164G Chr.22: 42693460-42693460 on Build GRCh38
1682306 c.635C>T p.S212F Chr.22: 42693317-42693317 on Build GRCh38
7224487 c.767G>T p.R256L Chr.22: 42693185-42693185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747585101 Chr.22: 42693400-42693400 on Build GRCh38
rs374341081 Chr.22: 42693508-42693508 on Build GRCh38
rs776140143 Chr.22: 42693577-42693577 on Build GRCh38
rs756476441 Chr.22: 42693193-42693193 on Build GRCh38
rs397514502 Chr.22: 42693321-42693321 on Build GRCh38
rs13058256 Chr.22: 42693280-42693280 on Build GRCh38
rs769709632 Chr.22: 42693309-42693309 on Build GRCh38
rs755543231 Chr.22: 42693187-42693187 on Build GRCh38
rs774598242 Chr.22: 42693325-42693325 on Build GRCh38
rs768659084 Chr.22: 42693226-42693226 on Build GRCh38
rs778860281 Chr.22: 42693516-42693516 on Build GRCh38
rs764245973 Chr.22: 42693422-42693422 on Build GRCh38
rs768937923 Chr.22: 42693522-42693522 on Build GRCh38
rs763428888 Chr.22: 42693500-42693500 on Build GRCh38
rs754483595 Chr.22: 42693291-42693291 on Build GRCh38
rs769744009 Chr.22: 42693542-42693542 on Build GRCh38
rs28940571 Chr.22: 42693200-42693200 on Build GRCh38
rs779714575 Chr.22: 42693217-42693217 on Build GRCh38
rs938590081 Chr.22: 42693243-42693243 on Build GRCh38
rs1019748688 Chr.22: 42693354-42693354 on Build GRCh38
rs570317019 Chr.22: 42693575-42693575 on Build GRCh38
rs28915383 Chr.22: 42693464-42693464 on Build GRCh38
rs550617900 Chr.22: 42693512-42693512 on Build GRCh38
rs769540282 Chr.22: 42693238-42693238 on Build GRCh38
rs915904656 Chr.22: 42693497-42693497 on Build GRCh38
rs775582651 Chr.22: 42693333-42693333 on Build GRCh38
rs376097487 Chr.22: 42693570-42693570 on Build GRCh38
rs753279376 Chr.22: 42693513-42693513 on Build GRCh38
rs1025296242 Chr.22: 42693610-42693610 on Build GRCh38
rs766552140 Chr.22: 42693249-42693249 on Build GRCh38
rs28940572 Chr.22: 42693392-42693392 on Build GRCh38
rs754189348 Chr.22: 42693279-42693279 on Build GRCh38
rs28915382 Chr.22: 42693511-42693511 on Build GRCh38
rs763170320 Chr.22: 42693248-42693248 on Build GRCh38
rs759146225 Chr.22: 42693438-42693438 on Build GRCh38
rs1033003135 Chr.22: 42693312-42693312 on Build GRCh38
rs1039182978 Chr.22: 42693476-42693476 on Build GRCh38
rs771507737 Chr.22: 42693404-42693405 on Build GRCh38
rs772269642 Chr.22: 42693405-42693405 on Build GRCh38
rs147133867 Chr.22: 42693305-42693305 on Build GRCh38
rs767605798 Chr.22: 42693265-42693265 on Build GRCh38
rs1041172195 Chr.22: 42693578-42693578 on Build GRCh38
rs777158954 Chr.22: 42693446-42693447 on Build GRCh38
rs928499253 Chr.22: 42693210-42693210 on Build GRCh38
rs781421948 Chr.22: 42693524-42693524 on Build GRCh38
rs748091098 Chr.22: 42693301-42693301 on Build GRCh38
rs1000201351 Chr.22: 42693300-42693300 on Build GRCh38
rs772764072 Chr.22: 42693183-42693183 on Build GRCh38
rs767153887 Chr.22: 42693441-42693441 on Build GRCh38
rs530734646 Chr.22: 42693241-42693241 on Build GRCh38
rs371893172 Chr.22: 42693426-42693426 on Build GRCh38
rs761090113 Chr.22: 42693334-42693334 on Build GRCh38
rs114722809 Chr.22: 42693585-42693585 on Build GRCh38
rs766205721 Chr.22: 42693599-42693599 on Build GRCh38
rs753227800 Chr.22: 42693190-42693190 on Build GRCh38
rs962566898 Chr.22: 42693384-42693384 on Build GRCh38
rs758251960 Chr.22: 42693517-42693517 on Build GRCh38
rs761776405 Chr.22: 42693234-42693234 on Build GRCh38
rs754641713 Chr.22: 42693367-42693367 on Build GRCh38
rs201965177 Chr.22: 42693427-42693427 on Build GRCh38
rs779713387 Chr.22: 42693332-42693332 on Build GRCh38
rs755279796 Chr.22: 42693609-42693609 on Build GRCh38
rs772205920 Chr.22: 42693471-42693471 on Build GRCh38
rs1034139444 Chr.22: 42693589-42693589 on Build GRCh38
rs150748224 Chr.22: 42693282-42693282 on Build GRCh38
rs757128526 Chr.22: 42693466-42693466 on Build GRCh38
rs141234083 Chr.22: 42693277-42693277 on Build GRCh38
rs755664626 Chr.22: 42693451-42693451 on Build GRCh38
rs780370330 Chr.22: 42693472-42693472 on Build GRCh38
rs1036278427 Chr.22: 42693205-42693205 on Build GRCh38
rs747658770 Chr.22: 42693389-42693389 on Build GRCh38
rs372494764 Chr.22: 42693567-42693567 on Build GRCh38
rs201411551 Chr.22: 42693275-42693275 on Build GRCh38
rs749195557 Chr.22: 42693393-42693393 on Build GRCh38
rs750440854 Chr.22: 42693588-42693588 on Build GRCh38
rs754275300 Chr.22: 42693348-42693348 on Build GRCh38
rs768921574 Chr.22: 42693474-42693474 on Build GRCh38
rs760757454 Chr.22: 42693417-42693417 on Build GRCh38
rs140487716 Chr.22: 42693594-42693594 on Build GRCh38
rs867333011 Chr.22: 42693421-42693421 on Build GRCh38
rs903347202 Chr.22: 42693341-42693341 on Build GRCh38
rs374580731 Chr.22: 42693296-42693296 on Build GRCh38
rs751330072 Chr.22: 42693604-42693604 on Build GRCh38
rs770354946 Chr.22: 42693482-42693482 on Build GRCh38
rs748193828 Chr.22: 42693541-42693541 on Build GRCh38
rs370246921 Chr.22: 42693624-42693624 on Build GRCh38
rs576019459 Chr.22: 42693436-42693436 on Build GRCh38
rs765750050 Chr.22: 42693432-42693432 on Build GRCh38
rs747290435 Chr.22: 42693520-42693520 on Build GRCh38
rs773475971 Chr.22: 42693492-42693492 on Build GRCh38
rs761360691 Chr.22: 42693580-42693580 on Build GRCh38
rs28915384 Chr.22: 42693283-42693283 on Build GRCh38
rs766624269 Chr.22: 42693501-42693501 on Build GRCh38
rs756072106 Chr.22: 42693390-42693390 on Build GRCh38
rs769360984 Chr.22: 42693582-42693582 on Build GRCh38
rs780577926 Chr.22: 42693374-42693374 on Build GRCh38
rs753211195 Chr.22: 42693270-42693270 on Build GRCh38
rs781617781 Chr.22: 42693618-42693618 on Build GRCh38
rs759801808 Chr.22: 42693326-42693326 on Build GRCh38
rs751576133 Chr.22: 42693178-42693178 on Build GRCh38
rs142381337 Chr.22: 42693576-42693576 on Build GRCh38
rs747167329 Chr.22: 42693473-42693473 on Build GRCh38
rs758708941 Chr.22: 42693315-42693315 on Build GRCh38
rs1037960020 Chr.22: 42693240-42693240 on Build GRCh38
rs980317710 Chr.22: 42693364-42693364 on Build GRCh38
rs751235324 Chr.22: 42693366-42693366 on Build GRCh38
rs762419257 Chr.22: 42693475-42693475 on Build GRCh38
rs139550090 Chr.22: 42693394-42693394 on Build GRCh38
rs778577677 Chr.22: 42693469-42693469 on Build GRCh38
rs776479206 Chr.22: 42693227-42693227 on Build GRCh38
rs765662216 Chr.22: 42693360-42693360 on Build GRCh38
rs746475139 Chr.22: 42693220-42693220 on Build GRCh38
rs145649275 Chr.22: 42693458-42693458 on Build GRCh38
rs369483198 Chr.22: 42693252-42693252 on Build GRCh38
rs773811911 Chr.22: 42693558-42693558 on Build GRCh38
rs74315453 Chr.22: 42693404-42693404 on Build GRCh38
rs775462635 Chr.22: 42693408-42693408 on Build GRCh38
rs368919958 Chr.22: 42693347-42693347 on Build GRCh38
rs750737826 Chr.22: 42693435-42693435 on Build GRCh38
rs781505090 Chr.22: 42693188-42693188 on Build GRCh38
rs752272593 Chr.22: 42693448-42693448 on Build GRCh38
rs757743963 Chr.22: 42693357-42693357 on Build GRCh38
rs778195937 Chr.22: 42693199-42693199 on Build GRCh38
rs756287846 Chr.22: 42693625-42693625 on Build GRCh38
rs779058273 Chr.22: 42693211-42693211 on Build GRCh38
rs770694629 Chr.22: 42693324-42693324 on Build GRCh38
rs149462925 Chr.22: 42693298-42693298 on Build GRCh38
rs891788462 Chr.22: 42693530-42693530 on Build GRCh38

View Full Product Details