Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44736686-44737188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR5
Entrez Gene ID
55615
Gene Name
proline rich 5
Gene Aliases
FLJ20185k, PP610, PROTOR-1, PROTOR1
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44668547-44737681 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017528.2 NM_001017529.2 NM_001017530.1 NM_001198721.1 NM_015366.3 NM_181333.3 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4388214 c.1008C>G p.P336P Chr.22: 44737019-44737019 on Build GRCh38
4460332 c.1161C>T p.F387F Chr.22: 44737172-44737172 on Build GRCh38
7081647 c.962G>C p.R321T Chr.22: 44737042-44737042 on Build GRCh38
7324185 c.1133C>T p.S378F Chr.22: 44737144-44737144 on Build GRCh38
1616540 c.820C>G p.R274G Chr.22: 44736831-44736831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745572575 Chr.22: 44736794-44736794 on Build GRCh38
rs773758546 Chr.22: 44736985-44736985 on Build GRCh38
rs758391118 Chr.22: 44737065-44737065 on Build GRCh38
rs375987474 Chr.22: 44736707-44736707 on Build GRCh38
rs376477477 Chr.22: 44736900-44736900 on Build GRCh38
rs377514542 Chr.22: 44736947-44736947 on Build GRCh38
rs745384019 Chr.22: 44737050-44737050 on Build GRCh38
rs767279783 Chr.22: 44736817-44736817 on Build GRCh38
rs759768271 Chr.22: 44737145-44737145 on Build GRCh38
rs551482108 Chr.22: 44737153-44737153 on Build GRCh38
rs780229778 Chr.22: 44736863-44736863 on Build GRCh38
rs192702673 Chr.22: 44736845-44736845 on Build GRCh38
rs955948035 Chr.22: 44736738-44736738 on Build GRCh38
rs771575780 Chr.22: 44736907-44736908 on Build GRCh38
rs767163318 Chr.22: 44736993-44736993 on Build GRCh38
rs781717773 Chr.22: 44736869-44736869 on Build GRCh38
rs200332751 Chr.22: 44736977-44736977 on Build GRCh38
rs980533615 Chr.22: 44737125-44737125 on Build GRCh38
rs985590783 Chr.22: 44736781-44736781 on Build GRCh38
rs368240295 Chr.22: 44736881-44736881 on Build GRCh38
rs747794043 Chr.22: 44736838-44736839 on Build GRCh38
rs1042503270 Chr.22: 44737116-44737116 on Build GRCh38
rs773421976 Chr.22: 44736809-44736809 on Build GRCh38
rs575116707 Chr.22: 44737075-44737075 on Build GRCh38
rs568772935 Chr.22: 44736950-44736950 on Build GRCh38
rs746141969 Chr.22: 44736913-44736913 on Build GRCh38
rs758576173 Chr.22: 44737018-44737018 on Build GRCh38
rs761889076 Chr.22: 44736777-44736777 on Build GRCh38
rs201222885 Chr.22: 44736974-44736974 on Build GRCh38
rs755944988 Chr.22: 44737033-44737033 on Build GRCh38
rs764098166 Chr.22: 44736831-44736831 on Build GRCh38
rs948693368 Chr.22: 44736948-44736948 on Build GRCh38
rs370424313 Chr.22: 44737146-44737146 on Build GRCh38
rs6006865 Chr.22: 44736905-44736905 on Build GRCh38
rs777674202 Chr.22: 44737085-44737085 on Build GRCh38
rs764816049 Chr.22: 44736891-44736891 on Build GRCh38
rs569490184 Chr.22: 44736896-44736896 on Build GRCh38
rs769793784 Chr.22: 44736975-44736975 on Build GRCh38
rs770807880 Chr.22: 44737039-44737039 on Build GRCh38
rs777881209 Chr.22: 44736880-44736880 on Build GRCh38
rs368882309 Chr.22: 44736902-44736902 on Build GRCh38
rs867365475 Chr.22: 44736897-44736897 on Build GRCh38
rs753023449 Chr.22: 44737149-44737149 on Build GRCh38
rs763205024 Chr.22: 44736814-44736814 on Build GRCh38
rs187629442 Chr.22: 44736973-44736973 on Build GRCh38
rs756998979 Chr.22: 44737151-44737151 on Build GRCh38
rs753229319 Chr.22: 44736832-44736832 on Build GRCh38
rs764509010 Chr.22: 44737012-44737012 on Build GRCh38
rs771175008 Chr.22: 44736882-44736882 on Build GRCh38
rs6006864 Chr.22: 44736824-44736824 on Build GRCh38
rs775081608 Chr.22: 44736883-44736883 on Build GRCh38
rs745884947 Chr.22: 44737098-44737098 on Build GRCh38
rs761791524 Chr.22: 44736932-44736932 on Build GRCh38
rs758171166 Chr.22: 44736859-44736859 on Build GRCh38
rs746239711 Chr.22: 44736737-44736737 on Build GRCh38
rs746523166 Chr.22: 44737155-44737155 on Build GRCh38
rs56299305 Chr.22: 44736951-44736951 on Build GRCh38
rs780152115 Chr.22: 44736866-44736866 on Build GRCh38
rs772648806 Chr.22: 44736744-44736744 on Build GRCh38
rs1044833148 Chr.22: 44736852-44736852 on Build GRCh38
rs761015439 Chr.22: 44737011-44737011 on Build GRCh38
rs372443410 Chr.22: 44736884-44736884 on Build GRCh38
rs145508795 Chr.22: 44736851-44736851 on Build GRCh38
rs376789084 Chr.22: 44736720-44736720 on Build GRCh38
rs373924360 Chr.22: 44736899-44736899 on Build GRCh38
rs200176020 Chr.22: 44736758-44736758 on Build GRCh38
rs113174550 Chr.22: 44736718-44736718 on Build GRCh38
rs373328428 Chr.22: 44736903-44736903 on Build GRCh38
rs763607745 Chr.22: 44737128-44737128 on Build GRCh38
rs747242549 Chr.22: 44736893-44736893 on Build GRCh38
rs575155687 Chr.22: 44736757-44736757 on Build GRCh38
rs756902571 Chr.22: 44736793-44736793 on Build GRCh38
rs867319528 Chr.22: 44736915-44736915 on Build GRCh38
rs149767702 Chr.22: 44736800-44736800 on Build GRCh38
rs769836444 Chr.22: 44736879-44736879 on Build GRCh38
rs755369530 Chr.22: 44736867-44736867 on Build GRCh38
rs148215157 Chr.22: 44736967-44736967 on Build GRCh38
rs757750907 Chr.22: 44737014-44737014 on Build GRCh38
rs776615270 Chr.22: 44736931-44736931 on Build GRCh38
rs750455297 Chr.22: 44737063-44737063 on Build GRCh38
rs756128125 Chr.22: 44736786-44736786 on Build GRCh38
rs36082900 Chr.22: 44736807-44736807 on Build GRCh38
rs751233180 Chr.22: 44736895-44736895 on Build GRCh38
rs201958852 Chr.22: 44737016-44737016 on Build GRCh38
rs369157990 Chr.22: 44736894-44736894 on Build GRCh38
rs749202808 Chr.22: 44737035-44737035 on Build GRCh38
rs765875767 Chr.22: 44736946-44736946 on Build GRCh38
rs769880055 Chr.22: 44736933-44736933 on Build GRCh38
rs146928678 Chr.22: 44737154-44737154 on Build GRCh38
rs759316874 Chr.22: 44736782-44736782 on Build GRCh38
rs769225171 Chr.22: 44736804-44736804 on Build GRCh38
rs773846550 Chr.22: 44737048-44737048 on Build GRCh38
rs753093304 Chr.22: 44736953-44736953 on Build GRCh38
rs778593611 Chr.22: 44736743-44736744 on Build GRCh38
rs749375092 Chr.22: 44736904-44736904 on Build GRCh38
rs767369731 Chr.22: 44736783-44736783 on Build GRCh38
rs374084190 Chr.22: 44737076-44737076 on Build GRCh38
rs777209182 Chr.22: 44736926-44736927 on Build GRCh38
rs776817721 Chr.22: 44736764-44736764 on Build GRCh38
rs748724459 Chr.22: 44736875-44736875 on Build GRCh38
rs749045779 Chr.22: 44736978-44736978 on Build GRCh38
rs929528475 Chr.22: 44736822-44736822 on Build GRCh38
rs757352288 Chr.22: 44736966-44736966 on Build GRCh38
rs113430838 Chr.22: 44736778-44736778 on Build GRCh38
rs199815736 Chr.22: 44736969-44736969 on Build GRCh38
rs780505145 Chr.22: 44736796-44736796 on Build GRCh38
rs761879773 Chr.22: 44736890-44736890 on Build GRCh38
rs766610770 Chr.22: 44737070-44737070 on Build GRCh38
rs774079458 Chr.22: 44736780-44736780 on Build GRCh38
rs544800634 Chr.22: 44736723-44736723 on Build GRCh38
rs564741534 Chr.22: 44736724-44736724 on Build GRCh38
rs751722922 Chr.22: 44737141-44737141 on Build GRCh38
rs548581539 Chr.22: 44736914-44736914 on Build GRCh38
rs369549687 Chr.22: 44737104-44737104 on Build GRCh38
rs747496256 Chr.22: 44736991-44736992 on Build GRCh38
rs776530226 Chr.22: 44736806-44736806 on Build GRCh38
rs776125406 Chr.22: 44736750-44736750 on Build GRCh38
rs780432497 Chr.22: 44737025-44737025 on Build GRCh38
rs766522809 Chr.22: 44737140-44737140 on Build GRCh38
rs770454822 Chr.22: 44736984-44736984 on Build GRCh38
rs375688714 Chr.22: 44736892-44736892 on Build GRCh38
rs950956217 Chr.22: 44736909-44736909 on Build GRCh38
rs773584515 Chr.22: 44737126-44737126 on Build GRCh38
rs756548067 Chr.22: 44736955-44736955 on Build GRCh38
rs778702308 Chr.22: 44737091-44737091 on Build GRCh38
rs558295511 Chr.22: 44737055-44737055 on Build GRCh38
rs765310412 Chr.22: 44737015-44737015 on Build GRCh38
rs1000069395 Chr.22: 44736733-44736733 on Build GRCh38
rs1037663986 Chr.22: 44736929-44736929 on Build GRCh38
rs771629067 Chr.22: 44737054-44737054 on Build GRCh38
rs779381637 Chr.22: 44736729-44736729 on Build GRCh38
rs556434563 Chr.22: 44736710-44736710 on Build GRCh38
rs753420006 Chr.22: 44736790-44736790 on Build GRCh38
rs770248126 Chr.22: 44737124-44737124 on Build GRCh38
rs754312502 Chr.22: 44737013-44737013 on Build GRCh38
rs753581449 Chr.22: 44737089-44737089 on Build GRCh38
rs370225939 Chr.22: 44737115-44737115 on Build GRCh38
rs770705854 Chr.22: 44737090-44737090 on Build GRCh38
rs900399194 Chr.22: 44736935-44736935 on Build GRCh38
rs762338085 Chr.22: 44737058-44737058 on Build GRCh38
rs375639228 Chr.22: 44737109-44737109 on Build GRCh38
rs778266321 Chr.22: 44736960-44736960 on Build GRCh38
rs760489840 Chr.22: 44736825-44736825 on Build GRCh38
rs759190486 Chr.22: 44736992-44736992 on Build GRCh38
rs776976590 Chr.22: 44737121-44737135 on Build GRCh38
rs139706999 Chr.22: 44736785-44736785 on Build GRCh38
rs776880552 Chr.22: 44737056-44737056 on Build GRCh38
rs373862910 Chr.22: 44736963-44736963 on Build GRCh38
rs763464271 Chr.22: 44736789-44736789 on Build GRCh38
rs775774233 Chr.22: 44736997-44736997 on Build GRCh38
rs970474448 Chr.22: 44737007-44737007 on Build GRCh38
rs528817849 Chr.22: 44736968-44736968 on Build GRCh38
rs543044954 Chr.22: 44736878-44736878 on Build GRCh38
rs762562267 Chr.22: 44736941-44736941 on Build GRCh38
rs764906131 Chr.22: 44736850-44736850 on Build GRCh38
rs772777832 Chr.22: 44736940-44736940 on Build GRCh38
rs776983827 Chr.22: 44737114-44737114 on Build GRCh38
rs941896507 Chr.22: 44737022-44737022 on Build GRCh38
rs979672097 Chr.22: 44736888-44736888 on Build GRCh38
rs141096517 Chr.22: 44737105-44737105 on Build GRCh38
rs370409725 Chr.22: 44737072-44737072 on Build GRCh38
rs771431872 Chr.22: 44737062-44737062 on Build GRCh38
rs779182167 Chr.22: 44736907-44736907 on Build GRCh38
rs576965839 Chr.22: 44736719-44736719 on Build GRCh38
rs776625029 Chr.22: 44736887-44736887 on Build GRCh38
rs921864641 Chr.22: 44737088-44737088 on Build GRCh38
rs771538941 Chr.22: 44737103-44737103 on Build GRCh38
rs747288609 Chr.22: 44737029-44737029 on Build GRCh38
rs529636044 Chr.22: 44736795-44736795 on Build GRCh38
rs768465019 Chr.22: 44736916-44736916 on Build GRCh38

View Full Product Details