Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15568272-15568790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HACL1
Entrez Gene ID
26061
Gene Name
2-hydroxyacyl-CoA lyase 1
Gene Aliases
2-HPCL, HPCL, HPCL2, PHYH2
UniGene
Hs.63290
Gene Chromosome Location
Chr.3: 15560704-15601852 on Build GRCh38
Transcripts
NM_001284413.1 NM_001284415.1 NM_001284416.1 NM_012260.3 NR_104315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495419 c.1215A>G p.G405G Chr.3: 15568467-15568467 on Build GRCh38
7193589 c.1121C>G p.P374R Chr.3: 15568561-15568561 on Build GRCh38
4523434 c.1214G>A p.G405E Chr.3: 15568468-15568468 on Build GRCh38
5989256 c.1234T>C p.Y412H Chr.3: 15568448-15568448 on Build GRCh38
3005121 c.1113delA p.K371fs*5 Chr.3: 15568569-15568569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574425967 Chr.3: 15568375-15568375 on Build GRCh38
rs144328498 Chr.3: 15568466-15568466 on Build GRCh38
rs770803683 Chr.3: 15568557-15568557 on Build GRCh38
rs773754383 Chr.3: 15568501-15568501 on Build GRCh38
rs1012773819 Chr.3: 15568766-15568766 on Build GRCh38
rs754323050 Chr.3: 15568510-15568510 on Build GRCh38
rs1039114004 Chr.3: 15568735-15568735 on Build GRCh38
rs200222129 Chr.3: 15568404-15568404 on Build GRCh38
rs887970314 Chr.3: 15568326-15568326 on Build GRCh38
rs371801877 Chr.3: 15568472-15568472 on Build GRCh38
rs147975662 Chr.3: 15568693-15568693 on Build GRCh38
rs769678407 Chr.3: 15568542-15568542 on Build GRCh38
rs923611970 Chr.3: 15568642-15568642 on Build GRCh38
rs981144058 Chr.3: 15568482-15568482 on Build GRCh38
rs937746636 Chr.3: 15568332-15568332 on Build GRCh38
rs753033322 Chr.3: 15568436-15568436 on Build GRCh38
rs144932943 Chr.3: 15568533-15568533 on Build GRCh38
rs201128934 Chr.3: 15568405-15568405 on Build GRCh38
rs186555110 Chr.3: 15568350-15568350 on Build GRCh38
rs748587626 Chr.3: 15568431-15568431 on Build GRCh38
rs370187154 Chr.3: 15568761-15568761 on Build GRCh38
rs764484510 Chr.3: 15568508-15568508 on Build GRCh38
rs765889307 Chr.3: 15568489-15568489 on Build GRCh38
rs1042455512 Chr.3: 15568680-15568680 on Build GRCh38
rs773187144 Chr.3: 15568550-15568550 on Build GRCh38
rs564680419 Chr.3: 15568728-15568728 on Build GRCh38
rs144301413 Chr.3: 15568517-15568517 on Build GRCh38
rs377278680 Chr.3: 15568576-15568576 on Build GRCh38
rs747973460 Chr.3: 15568473-15568473 on Build GRCh38
rs886460157 Chr.3: 15568763-15568763 on Build GRCh38
rs772737377 Chr.3: 15568568-15568569 on Build GRCh38
rs773273594 Chr.3: 15568484-15568484 on Build GRCh38
rs778910688 Chr.3: 15568465-15568465 on Build GRCh38
rs151003425 Chr.3: 15568559-15568559 on Build GRCh38
rs538291448 Chr.3: 15568504-15568504 on Build GRCh38
rs779832710 Chr.3: 15568395-15568395 on Build GRCh38
rs191062531 Chr.3: 15568649-15568649 on Build GRCh38
rs770633282 Chr.3: 15568387-15568387 on Build GRCh38
rs775521394 Chr.3: 15568644-15568644 on Build GRCh38
rs768995983 Chr.3: 15568415-15568422 on Build GRCh38
rs762338863 Chr.3: 15568637-15568638 on Build GRCh38
rs368794326 Chr.3: 15568408-15568408 on Build GRCh38
rs574738860 Chr.3: 15568537-15568537 on Build GRCh38
rs752404751 Chr.3: 15568503-15568503 on Build GRCh38
rs866896364 Chr.3: 15568431-15568432 on Build GRCh38
rs757718682 Chr.3: 15568607-15568607 on Build GRCh38
rs754634438 Chr.3: 15568411-15568411 on Build GRCh38
rs977714843 Chr.3: 15568615-15568615 on Build GRCh38
rs1026244366 Chr.3: 15568759-15568759 on Build GRCh38
rs370023923 Chr.3: 15568502-15568502 on Build GRCh38
rs756546304 Chr.3: 15568438-15568438 on Build GRCh38
rs905650 Chr.3: 15568563-15568563 on Build GRCh38
rs777190733 Chr.3: 15568429-15568430 on Build GRCh38
rs62243579 Chr.3: 15568737-15568737 on Build GRCh38
rs140972844 Chr.3: 15568478-15568478 on Build GRCh38
rs760337033 Chr.3: 15568352-15568352 on Build GRCh38
rs950600321 Chr.3: 15568519-15568519 on Build GRCh38
rs914892748 Chr.3: 15568346-15568346 on Build GRCh38
rs778239828 Chr.3: 15568439-15568439 on Build GRCh38
rs1039130681 Chr.3: 15568349-15568349 on Build GRCh38
rs776000265 Chr.3: 15568464-15568464 on Build GRCh38
rs566971734 Chr.3: 15568389-15568389 on Build GRCh38
rs1023191273 Chr.3: 15568767-15568767 on Build GRCh38
rs199661326 Chr.3: 15568532-15568532 on Build GRCh38
rs751456852 Chr.3: 15568764-15568764 on Build GRCh38
rs1054381055 Chr.3: 15568742-15568742 on Build GRCh38
rs749860820 Chr.3: 15568454-15568454 on Build GRCh38
rs749185994 Chr.3: 15568552-15568552 on Build GRCh38
rs1052466020 Chr.3: 15568654-15568654 on Build GRCh38
rs890612401 Chr.3: 15568679-15568679 on Build GRCh38
rs767121570 Chr.3: 15568574-15568574 on Build GRCh38
rs933427945 Chr.3: 15568673-15568673 on Build GRCh38
rs543169889 Chr.3: 15568451-15568451 on Build GRCh38
rs772460206 Chr.3: 15568320-15568320 on Build GRCh38
rs765679506 Chr.3: 15568630-15568630 on Build GRCh38
rs747425954 Chr.3: 15568408-15568408 on Build GRCh38
rs764394871 Chr.3: 15568584-15568584 on Build GRCh38
rs779013598 Chr.3: 15568515-15568515 on Build GRCh38
rs769201368 Chr.3: 15568432-15568433 on Build GRCh38
rs762768057 Chr.3: 15568710-15568710 on Build GRCh38
rs572981785 Chr.3: 15568687-15568687 on Build GRCh38
rs763021809 Chr.3: 15568486-15568486 on Build GRCh38
rs781569571 Chr.3: 15568418-15568418 on Build GRCh38
rs772218005 Chr.3: 15568469-15568469 on Build GRCh38
rs772788737 Chr.3: 15568336-15568336 on Build GRCh38
rs754235035 Chr.3: 15568595-15568595 on Build GRCh38
rs760369822 Chr.3: 15568581-15568581 on Build GRCh38

View Full Product Details