Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 17009502-17010045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLCL2
Entrez Gene ID
23228
Gene Name
phospholipase C like 2
Gene Aliases
PLCE2
UniGene
Hs.202010
Gene Chromosome Location
Chr.3: 16802691-17090606 on Build GRCh38
Transcripts
NM_001144382.1 NM_015184.5 XM_006713073.3 XM_017006022.1 XM_017006023.1 XM_017006024.1 XM_017006025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3590074 c.278C>T p.S93F Chr.3: 17010002-17010002 on Build GRCh38
4421849 c.89C>G p.S30C Chr.3: 17009813-17009813 on Build GRCh38
4002512 c.234C>A p.R78R Chr.3: 17009958-17009958 on Build GRCh38
3590065 c.79C>T p.R27C Chr.3: 17009803-17009803 on Build GRCh38
1720044 c.160G>A p.D54N Chr.3: 17009884-17009884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540695724 Chr.3: 17009860-17009860 on Build GRCh38
rs761824231 Chr.3: 17009719-17009719 on Build GRCh38
rs376021613 Chr.3: 17009654-17009654 on Build GRCh38
rs749373345 Chr.3: 17009810-17009810 on Build GRCh38
rs570594685 Chr.3: 17009849-17009849 on Build GRCh38
rs765769300 Chr.3: 17009928-17009928 on Build GRCh38
rs774451232 Chr.3: 17009693-17009693 on Build GRCh38
rs753054859 Chr.3: 17009976-17009976 on Build GRCh38
rs116233779 Chr.3: 17009664-17009664 on Build GRCh38
rs759132742 Chr.3: 17009956-17009956 on Build GRCh38
rs780485591 Chr.3: 17010013-17010013 on Build GRCh38
rs754662964 Chr.3: 17009787-17009787 on Build GRCh38
rs760499396 Chr.3: 17009698-17009698 on Build GRCh38
rs1016489666 Chr.3: 17009776-17009776 on Build GRCh38
rs187412128 Chr.3: 17009575-17009575 on Build GRCh38
rs762379829 Chr.3: 17009745-17009745 on Build GRCh38
rs769761633 Chr.3: 17009900-17009900 on Build GRCh38
rs200484259 Chr.3: 17009825-17009825 on Build GRCh38
rs28932173 Chr.3: 17009958-17009958 on Build GRCh38
rs758060101 Chr.3: 17009639-17009639 on Build GRCh38
rs1041557194 Chr.3: 17009628-17009628 on Build GRCh38
rs953039293 Chr.3: 17009715-17009715 on Build GRCh38
rs372927646 Chr.3: 17009997-17009997 on Build GRCh38
rs147244755 Chr.3: 17009934-17009934 on Build GRCh38
rs781707143 Chr.3: 17009669-17009669 on Build GRCh38
rs768605034 Chr.3: 17009700-17009700 on Build GRCh38
rs748421376 Chr.3: 17009977-17009977 on Build GRCh38
rs749955181 Chr.3: 17009638-17009638 on Build GRCh38
rs926704167 Chr.3: 17009931-17009931 on Build GRCh38
rs751240748 Chr.3: 17009754-17009754 on Build GRCh38
rs149142356 Chr.3: 17009712-17009712 on Build GRCh38
rs938020176 Chr.3: 17009581-17009581 on Build GRCh38
rs1041694597 Chr.3: 17010004-17010004 on Build GRCh38
rs781632971 Chr.3: 17009655-17009655 on Build GRCh38
rs781512811 Chr.3: 17009788-17009788 on Build GRCh38
rs779573986 Chr.3: 17009646-17009646 on Build GRCh38
rs745955226 Chr.3: 17010010-17010010 on Build GRCh38
rs141837634 Chr.3: 17009800-17009800 on Build GRCh38
rs374754114 Chr.3: 17009985-17009985 on Build GRCh38
rs1050968661 Chr.3: 17009560-17009560 on Build GRCh38
rs933987581 Chr.3: 17010016-17010016 on Build GRCh38
rs772349922 Chr.3: 17010012-17010012 on Build GRCh38
rs768818518 Chr.3: 17009906-17009906 on Build GRCh38
rs761771277 Chr.3: 17009885-17009885 on Build GRCh38
rs77416948 Chr.3: 17009936-17009936 on Build GRCh38
rs563475580 Chr.3: 17010003-17010003 on Build GRCh38
rs772442848 Chr.3: 17009859-17009859 on Build GRCh38
rs773327702 Chr.3: 17009665-17009665 on Build GRCh38
rs1011384102 Chr.3: 17009625-17009625 on Build GRCh38
rs762497210 Chr.3: 17009908-17009908 on Build GRCh38
rs754340410 Chr.3: 17009992-17009992 on Build GRCh38
rs947266828 Chr.3: 17010000-17010000 on Build GRCh38
rs893060090 Chr.3: 17009608-17009608 on Build GRCh38
rs1046506647 Chr.3: 17009563-17009563 on Build GRCh38
rs28498854 Chr.3: 17009589-17009589 on Build GRCh38
rs748622014 Chr.3: 17009656-17009656 on Build GRCh38
rs1035257129 Chr.3: 17009772-17009772 on Build GRCh38
rs148021156 Chr.3: 17009826-17009826 on Build GRCh38
rs749321876 Chr.3: 17009839-17009839 on Build GRCh38
rs773075259 Chr.3: 17009907-17009907 on Build GRCh38
rs17853614 Chr.3: 17009978-17009978 on Build GRCh38
rs368088540 Chr.3: 17009721-17009721 on Build GRCh38
rs749876948 Chr.3: 17009732-17009732 on Build GRCh38
rs868479386 Chr.3: 17009714-17009714 on Build GRCh38
rs7653834 Chr.3: 17009836-17009836 on Build GRCh38
rs1054721327 Chr.3: 17009617-17009617 on Build GRCh38
rs1002572945 Chr.3: 17009729-17009729 on Build GRCh38
rs781573166 Chr.3: 17009658-17009659 on Build GRCh38
rs778209736 Chr.3: 17009807-17009807 on Build GRCh38
rs771118894 Chr.3: 17009678-17009678 on Build GRCh38

View Full Product Details