Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25569666-25570161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARB
Entrez Gene ID
5915
Gene Name
retinoic acid receptor beta
Gene Aliases
HAP, MCOPS12, NR1B2, RARbeta1, RRB2
UniGene
Hs.543218
Gene Chromosome Location
Chr.3: 24829323-25597932 on Build GRCh38
Transcripts
NM_000965.4 NM_001290216.2 NM_001290217.1 NM_001290266.1 NM_001290276.1 NM_001290277.1 NM_001290300.1 NM_016152.3 NR_110892.1 NR_110893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1422187 c.559C>T p.L187F Chr.3: 25569847-25569847 on Build GRCh38
4500248 c.560C>T p.A187V Chr.3: 25569869-25569869 on Build GRCh38
3592098 c.554G>A p.R185Q Chr.3: 25569863-25569863 on Build GRCh38
3592099 c.575G>A p.R192Q Chr.3: 25569863-25569863 on Build GRCh38
4500249 c.581C>T p.A194V Chr.3: 25569869-25569869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754683921 Chr.3: 25569888-25569888 on Build GRCh38
rs756224180 Chr.3: 25569841-25569841 on Build GRCh38
rs930719246 Chr.3: 25569930-25569930 on Build GRCh38
rs61195279 Chr.3: 25569982-25569985 on Build GRCh38
rs966888167 Chr.3: 25569990-25569990 on Build GRCh38
rs866881934 Chr.3: 25569993-25569993 on Build GRCh38
rs753088209 Chr.3: 25569837-25569837 on Build GRCh38
rs968122923 Chr.3: 25569692-25569692 on Build GRCh38
rs775765472 Chr.3: 25569988-25569991 on Build GRCh38
rs761412606 Chr.3: 25569798-25569798 on Build GRCh38
rs200899531 Chr.3: 25569935-25569935 on Build GRCh38
rs369808161 Chr.3: 25569982-25569991 on Build GRCh38
rs1022440319 Chr.3: 25569995-25569995 on Build GRCh38
rs772764423 Chr.3: 25569917-25569917 on Build GRCh38
rs1022421744 Chr.3: 25569972-25569972 on Build GRCh38
rs779249054 Chr.3: 25569885-25569885 on Build GRCh38
rs746323284 Chr.3: 25569990-25569991 on Build GRCh38
rs778155399 Chr.3: 25569743-25569743 on Build GRCh38
rs796553015 Chr.3: 25570018-25570021 on Build GRCh38
rs757604943 Chr.3: 25569862-25569862 on Build GRCh38
rs1019642090 Chr.3: 25570009-25570009 on Build GRCh38
rs762658175 Chr.3: 25569818-25569818 on Build GRCh38
rs778067457 Chr.3: 25569843-25569843 on Build GRCh38
rs372148180 Chr.3: 25569797-25569797 on Build GRCh38
rs878903574 Chr.3: 25570073-25570073 on Build GRCh38
rs563296763 Chr.3: 25570106-25570106 on Build GRCh38
rs397988973 Chr.3: 25570020-25570021 on Build GRCh38
rs548423302 Chr.3: 25570107-25570107 on Build GRCh38
rs1656455 Chr.3: 25570135-25570135 on Build GRCh38
rs761455977 Chr.3: 25570114-25570114 on Build GRCh38
rs780537028 Chr.3: 25570013-25570022 on Build GRCh38
rs922031826 Chr.3: 25569891-25569891 on Build GRCh38
rs768100812 Chr.3: 25569825-25569825 on Build GRCh38
rs759373266 Chr.3: 25569788-25569790 on Build GRCh38
rs535282454 Chr.3: 25570047-25570047 on Build GRCh38
rs775919820 Chr.3: 25569940-25569940 on Build GRCh38
rs996836222 Chr.3: 25569991-25569991 on Build GRCh38
rs750549174 Chr.3: 25569962-25569962 on Build GRCh38
rs765583131 Chr.3: 25569961-25569961 on Build GRCh38
rs869073242 Chr.3: 25570098-25570098 on Build GRCh38
rs577093267 Chr.3: 25569725-25569725 on Build GRCh38
rs12490204 Chr.3: 25570083-25570083 on Build GRCh38
rs763093963 Chr.3: 25570013-25570015 on Build GRCh38
rs566023799 Chr.3: 25570137-25570137 on Build GRCh38
rs1044203022 Chr.3: 25569854-25569854 on Build GRCh38
rs969960503 Chr.3: 25570117-25570117 on Build GRCh38
rs556962373 Chr.3: 25569690-25569690 on Build GRCh38
rs773834116 Chr.3: 25569929-25569929 on Build GRCh38
rs960821845 Chr.3: 25570051-25570051 on Build GRCh38
rs749456039 Chr.3: 25569749-25569749 on Build GRCh38
rs56862219 Chr.3: 25569982-25569989 on Build GRCh38
rs67669779 Chr.3: 25569982-25569983 on Build GRCh38
rs746760676 Chr.3: 25569773-25569773 on Build GRCh38
rs770609032 Chr.3: 25569927-25569927 on Build GRCh38
rs978222169 Chr.3: 25570005-25570005 on Build GRCh38
rs930728683 Chr.3: 25570030-25570030 on Build GRCh38
rs916574564 Chr.3: 25570082-25570082 on Build GRCh38
rs78456013 Chr.3: 25570139-25570139 on Build GRCh38
rs780919447 Chr.3: 25569967-25569967 on Build GRCh38
rs966299460 Chr.3: 25570016-25570016 on Build GRCh38
rs201331941 Chr.3: 25570019-25570022 on Build GRCh38
rs781198504 Chr.3: 25569894-25569894 on Build GRCh38
rs755565546 Chr.3: 25569723-25569723 on Build GRCh38
rs780055646 Chr.3: 25569755-25569755 on Build GRCh38
rs769232649 Chr.3: 25569804-25569804 on Build GRCh38
rs200850244 Chr.3: 25569821-25569821 on Build GRCh38
rs751913758 Chr.3: 25569716-25569716 on Build GRCh38
rs375687313 Chr.3: 25569925-25569925 on Build GRCh38
rs754013803 Chr.3: 25569953-25569953 on Build GRCh38
rs796858774 Chr.3: 25570014-25570021 on Build GRCh38
rs757375575 Chr.3: 25569957-25569957 on Build GRCh38
rs367760865 Chr.3: 25569750-25569750 on Build GRCh38
rs766001096 Chr.3: 25569819-25569819 on Build GRCh38
rs560811696 Chr.3: 25570027-25570027 on Build GRCh38
rs766660422 Chr.3: 25569712-25569712 on Build GRCh38
rs149559768 Chr.3: 25569912-25569912 on Build GRCh38
rs146549207 Chr.3: 25569822-25569822 on Build GRCh38
rs879247724 Chr.3: 25569989-25569990 on Build GRCh38
rs201006106 Chr.3: 25569839-25569840 on Build GRCh38
rs747965385 Chr.3: 25569968-25569968 on Build GRCh38
rs371805717 Chr.3: 25569937-25569937 on Build GRCh38
rs1011253235 Chr.3: 25569971-25569971 on Build GRCh38
rs747964713 Chr.3: 25569903-25569903 on Build GRCh38
rs56255512 Chr.3: 25570012-25570021 on Build GRCh38
rs199786350 Chr.3: 25570043-25570043 on Build GRCh38
rs549490224 Chr.3: 25570062-25570062 on Build GRCh38
rs892896546 Chr.3: 25569970-25569970 on Build GRCh38
rs56277016 Chr.3: 25569731-25569731 on Build GRCh38
rs772852342 Chr.3: 25569811-25569811 on Build GRCh38
rs921924988 Chr.3: 25570020-25570020 on Build GRCh38
rs879129265 Chr.3: 25569988-25569989 on Build GRCh38
rs753955955 Chr.3: 25569849-25569849 on Build GRCh38
rs975164165 Chr.3: 25570019-25570019 on Build GRCh38
rs141390071 Chr.3: 25569963-25569963 on Build GRCh38
rs575945786 Chr.3: 25569697-25569697 on Build GRCh38
rs1031074643 Chr.3: 25570034-25570034 on Build GRCh38
rs201595452 Chr.3: 25569944-25569944 on Build GRCh38
rs944738824 Chr.3: 25569856-25569856 on Build GRCh38
rs144176800 Chr.3: 25569726-25569726 on Build GRCh38
rs990079045 Chr.3: 25569686-25569686 on Build GRCh38
rs764677514 Chr.3: 25569950-25569950 on Build GRCh38

View Full Product Details