Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47418230-47418719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAP
Entrez Gene ID
22937
Gene Name
SREBF chaperone
Gene Aliases
-
UniGene
Hs.531789
Gene Chromosome Location
Chr.3: 47413685-47477126 on Build GRCh38
Transcripts
NM_001320044.1 NM_012235.3 XM_005264967.1 XM_005264968.1 XM_005264971.1 XM_011533501.1 XM_011533502.2 XM_017005918.1 XM_017005919.1 XM_017005920.1 XM_017005921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4653701 c.2195T>C p.V732A Chr.3: 47418457-47418457 on Build GRCh38
4879759 c.2130-9C>T p.? Chr.3: 47418531-47418531 on Build GRCh38
1224753 c.2206C>T p.R736C Chr.3: 47418446-47418446 on Build GRCh38
5819652 c.2307G>C p.P769P Chr.3: 47418345-47418345 on Build GRCh38
1045604 c.2115C>T p.D705D Chr.3: 47418669-47418669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910050687 Chr.3: 47418258-47418258 on Build GRCh38
rs371849082 Chr.3: 47418513-47418513 on Build GRCh38
rs370895515 Chr.3: 47418345-47418345 on Build GRCh38
rs760837280 Chr.3: 47418461-47418461 on Build GRCh38
rs745442359 Chr.3: 47418363-47418363 on Build GRCh38
rs867079103 Chr.3: 47418606-47418606 on Build GRCh38
rs1010042501 Chr.3: 47418533-47418533 on Build GRCh38
rs200867262 Chr.3: 47418615-47418615 on Build GRCh38
rs867962723 Chr.3: 47418331-47418331 on Build GRCh38
rs370092118 Chr.3: 47418263-47418263 on Build GRCh38
rs773690096 Chr.3: 47418335-47418336 on Build GRCh38
rs1033297853 Chr.3: 47418351-47418351 on Build GRCh38
rs570896201 Chr.3: 47418504-47418504 on Build GRCh38
rs892838001 Chr.3: 47418531-47418531 on Build GRCh38
rs761628759 Chr.3: 47418378-47418378 on Build GRCh38
rs772986145 Chr.3: 47418448-47418448 on Build GRCh38
rs111955430 Chr.3: 47418407-47418407 on Build GRCh38
rs780667235 Chr.3: 47418529-47418529 on Build GRCh38
rs373538536 Chr.3: 47418273-47418273 on Build GRCh38
rs762827179 Chr.3: 47418546-47418546 on Build GRCh38
rs761001660 Chr.3: 47418650-47418650 on Build GRCh38
rs748659730 Chr.3: 47418348-47418348 on Build GRCh38
rs555711337 Chr.3: 47418482-47418482 on Build GRCh38
rs576433381 Chr.3: 47418689-47418689 on Build GRCh38
rs749769930 Chr.3: 47418409-47418409 on Build GRCh38
rs867346192 Chr.3: 47418346-47418346 on Build GRCh38
rs541369635 Chr.3: 47418668-47418668 on Build GRCh38
rs766611578 Chr.3: 47418332-47418332 on Build GRCh38
rs758670894 Chr.3: 47418302-47418302 on Build GRCh38
rs753941312 Chr.3: 47418574-47418574 on Build GRCh38
rs768013421 Chr.3: 47418441-47418441 on Build GRCh38
rs765343510 Chr.3: 47418665-47418665 on Build GRCh38
rs528093394 Chr.3: 47418401-47418401 on Build GRCh38
rs773611737 Chr.3: 47418268-47418268 on Build GRCh38
rs1045394129 Chr.3: 47418326-47418326 on Build GRCh38
rs370331755 Chr.3: 47418370-47418370 on Build GRCh38
rs565303881 Chr.3: 47418375-47418375 on Build GRCh38
rs761681586 Chr.3: 47418446-47418446 on Build GRCh38
rs757697127 Chr.3: 47418410-47418410 on Build GRCh38
rs566797657 Chr.3: 47418403-47418403 on Build GRCh38
rs755179051 Chr.3: 47418386-47418386 on Build GRCh38
rs376996018 Chr.3: 47418492-47418492 on Build GRCh38
rs777219119 Chr.3: 47418623-47418623 on Build GRCh38
rs770674034 Chr.3: 47418279-47418279 on Build GRCh38
rs529398318 Chr.3: 47418626-47418626 on Build GRCh38
rs867157588 Chr.3: 47418523-47418523 on Build GRCh38
rs939100017 Chr.3: 47418621-47418622 on Build GRCh38
rs751741789 Chr.3: 47418339-47418339 on Build GRCh38
rs773999691 Chr.3: 47418640-47418640 on Build GRCh38
rs531845963 Chr.3: 47418299-47418299 on Build GRCh38
rs542816229 Chr.3: 47418627-47418627 on Build GRCh38
rs377478393 Chr.3: 47418296-47418296 on Build GRCh38
rs866383001 Chr.3: 47418496-47418496 on Build GRCh38
rs770769434 Chr.3: 47418638-47418638 on Build GRCh38
rs766890714 Chr.3: 47418627-47418631 on Build GRCh38
rs757884343 Chr.3: 47418536-47418536 on Build GRCh38
rs997386954 Chr.3: 47418543-47418543 on Build GRCh38
rs746555026 Chr.3: 47418438-47418438 on Build GRCh38
rs767092316 Chr.3: 47418289-47418289 on Build GRCh38
rs1050372965 Chr.3: 47418249-47418249 on Build GRCh38
rs751838887 Chr.3: 47418688-47418688 on Build GRCh38
rs369222944 Chr.3: 47418498-47418498 on Build GRCh38
rs774051545 Chr.3: 47418284-47418284 on Build GRCh38
rs768982772 Chr.3: 47418659-47418659 on Build GRCh38
rs773872046 Chr.3: 47418671-47418671 on Build GRCh38
rs776385822 Chr.3: 47418377-47418377 on Build GRCh38
rs922588557 Chr.3: 47418399-47418399 on Build GRCh38
rs766906645 Chr.3: 47418471-47418472 on Build GRCh38
rs774915990 Chr.3: 47418417-47418417 on Build GRCh38
rs369991877 Chr.3: 47418693-47418693 on Build GRCh38
rs768225701 Chr.3: 47418528-47418528 on Build GRCh38
rs764289267 Chr.3: 47418566-47418566 on Build GRCh38
rs373741294 Chr.3: 47418383-47418383 on Build GRCh38
rs775782835 Chr.3: 47418553-47418553 on Build GRCh38
rs748621732 Chr.3: 47418262-47418262 on Build GRCh38
rs527488110 Chr.3: 47418406-47418406 on Build GRCh38
rs75114530 Chr.3: 47418357-47418357 on Build GRCh38
rs375407546 Chr.3: 47418559-47418559 on Build GRCh38
rs776946620 Chr.3: 47418661-47418661 on Build GRCh38
rs752304672 Chr.3: 47418694-47418694 on Build GRCh38
rs763611933 Chr.3: 47418412-47418418 on Build GRCh38
rs769687752 Chr.3: 47418537-47418537 on Build GRCh38
rs753641503 Chr.3: 47418300-47418300 on Build GRCh38
rs779359225 Chr.3: 47418412-47418412 on Build GRCh38
rs762463972 Chr.3: 47418605-47418605 on Build GRCh38
rs771728213 Chr.3: 47418416-47418416 on Build GRCh38
rs1013946027 Chr.3: 47418312-47418312 on Build GRCh38
rs555150526 Chr.3: 47418663-47418663 on Build GRCh38
rs777715695 Chr.3: 47418633-47418633 on Build GRCh38
rs113000966 Chr.3: 47418617-47418617 on Build GRCh38
rs865875852 Chr.3: 47418321-47418321 on Build GRCh38
rs759136549 Chr.3: 47418285-47418285 on Build GRCh38
rs11546518 Chr.3: 47418359-47418359 on Build GRCh38
rs754581570 Chr.3: 47418621-47418621 on Build GRCh38
rs372058889 Chr.3: 47418476-47418476 on Build GRCh38
rs776635364 Chr.3: 47418445-47418445 on Build GRCh38
rs771769586 Chr.3: 47418366-47418366 on Build GRCh38
rs149033051 Chr.3: 47418402-47418402 on Build GRCh38
rs377014914 Chr.3: 47418618-47418618 on Build GRCh38
rs746578522 Chr.3: 47418514-47418514 on Build GRCh38
rs201478816 Chr.3: 47418447-47418447 on Build GRCh38
rs867345228 Chr.3: 47418664-47418664 on Build GRCh38
rs774174292 Chr.3: 47418362-47418362 on Build GRCh38
rs769243198 Chr.3: 47418630-47418630 on Build GRCh38
rs749110335 Chr.3: 47418635-47418635 on Build GRCh38
rs368756779 Chr.3: 47418459-47418459 on Build GRCh38
rs763355897 Chr.3: 47418677-47418677 on Build GRCh38
rs751943955 Chr.3: 47418570-47418571 on Build GRCh38
rs770589028 Chr.3: 47418547-47418547 on Build GRCh38
rs376014393 Chr.3: 47418358-47418358 on Build GRCh38
rs765869950 Chr.3: 47418608-47418608 on Build GRCh38
rs760746234 Chr.3: 47418558-47418558 on Build GRCh38
rs754531613 Chr.3: 47418607-47418608 on Build GRCh38
rs756639053 Chr.3: 47418350-47418350 on Build GRCh38
rs374707747 Chr.3: 47418453-47418453 on Build GRCh38
rs766835027 Chr.3: 47418685-47418685 on Build GRCh38
rs371768067 Chr.3: 47418413-47418413 on Build GRCh38
rs538275805 Chr.3: 47418538-47418538 on Build GRCh38
rs767756854 Chr.3: 47418458-47418458 on Build GRCh38
rs377388205 Chr.3: 47418369-47418369 on Build GRCh38

View Full Product Details