Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49809737-49810244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBA7
Entrez Gene ID
7318
Gene Name
ubiquitin like modifier activating enzyme 7
Gene Aliases
D8, UBA1B, UBE1L, UBE2
UniGene
Hs.16695
Gene Chromosome Location
Chr.3: 49805205-49813958 on Build GRCh38
Transcripts
NM_003335.2 XM_005265430.2 XM_006713321.2 XM_011534070.1 XM_011534071.1 XM_011534072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4738598 c.1861G>T p.E621* Chr.3: 49809858-49809858 on Build GRCh38
1231522 c.1722C>A p.F574L Chr.3: 49810095-49810095 on Build GRCh38
4738599 c.1833C>A p.T611T Chr.3: 49809984-49809984 on Build GRCh38
4532442 c.1884G>A p.E628E Chr.3: 49809835-49809835 on Build GRCh38
446727 c.1760C>T p.S587L Chr.3: 49810057-49810057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191072140 Chr.3: 49809967-49809967 on Build GRCh38
rs752904555 Chr.3: 49809993-49809993 on Build GRCh38
rs746423288 Chr.3: 49810202-49810202 on Build GRCh38
rs780279258 Chr.3: 49809798-49809799 on Build GRCh38
rs759841744 Chr.3: 49809981-49809981 on Build GRCh38
rs190943943 Chr.3: 49810196-49810196 on Build GRCh38
rs1040183618 Chr.3: 49809984-49809984 on Build GRCh38
rs781015011 Chr.3: 49810180-49810180 on Build GRCh38
rs371092189 Chr.3: 49809848-49809848 on Build GRCh38
rs761900523 Chr.3: 49810165-49810165 on Build GRCh38
rs769025431 Chr.3: 49809926-49809926 on Build GRCh38
rs752775958 Chr.3: 49809876-49809876 on Build GRCh38
rs757285202 Chr.3: 49809912-49809912 on Build GRCh38
rs775769899 Chr.3: 49810213-49810213 on Build GRCh38
rs764300731 Chr.3: 49810159-49810159 on Build GRCh38
rs771421520 Chr.3: 49810115-49810115 on Build GRCh38
rs768813898 Chr.3: 49810214-49810214 on Build GRCh38
rs777711563 Chr.3: 49809798-49809798 on Build GRCh38
rs757115951 Chr.3: 49810013-49810013 on Build GRCh38
rs370839169 Chr.3: 49809962-49809962 on Build GRCh38
rs746504176 Chr.3: 49809947-49809947 on Build GRCh38
rs201738297 Chr.3: 49810122-49810122 on Build GRCh38
rs749063551 Chr.3: 49809805-49809805 on Build GRCh38
rs200111600 Chr.3: 49809914-49809914 on Build GRCh38
rs766916482 Chr.3: 49809849-49809849 on Build GRCh38
rs200258650 Chr.3: 49809845-49809845 on Build GRCh38
rs374264419 Chr.3: 49809908-49809908 on Build GRCh38
rs766143595 Chr.3: 49810173-49810173 on Build GRCh38
rs559148432 Chr.3: 49809992-49809992 on Build GRCh38
rs370509752 Chr.3: 49810164-49810164 on Build GRCh38
rs751884308 Chr.3: 49809872-49809872 on Build GRCh38
rs925447151 Chr.3: 49809762-49809762 on Build GRCh38
rs777660112 Chr.3: 49809909-49809909 on Build GRCh38
rs957796130 Chr.3: 49810138-49810138 on Build GRCh38
rs371312611 Chr.3: 49809778-49809778 on Build GRCh38
rs772641963 Chr.3: 49810012-49810012 on Build GRCh38
rs754520929 Chr.3: 49810061-49810061 on Build GRCh38
rs374217231 Chr.3: 49809784-49809784 on Build GRCh38
rs746724264 Chr.3: 49810048-49810048 on Build GRCh38
rs1035199837 Chr.3: 49810176-49810176 on Build GRCh38
rs1025059270 Chr.3: 49810034-49810034 on Build GRCh38
rs565989188 Chr.3: 49809831-49809831 on Build GRCh38
rs371659072 Chr.3: 49809956-49809956 on Build GRCh38
rs750365727 Chr.3: 49810169-49810169 on Build GRCh38
rs536133977 Chr.3: 49809820-49809820 on Build GRCh38
rs146190304 Chr.3: 49810123-49810123 on Build GRCh38
rs779592568 Chr.3: 49809938-49809938 on Build GRCh38
rs1001827609 Chr.3: 49810113-49810113 on Build GRCh38
rs747156603 Chr.3: 49809880-49809880 on Build GRCh38
rs759738405 Chr.3: 49809873-49809873 on Build GRCh38
rs143122035 Chr.3: 49810181-49810181 on Build GRCh38
rs759786104 Chr.3: 49810215-49810215 on Build GRCh38
rs763612103 Chr.3: 49809965-49809965 on Build GRCh38
rs778708608 Chr.3: 49809922-49809922 on Build GRCh38
rs760131776 Chr.3: 49809839-49809839 on Build GRCh38
rs574375788 Chr.3: 49810044-49810044 on Build GRCh38
rs774738018 Chr.3: 49809974-49809974 on Build GRCh38
rs761270825 Chr.3: 49810065-49810065 on Build GRCh38
rs182541079 Chr.3: 49809995-49809995 on Build GRCh38
rs2234383 Chr.3: 49809935-49809935 on Build GRCh38
rs756039926 Chr.3: 49809797-49809797 on Build GRCh38
rs932712387 Chr.3: 49809891-49809891 on Build GRCh38
rs1799845 Chr.3: 49810190-49810190 on Build GRCh38
rs756237742 Chr.3: 49809897-49809897 on Build GRCh38
rs758173925 Chr.3: 49810036-49810036 on Build GRCh38
rs754637252 Chr.3: 49810177-49810177 on Build GRCh38
rs755621044 Chr.3: 49810187-49810187 on Build GRCh38
rs758206937 Chr.3: 49810170-49810170 on Build GRCh38
rs770756183 Chr.3: 49809818-49809818 on Build GRCh38
rs111416468 Chr.3: 49809816-49809816 on Build GRCh38
rs367943954 Chr.3: 49810016-49810016 on Build GRCh38
rs142469290 Chr.3: 49810153-49810153 on Build GRCh38
rs747692747 Chr.3: 49810101-49810101 on Build GRCh38
rs775770831 Chr.3: 49810134-49810134 on Build GRCh38
rs199938849 Chr.3: 49810007-49810007 on Build GRCh38
rs772387728 Chr.3: 49810132-49810132 on Build GRCh38
rs532421188 Chr.3: 49809777-49809777 on Build GRCh38
rs771713527 Chr.3: 49809936-49809936 on Build GRCh38
rs149089184 Chr.3: 49809789-49809789 on Build GRCh38
rs141563072 Chr.3: 49809835-49809835 on Build GRCh38
rs1799844 Chr.3: 49810209-49810209 on Build GRCh38
rs186585671 Chr.3: 49810175-49810175 on Build GRCh38
rs750181191 Chr.3: 49810017-49810017 on Build GRCh38
rs988285970 Chr.3: 49809792-49809792 on Build GRCh38
rs554492794 Chr.3: 49809980-49809980 on Build GRCh38

View Full Product Details