Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50297047-50297550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL3
Entrez Gene ID
8372
Gene Name
hyaluronoglucosaminidase 3
Gene Aliases
HYAL-3, LUCA-3, LUCA3
UniGene
Hs.129910
Gene Chromosome Location
Chr.3: 50292828-50299468 on Build GRCh38
Transcripts
NM_001200029.1 NM_001200030.1 NM_001200031.1 NM_001200032.1 NM_003549.3 NM_001200016.1 NM_001200018.1 NM_012191.3

Gene Symbol
NAT6
Entrez Gene ID
24142
Gene Name
N-acetyltransferase 6
Gene Aliases
FUS-2, FUS2
UniGene
Hs.129910
Gene Chromosome Location
Chr.3: 50296402-50299421 on Build GRCh38
Transcripts
NM_001200029.1 NM_001200030.1 NM_001200031.1 NM_001200032.1 NM_003549.3 NM_001200016.1 NM_001200018.1 NM_012191.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864887 c.223C>T p.P75S Chr.3: 50297307-50297307 on Build GRCh38
6389451 c.325C>A p.R109S Chr.3: 50297205-50297205 on Build GRCh38
2851264 c.310C>G p.Q104E Chr.3: 50297220-50297220 on Build GRCh38
2851263 c.320G>A p.R107H Chr.3: 50297210-50297210 on Build GRCh38
6202756 c.39G>T p.L13L Chr.3: 50297491-50297491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781987871 Chr.3: 50297219-50297219 on Build GRCh38
rs782627339 Chr.3: 50297313-50297313 on Build GRCh38
rs371706289 Chr.3: 50297225-50297225 on Build GRCh38
rs372709348 Chr.3: 50297325-50297325 on Build GRCh38
rs376015708 Chr.3: 50297371-50297371 on Build GRCh38
rs781884317 Chr.3: 50297280-50297280 on Build GRCh38
rs587730310 Chr.3: 50297413-50297413 on Build GRCh38
rs782641751 Chr.3: 50297414-50297414 on Build GRCh38
rs781851330 Chr.3: 50297139-50297139 on Build GRCh38
rs782356090 Chr.3: 50297067-50297067 on Build GRCh38
rs587661850 Chr.3: 50297476-50297476 on Build GRCh38
rs782615056 Chr.3: 50297512-50297512 on Build GRCh38
rs587643318 Chr.3: 50297133-50297133 on Build GRCh38
rs782143664 Chr.3: 50297464-50297464 on Build GRCh38
rs782774866 Chr.3: 50297265-50297265 on Build GRCh38
rs782784164 Chr.3: 50297397-50297397 on Build GRCh38
rs369969298 Chr.3: 50297465-50297465 on Build GRCh38
rs1040889161 Chr.3: 50297528-50297528 on Build GRCh38
rs782815866 Chr.3: 50297211-50297211 on Build GRCh38
rs1053028418 Chr.3: 50297481-50297481 on Build GRCh38
rs892400286 Chr.3: 50297483-50297483 on Build GRCh38
rs71326924 Chr.3: 50297198-50297198 on Build GRCh38
rs782453794 Chr.3: 50297400-50297400 on Build GRCh38
rs781953481 Chr.3: 50297107-50297107 on Build GRCh38
rs904485250 Chr.3: 50297179-50297179 on Build GRCh38
rs782062604 Chr.3: 50297366-50297366 on Build GRCh38
rs781944387 Chr.3: 50297347-50297347 on Build GRCh38
rs782264090 Chr.3: 50297485-50297485 on Build GRCh38
rs369059750 Chr.3: 50297383-50297383 on Build GRCh38
rs375660347 Chr.3: 50297438-50297438 on Build GRCh38
rs782695530 Chr.3: 50297449-50297449 on Build GRCh38
rs782407896 Chr.3: 50297255-50297255 on Build GRCh38
rs199554290 Chr.3: 50297137-50297137 on Build GRCh38
rs782290237 Chr.3: 50297189-50297189 on Build GRCh38
rs372606739 Chr.3: 50297398-50297398 on Build GRCh38
rs782571735 Chr.3: 50297430-50297430 on Build GRCh38
rs782328911 Chr.3: 50297428-50297428 on Build GRCh38
rs782052987 Chr.3: 50297104-50297104 on Build GRCh38
rs782545983 Chr.3: 50297292-50297292 on Build GRCh38
rs782316757 Chr.3: 50297346-50297346 on Build GRCh38
rs782004248 Chr.3: 50297121-50297121 on Build GRCh38
rs371728332 Chr.3: 50297358-50297358 on Build GRCh38
rs587697098 Chr.3: 50297368-50297368 on Build GRCh38
rs782215726 Chr.3: 50297204-50297204 on Build GRCh38
rs1019840790 Chr.3: 50297243-50297243 on Build GRCh38
rs782689978 Chr.3: 50297301-50297301 on Build GRCh38
rs374560438 Chr.3: 50297161-50297161 on Build GRCh38
rs782408874 Chr.3: 50297436-50297436 on Build GRCh38
rs782117020 Chr.3: 50297377-50297377 on Build GRCh38
rs782660206 Chr.3: 50297126-50297127 on Build GRCh38
rs782249745 Chr.3: 50297318-50297318 on Build GRCh38
rs35565547 Chr.3: 50297257-50297257 on Build GRCh38
rs782352104 Chr.3: 50297444-50297444 on Build GRCh38
rs782151358 Chr.3: 50297127-50297127 on Build GRCh38
rs200704593 Chr.3: 50297307-50297307 on Build GRCh38
rs782442485 Chr.3: 50297157-50297157 on Build GRCh38
rs782400924 Chr.3: 50297089-50297089 on Build GRCh38
rs782362952 Chr.3: 50297205-50297205 on Build GRCh38
rs782329670 Chr.3: 50297105-50297105 on Build GRCh38
rs782268191 Chr.3: 50297250-50297250 on Build GRCh38
rs782374858 Chr.3: 50297488-50297488 on Build GRCh38
rs368619998 Chr.3: 50297308-50297308 on Build GRCh38
rs781999866 Chr.3: 50297356-50297356 on Build GRCh38
rs369623576 Chr.3: 50297348-50297348 on Build GRCh38
rs781793341 Chr.3: 50297475-50297475 on Build GRCh38
rs377153901 Chr.3: 50297339-50297339 on Build GRCh38
rs781901007 Chr.3: 50297455-50297455 on Build GRCh38
rs782299613 Chr.3: 50297434-50297434 on Build GRCh38
rs782465332 Chr.3: 50297309-50297309 on Build GRCh38
rs77944398 Chr.3: 50297081-50297081 on Build GRCh38
rs782580769 Chr.3: 50297468-50297468 on Build GRCh38
rs782524481 Chr.3: 50297322-50297322 on Build GRCh38
rs782575281 Chr.3: 50297411-50297411 on Build GRCh38
rs201566424 Chr.3: 50297360-50297360 on Build GRCh38
rs35833373 Chr.3: 50297109-50297109 on Build GRCh38
rs587603059 Chr.3: 50297261-50297261 on Build GRCh38
rs782673275 Chr.3: 50297329-50297329 on Build GRCh38
rs782283737 Chr.3: 50297196-50297196 on Build GRCh38
rs374757971 Chr.3: 50297264-50297264 on Build GRCh38
rs781838773 Chr.3: 50297373-50297373 on Build GRCh38
rs781967864 Chr.3: 50297262-50297262 on Build GRCh38
rs587610803 Chr.3: 50297417-50297417 on Build GRCh38
rs782810908 Chr.3: 50297131-50297131 on Build GRCh38
rs587741919 Chr.3: 50297349-50297349 on Build GRCh38
rs372643040 Chr.3: 50297090-50297090 on Build GRCh38
rs200708727 Chr.3: 50297364-50297364 on Build GRCh38
rs782033034 Chr.3: 50297098-50297098 on Build GRCh38
rs782558690 Chr.3: 50297410-50297410 on Build GRCh38
rs782213113 Chr.3: 50297423-50297423 on Build GRCh38
rs781951279 Chr.3: 50297445-50297445 on Build GRCh38
rs782111332 Chr.3: 50297439-50297439 on Build GRCh38
rs782058461 Chr.3: 50297447-50297447 on Build GRCh38
rs782256777 Chr.3: 50297082-50297082 on Build GRCh38
rs377325247 Chr.3: 50297110-50297110 on Build GRCh38

View Full Product Details