Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52258443-52258989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR82
Entrez Gene ID
80335
Gene Name
WD repeat domain 82
Gene Aliases
MST107, MSTP107, PRO2730, PRO34047, SWD2, TMEM113, WDR82A
UniGene
Hs.194110
Gene Chromosome Location
Chr.3: 52254422-52278643 on Build GRCh38
Transcripts
NM_025222.3 XM_011534136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4741097 c.899C>T p.A300V Chr.3: 52258549-52258549 on Build GRCh38
6342015 c.803G>A p.G268E Chr.3: 52258645-52258645 on Build GRCh38
5491654 c.819delA p.V274fs*1 Chr.3: 52258629-52258629 on Build GRCh38
6361672 c.792T>A p.H264Q Chr.3: 52258656-52258656 on Build GRCh38
4118976 c.900G>A p.A300A Chr.3: 52258548-52258548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767459021 Chr.3: 52258723-52258728 on Build GRCh38
rs923997394 Chr.3: 52258862-52258862 on Build GRCh38
rs368349772 Chr.3: 52258623-52258623 on Build GRCh38
rs568305709 Chr.3: 52258848-52258848 on Build GRCh38
rs747540622 Chr.3: 52258681-52258681 on Build GRCh38
rs904647858 Chr.3: 52258941-52258941 on Build GRCh38
rs972482178 Chr.3: 52258777-52258777 on Build GRCh38
rs895150066 Chr.3: 52258922-52258922 on Build GRCh38
rs753326322 Chr.3: 52258657-52258657 on Build GRCh38
rs1019713055 Chr.3: 52258767-52258767 on Build GRCh38
rs766958095 Chr.3: 52258619-52258619 on Build GRCh38
rs752847442 Chr.3: 52258724-52258724 on Build GRCh38
rs758760178 Chr.3: 52258656-52258656 on Build GRCh38
rs768354064 Chr.3: 52258517-52258517 on Build GRCh38
rs377234087 Chr.3: 52258620-52258620 on Build GRCh38
rs562861431 Chr.3: 52258596-52258596 on Build GRCh38
rs772826162 Chr.3: 52258717-52258717 on Build GRCh38
rs952516729 Chr.3: 52258792-52258792 on Build GRCh38
rs763392405 Chr.3: 52258557-52258557 on Build GRCh38
rs1004055174 Chr.3: 52258478-52258478 on Build GRCh38
rs372394905 Chr.3: 52258695-52258695 on Build GRCh38
rs946112786 Chr.3: 52258891-52258891 on Build GRCh38
rs1025438770 Chr.3: 52258786-52258786 on Build GRCh38
rs565522711 Chr.3: 52258829-52258829 on Build GRCh38
rs531883623 Chr.3: 52258638-52258638 on Build GRCh38
rs1041780104 Chr.3: 52258921-52258921 on Build GRCh38
rs752968728 Chr.3: 52258653-52258653 on Build GRCh38
rs765234433 Chr.3: 52258511-52258511 on Build GRCh38
rs566098743 Chr.3: 52258940-52258940 on Build GRCh38
rs762562168 Chr.3: 52258504-52258504 on Build GRCh38
rs150340112 Chr.3: 52258855-52258855 on Build GRCh38
rs187832542 Chr.3: 52258838-52258838 on Build GRCh38
rs774123117 Chr.3: 52258703-52258703 on Build GRCh38
rs760523474 Chr.3: 52258718-52258718 on Build GRCh38
rs779059418 Chr.3: 52258641-52258641 on Build GRCh38
rs771895676 Chr.3: 52258707-52258707 on Build GRCh38
rs917246402 Chr.3: 52258869-52258869 on Build GRCh38
rs141471824 Chr.3: 52258548-52258548 on Build GRCh38
rs983984868 Chr.3: 52258824-52258824 on Build GRCh38
rs771281738 Chr.3: 52258693-52258693 on Build GRCh38
rs545278479 Chr.3: 52258745-52258745 on Build GRCh38
rs1006536684 Chr.3: 52258763-52258763 on Build GRCh38
rs761412808 Chr.3: 52258704-52258704 on Build GRCh38
rs768302801 Chr.3: 52258696-52258696 on Build GRCh38
rs778070473 Chr.3: 52258663-52258663 on Build GRCh38
rs752357593 Chr.3: 52258960-52258960 on Build GRCh38
rs936157071 Chr.3: 52258964-52258964 on Build GRCh38
rs965125842 Chr.3: 52258768-52258768 on Build GRCh38
rs921693245 Chr.3: 52258736-52258736 on Build GRCh38
rs767263019 Chr.3: 52258581-52258581 on Build GRCh38
rs755399428 Chr.3: 52258684-52258684 on Build GRCh38
rs1029190574 Chr.3: 52258734-52258734 on Build GRCh38
rs574115666 Chr.3: 52258487-52258487 on Build GRCh38
rs755082869 Chr.3: 52258639-52258639 on Build GRCh38
rs939463742 Chr.3: 52258909-52258909 on Build GRCh38

View Full Product Details