Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53819444-53819936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHDH
Entrez Gene ID
55349
Gene Name
choline dehydrogenase
Gene Aliases
-
UniGene
Hs.126688
Gene Chromosome Location
Chr.3: 53813130-53846393 on Build GRCh38
Transcripts
NM_018397.4 XM_006713250.3 XM_006713251.3 XM_006713252.3 XM_011533938.2 XM_011533939.2 XM_017006797.1 XM_017006798.1 XM_017006799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5786192 c.1135G>T p.A379S Chr.3: 53819660-53819660 on Build GRCh38
5911853 c.1180C>T p.P394S Chr.3: 53819615-53819615 on Build GRCh38
5937077 c.1263+3G>A p.? Chr.3: 53819529-53819529 on Build GRCh38
7279689 c.1235G>A p.R412Q Chr.3: 53819560-53819560 on Build GRCh38
4402206 c.1127G>A p.G376E Chr.3: 53819668-53819668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978084681 Chr.3: 53819903-53819903 on Build GRCh38
rs763375422 Chr.3: 53819696-53819696 on Build GRCh38
rs539445107 Chr.3: 53819751-53819751 on Build GRCh38
rs989377301 Chr.3: 53819843-53819843 on Build GRCh38
rs764336584 Chr.3: 53819691-53819693 on Build GRCh38
rs756938431 Chr.3: 53819511-53819511 on Build GRCh38
rs760783984 Chr.3: 53819662-53819662 on Build GRCh38
rs908408829 Chr.3: 53819768-53819768 on Build GRCh38
rs759973032 Chr.3: 53819684-53819684 on Build GRCh38
rs769757666 Chr.3: 53819540-53819540 on Build GRCh38
rs974736074 Chr.3: 53819739-53819739 on Build GRCh38
rs964728670 Chr.3: 53819731-53819731 on Build GRCh38
rs866211521 Chr.3: 53819764-53819764 on Build GRCh38
rs948427946 Chr.3: 53819819-53819819 on Build GRCh38
rs1042883787 Chr.3: 53819665-53819665 on Build GRCh38
rs745384215 Chr.3: 53819534-53819534 on Build GRCh38
rs1011704662 Chr.3: 53819887-53819887 on Build GRCh38
rs373914197 Chr.3: 53819510-53819510 on Build GRCh38
rs767168457 Chr.3: 53819703-53819703 on Build GRCh38
rs928271669 Chr.3: 53819834-53819834 on Build GRCh38
rs532459227 Chr.3: 53819841-53819841 on Build GRCh38
rs780690887 Chr.3: 53819516-53819516 on Build GRCh38
rs375682171 Chr.3: 53819713-53819713 on Build GRCh38
rs755373038 Chr.3: 53819492-53819492 on Build GRCh38
rs908357485 Chr.3: 53819523-53819523 on Build GRCh38
rs898197187 Chr.3: 53819568-53819568 on Build GRCh38
rs376011719 Chr.3: 53819670-53819670 on Build GRCh38
rs200289247 Chr.3: 53819677-53819677 on Build GRCh38
rs772329240 Chr.3: 53819651-53819651 on Build GRCh38
rs749195979 Chr.3: 53819613-53819613 on Build GRCh38
rs754354587 Chr.3: 53819589-53819589 on Build GRCh38
rs76334678 Chr.3: 53819867-53819867 on Build GRCh38
rs775819157 Chr.3: 53819695-53819695 on Build GRCh38
rs973867424 Chr.3: 53819790-53819790 on Build GRCh38
rs765751306 Chr.3: 53819691-53819691 on Build GRCh38
rs117099761 Chr.3: 53819763-53819763 on Build GRCh38
rs372747407 Chr.3: 53819572-53819572 on Build GRCh38
rs759269606 Chr.3: 53819684-53819688 on Build GRCh38
rs199841801 Chr.3: 53819593-53819593 on Build GRCh38
rs1055261026 Chr.3: 53819716-53819716 on Build GRCh38
rs1031863034 Chr.3: 53819679-53819679 on Build GRCh38
rs144271109 Chr.3: 53819553-53819553 on Build GRCh38
rs762871358 Chr.3: 53819628-53819646 on Build GRCh38
rs892014437 Chr.3: 53819868-53819868 on Build GRCh38
rs560840677 Chr.3: 53819521-53819521 on Build GRCh38
rs772762599 Chr.3: 53819568-53819569 on Build GRCh38
rs570524358 Chr.3: 53819730-53819730 on Build GRCh38
rs777384961 Chr.3: 53819499-53819499 on Build GRCh38
rs771778664 Chr.3: 53819623-53819623 on Build GRCh38
rs747917464 Chr.3: 53819720-53819720 on Build GRCh38
rs766875892 Chr.3: 53819580-53819580 on Build GRCh38
rs1030716120 Chr.3: 53819754-53819754 on Build GRCh38
rs776807205 Chr.3: 53819672-53819672 on Build GRCh38
rs977028858 Chr.3: 53819814-53819814 on Build GRCh38
rs374272735 Chr.3: 53819601-53819601 on Build GRCh38
rs888097635 Chr.3: 53819757-53819757 on Build GRCh38
rs774317689 Chr.3: 53819561-53819561 on Build GRCh38
rs755620844 Chr.3: 53819610-53819610 on Build GRCh38
rs768521063 Chr.3: 53819560-53819560 on Build GRCh38
rs2289208 Chr.3: 53819715-53819715 on Build GRCh38
rs190376458 Chr.3: 53819717-53819717 on Build GRCh38
rs140429200 Chr.3: 53819829-53819829 on Build GRCh38
rs766306186 Chr.3: 53819632-53819632 on Build GRCh38
rs768719238 Chr.3: 53819618-53819618 on Build GRCh38
rs535371755 Chr.3: 53819832-53819832 on Build GRCh38
rs745670196 Chr.3: 53819487-53819487 on Build GRCh38
rs939808475 Chr.3: 53819535-53819535 on Build GRCh38
rs749080484 Chr.3: 53819558-53819558 on Build GRCh38
rs569334618 Chr.3: 53819709-53819709 on Build GRCh38
rs933875799 Chr.3: 53819512-53819512 on Build GRCh38
rs948374717 Chr.3: 53819621-53819621 on Build GRCh38
rs1043185577 Chr.3: 53819900-53819900 on Build GRCh38
rs202222617 Chr.3: 53819706-53819706 on Build GRCh38
rs939840031 Chr.3: 53819781-53819781 on Build GRCh38
rs368495308 Chr.3: 53819607-53819607 on Build GRCh38
rs142488912 Chr.3: 53819633-53819633 on Build GRCh38
rs370185031 Chr.3: 53819565-53819565 on Build GRCh38
rs375592914 Chr.3: 53819564-53819564 on Build GRCh38
rs773343548 Chr.3: 53819656-53819656 on Build GRCh38
rs754223371 Chr.3: 53819490-53819490 on Build GRCh38
rs921185152 Chr.3: 53819486-53819486 on Build GRCh38
rs775433355 Chr.3: 53819707-53819707 on Build GRCh38
rs779636890 Chr.3: 53819612-53819612 on Build GRCh38
rs751308751 Chr.3: 53819682-53819696 on Build GRCh38
rs749946328 Chr.3: 53819608-53819608 on Build GRCh38
rs919676817 Chr.3: 53819802-53819802 on Build GRCh38
rs765550893 Chr.3: 53819600-53819600 on Build GRCh38

View Full Product Details