Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108981431-108981936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR47
Entrez Gene ID
22911
Gene Name
WD repeat domain 47
Gene Aliases
-
UniGene
Hs.570055
Gene Chromosome Location
Chr.1: 108970211-109042228 on Build GRCh38
Transcripts
NM_001142550.1 NM_001142551.1 NM_014969.5 XM_006710460.3 XM_011541027.2 XM_011541028.2 XM_011541029.1 XM_011541030.1 XM_011541031.1 XM_017000696.1 XM_017000697.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667770 c.2353C>T p.R785* Chr.1: 108981781-108981781 on Build GRCh38
6821893 c.2356_2357delGT p.V786fs*1 Chr.1: 108981798-108981799 on Build GRCh38
6507595 c.2324G>T p.W775L Chr.1: 108981831-108981831 on Build GRCh38
4591240 c.2377G>A p.V793I Chr.1: 108981778-108981778 on Build GRCh38
5377104 c.2321C>T p.S774F Chr.1: 108981813-108981813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762919836 Chr.1: 108981746-108981748 on Build GRCh38
rs776104971 Chr.1: 108981854-108981854 on Build GRCh38
rs756848660 Chr.1: 108981707-108981707 on Build GRCh38
rs1541186 Chr.1: 108981588-108981588 on Build GRCh38
rs745891409 Chr.1: 108981829-108981829 on Build GRCh38
rs1048031047 Chr.1: 108981501-108981501 on Build GRCh38
rs147958674 Chr.1: 108981527-108981527 on Build GRCh38
rs781334141 Chr.1: 108981551-108981551 on Build GRCh38
rs995072704 Chr.1: 108981497-108981497 on Build GRCh38
rs144990719 Chr.1: 108981514-108981514 on Build GRCh38
rs920742035 Chr.1: 108981882-108981882 on Build GRCh38
rs772865475 Chr.1: 108981695-108981696 on Build GRCh38
rs764353095 Chr.1: 108981860-108981860 on Build GRCh38
rs548120030 Chr.1: 108981516-108981516 on Build GRCh38
rs776862464 Chr.1: 108981809-108981809 on Build GRCh38
rs749967209 Chr.1: 108981901-108981901 on Build GRCh38
rs991617778 Chr.1: 108981786-108981786 on Build GRCh38
rs370170653 Chr.1: 108981692-108981692 on Build GRCh38
rs201815469 Chr.1: 108981898-108981898 on Build GRCh38
rs117418268 Chr.1: 108981649-108981649 on Build GRCh38
rs1033942519 Chr.1: 108981639-108981639 on Build GRCh38
rs770451292 Chr.1: 108981853-108981853 on Build GRCh38
rs749879337 Chr.1: 108981712-108981712 on Build GRCh38
rs930837408 Chr.1: 108981890-108981890 on Build GRCh38
rs773443621 Chr.1: 108981664-108981664 on Build GRCh38
rs763424699 Chr.1: 108981856-108981856 on Build GRCh38
rs751149447 Chr.1: 108981702-108981702 on Build GRCh38
rs943711004 Chr.1: 108981601-108981601 on Build GRCh38
rs1048683107 Chr.1: 108981657-108981657 on Build GRCh38
rs367673828 Chr.1: 108981897-108981897 on Build GRCh38
rs778118289 Chr.1: 108981798-108981798 on Build GRCh38
rs774119398 Chr.1: 108981868-108981868 on Build GRCh38
rs747195754 Chr.1: 108981800-108981800 on Build GRCh38
rs899387025 Chr.1: 108981485-108981485 on Build GRCh38
rs369687461 Chr.1: 108981682-108981682 on Build GRCh38
rs77563817 Chr.1: 108981691-108981691 on Build GRCh38
rs113854203 Chr.1: 108981581-108981581 on Build GRCh38
rs35562428 Chr.1: 108981896-108981897 on Build GRCh38
rs779928378 Chr.1: 108981718-108981718 on Build GRCh38
rs755591250 Chr.1: 108981903-108981903 on Build GRCh38
rs914769577 Chr.1: 108981474-108981474 on Build GRCh38
rs373591442 Chr.1: 108981804-108981804 on Build GRCh38
rs754712052 Chr.1: 108981752-108981752 on Build GRCh38
rs1003915054 Chr.1: 108981524-108981524 on Build GRCh38
rs756069953 Chr.1: 108981715-108981715 on Build GRCh38
rs771120896 Chr.1: 108981806-108981806 on Build GRCh38
rs763847020 Chr.1: 108981697-108981697 on Build GRCh38
rs777315821 Chr.1: 108981768-108981768 on Build GRCh38
rs537759483 Chr.1: 108981515-108981515 on Build GRCh38
rs769383570 Chr.1: 108981692-108981693 on Build GRCh38
rs749121980 Chr.1: 108981735-108981735 on Build GRCh38

View Full Product Details