Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109216562-109217108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARS
Entrez Gene ID
6301
Gene Name
seryl-tRNA synthetase
Gene Aliases
SERRS, SERS
UniGene
Hs.531176
Gene Chromosome Location
Chr.1: 109213893-109238182 on Build GRCh38
Transcripts
NM_006513.3 NR_034072.1 XM_006710813.2

SNP Detail

SNP ID Chromosome Location
rs1038941424 Chr.1: 109217009-109217009 on Build GRCh38
rs949694152 Chr.1: 109216992-109216992 on Build GRCh38
rs920133149 Chr.1: 109216647-109216647 on Build GRCh38
rs537658099 Chr.1: 109216830-109216830 on Build GRCh38
rs146629385 Chr.1: 109216795-109216795 on Build GRCh38
rs767306486 Chr.1: 109216707-109216707 on Build GRCh38
rs893586173 Chr.1: 109216591-109216591 on Build GRCh38
rs760555267 Chr.1: 109217057-109217057 on Build GRCh38
rs758547425 Chr.1: 109216901-109216901 on Build GRCh38
rs1010250008 Chr.1: 109216782-109216782 on Build GRCh38
rs140146734 Chr.1: 109216828-109216828 on Build GRCh38
rs974613288 Chr.1: 109216894-109216894 on Build GRCh38
rs750655689 Chr.1: 109216770-109216770 on Build GRCh38
rs550239861 Chr.1: 109217046-109217046 on Build GRCh38
rs373377571 Chr.1: 109216813-109216813 on Build GRCh38
rs554418679 Chr.1: 109216861-109216861 on Build GRCh38
rs201229686 Chr.1: 109216638-109216639 on Build GRCh38
rs766075863 Chr.1: 109216627-109216629 on Build GRCh38
rs1009710372 Chr.1: 109216591-109216591 on Build GRCh38
rs765066144 Chr.1: 109216767-109216767 on Build GRCh38
rs1045497118 Chr.1: 109216712-109216712 on Build GRCh38
rs780316899 Chr.1: 109216979-109216979 on Build GRCh38
rs1009124362 Chr.1: 109216997-109216997 on Build GRCh38
rs546589948 Chr.1: 109216607-109216607 on Build GRCh38
rs890279069 Chr.1: 109216614-109216614 on Build GRCh38
rs757015681 Chr.1: 109216662-109216662 on Build GRCh38
rs930198394 Chr.1: 109216913-109216913 on Build GRCh38
rs116832207 Chr.1: 109217010-109217010 on Build GRCh38
rs186194158 Chr.1: 109216661-109216661 on Build GRCh38
rs1047821571 Chr.1: 109216669-109216669 on Build GRCh38
rs964032696 Chr.1: 109216702-109216703 on Build GRCh38
rs34647086 Chr.1: 109216600-109216601 on Build GRCh38
rs151293360 Chr.1: 109216890-109216890 on Build GRCh38
rs945182099 Chr.1: 109216930-109216930 on Build GRCh38
rs1042499658 Chr.1: 109216972-109216972 on Build GRCh38
rs532248025 Chr.1: 109216659-109216659 on Build GRCh38
rs911413253 Chr.1: 109216590-109216590 on Build GRCh38
rs886209038 Chr.1: 109216676-109216676 on Build GRCh38
rs1020217691 Chr.1: 109216788-109216788 on Build GRCh38
rs577310189 Chr.1: 109217012-109217012 on Build GRCh38
rs1018306490 Chr.1: 109216866-109216866 on Build GRCh38
rs775369475 Chr.1: 109216858-109216858 on Build GRCh38
rs974390693 Chr.1: 109216638-109216638 on Build GRCh38
rs942848656 Chr.1: 109216625-109216625 on Build GRCh38
rs117247296 Chr.1: 109217002-109217002 on Build GRCh38
rs983262826 Chr.1: 109216777-109216777 on Build GRCh38
rs1032929758 Chr.1: 109216766-109216766 on Build GRCh38
rs983014876 Chr.1: 109216957-109216957 on Build GRCh38
rs937635043 Chr.1: 109216590-109216591 on Build GRCh38
rs905730706 Chr.1: 109216720-109216720 on Build GRCh38
rs115619642 Chr.1: 109216733-109216733 on Build GRCh38
rs370071680 Chr.1: 109217047-109217047 on Build GRCh38

View Full Product Details