Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109311410-109311907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SORT1
Entrez Gene ID
6272
Gene Name
sortilin 1
Gene Aliases
Gp95, LDLCQ6, NT3, NTR3
UniGene
Hs.485195
Gene Chromosome Location
Chr.1: 109309565-109397945 on Build GRCh38
Transcripts
NM_001205228.1 NM_002959.6 XM_005271100.2 XM_005271101.2 XM_005271102.1 XM_006710812.1

SNP Detail

SNP ID Chromosome Location
rs961529509 Chr.1: 109311541-109311541 on Build GRCh38
rs934973244 Chr.1: 109311473-109311473 on Build GRCh38
rs558256804 Chr.1: 109311727-109311727 on Build GRCh38
rs1028294235 Chr.1: 109311514-109311514 on Build GRCh38
rs72647807 Chr.1: 109311644-109311644 on Build GRCh38
rs530748978 Chr.1: 109311555-109311555 on Build GRCh38
rs72647805 Chr.1: 109311761-109311761 on Build GRCh38
rs1040824781 Chr.1: 109311638-109311638 on Build GRCh38
rs780153471 Chr.1: 109311447-109311447 on Build GRCh38
rs915562834 Chr.1: 109311579-109311579 on Build GRCh38
rs935004568 Chr.1: 109311705-109311705 on Build GRCh38
rs1064598 Chr.1: 109311461-109311461 on Build GRCh38
rs560334109 Chr.1: 109311490-109311490 on Build GRCh38
rs947167646 Chr.1: 109311589-109311589 on Build GRCh38
rs982400081 Chr.1: 109311545-109311545 on Build GRCh38
rs558167050 Chr.1: 109311642-109311642 on Build GRCh38
rs924374063 Chr.1: 109311865-109311865 on Build GRCh38
rs975706719 Chr.1: 109311723-109311723 on Build GRCh38
rs72647806 Chr.1: 109311657-109311657 on Build GRCh38
rs538062119 Chr.1: 109311828-109311828 on Build GRCh38
rs1050211530 Chr.1: 109311656-109311656 on Build GRCh38
rs569970177 Chr.1: 109311838-109311838 on Build GRCh38
rs1040940935 Chr.1: 109311466-109311466 on Build GRCh38
rs934379949 Chr.1: 109311883-109311883 on Build GRCh38
rs567280929 Chr.1: 109311659-109311659 on Build GRCh38
rs867321080 Chr.1: 109311672-109311672 on Build GRCh38
rs924193102 Chr.1: 109311689-109311689 on Build GRCh38
rs554123171 Chr.1: 109311853-109311853 on Build GRCh38
rs66852370 Chr.1: 109311535-109311535 on Build GRCh38
rs540612634 Chr.1: 109311840-109311840 on Build GRCh38
rs186195609 Chr.1: 109311760-109311760 on Build GRCh38
rs1005672976 Chr.1: 109311709-109311709 on Build GRCh38
rs61696135 Chr.1: 109311554-109311554 on Build GRCh38
rs934499385 Chr.1: 109311617-109311617 on Build GRCh38
rs369570791 Chr.1: 109311602-109311602 on Build GRCh38
rs959914098 Chr.1: 109311766-109311766 on Build GRCh38
rs1005619096 Chr.1: 109311526-109311526 on Build GRCh38
rs567561029 Chr.1: 109311613-109311613 on Build GRCh38
rs941837373 Chr.1: 109311767-109311767 on Build GRCh38
rs554920955 Chr.1: 109311837-109311837 on Build GRCh38
rs752056352 Chr.1: 109311472-109311472 on Build GRCh38
rs767507562 Chr.1: 109311494-109311494 on Build GRCh38
rs190886762 Chr.1: 109311502-109311502 on Build GRCh38
rs540627895 Chr.1: 109311477-109311477 on Build GRCh38
rs1015682931 Chr.1: 109311725-109311725 on Build GRCh38
rs546571692 Chr.1: 109311650-109311650 on Build GRCh38
rs538328945 Chr.1: 109311699-109311699 on Build GRCh38
rs980095279 Chr.1: 109311686-109311686 on Build GRCh38
rs897713854 Chr.1: 109311821-109311821 on Build GRCh38
rs952923957 Chr.1: 109311516-109311516 on Build GRCh38
rs1036709118 Chr.1: 109311795-109311795 on Build GRCh38
rs901571741 Chr.1: 109311522-109311522 on Build GRCh38
rs991581644 Chr.1: 109311578-109311578 on Build GRCh38
rs961415292 Chr.1: 109311742-109311742 on Build GRCh38

View Full Product Details