Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55469952-55470448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT5A
Entrez Gene ID
7474
Gene Name
Wnt family member 5A
Gene Aliases
hWNT5A
UniGene
Hs.643085
Gene Chromosome Location
Chr.3: 55465715-55505261 on Build GRCh38
Transcripts
NM_001256105.1 NM_003392.4 XM_006713324.1 XM_011534085.2 XM_011534086.2 XM_011534087.2 XM_011534088.2 XM_011534089.1 XM_017007127.1 XM_017007128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
242269 c.1121G>A p.R374Q Chr.3: 55470393-55470393 on Build GRCh38
1047418 c.1398C>T p.I466I Chr.3: 55470116-55470116 on Build GRCh38
192113 c.1207G>A p.V403M Chr.3: 55470307-55470307 on Build GRCh38
6823264 c.1335G>A p.T445T Chr.3: 55470179-55470179 on Build GRCh38
420340 c.1372G>C p.V458L Chr.3: 55470142-55470142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754080980 Chr.3: 55470179-55470179 on Build GRCh38
rs768254997 Chr.3: 55470241-55470241 on Build GRCh38
rs762576376 Chr.3: 55470230-55470230 on Build GRCh38
rs755298573 Chr.3: 55470180-55470180 on Build GRCh38
rs763619433 Chr.3: 55470010-55470010 on Build GRCh38
rs747860258 Chr.3: 55470337-55470337 on Build GRCh38
rs747485846 Chr.3: 55470310-55470310 on Build GRCh38
rs773819361 Chr.3: 55470333-55470333 on Build GRCh38
rs751084010 Chr.3: 55470122-55470122 on Build GRCh38
rs546706293 Chr.3: 55470412-55470412 on Build GRCh38
rs1028601599 Chr.3: 55469976-55469976 on Build GRCh38
rs761800959 Chr.3: 55470124-55470124 on Build GRCh38
rs954400880 Chr.3: 55469980-55469980 on Build GRCh38
rs747534689 Chr.3: 55470211-55470211 on Build GRCh38
rs757813200 Chr.3: 55470324-55470324 on Build GRCh38
rs553825428 Chr.3: 55470139-55470139 on Build GRCh38
rs867852190 Chr.3: 55470027-55470027 on Build GRCh38
rs767068436 Chr.3: 55470253-55470253 on Build GRCh38
rs752974227 Chr.3: 55470303-55470303 on Build GRCh38
rs544912525 Chr.3: 55470353-55470353 on Build GRCh38
rs984934479 Chr.3: 55470123-55470123 on Build GRCh38
rs781617053 Chr.3: 55470410-55470410 on Build GRCh38
rs750281702 Chr.3: 55470255-55470255 on Build GRCh38
rs200583587 Chr.3: 55470416-55470416 on Build GRCh38
rs961827183 Chr.3: 55470031-55470031 on Build GRCh38
rs961757574 Chr.3: 55470251-55470251 on Build GRCh38
rs542775163 Chr.3: 55470229-55470229 on Build GRCh38
rs755916316 Chr.3: 55470155-55470155 on Build GRCh38
rs771445451 Chr.3: 55470343-55470343 on Build GRCh38
rs201975128 Chr.3: 55470252-55470252 on Build GRCh38
rs781527130 Chr.3: 55470328-55470328 on Build GRCh38
rs377043203 Chr.3: 55470419-55470419 on Build GRCh38
rs780358735 Chr.3: 55470045-55470045 on Build GRCh38
rs757185242 Chr.3: 55470023-55470023 on Build GRCh38
rs771248572 Chr.3: 55470228-55470228 on Build GRCh38
rs751696945 Chr.3: 55470393-55470393 on Build GRCh38
rs760675240 Chr.3: 55470348-55470348 on Build GRCh38
rs370410624 Chr.3: 55470425-55470425 on Build GRCh38
rs973604044 Chr.3: 55470033-55470033 on Build GRCh38
rs539885600 Chr.3: 55470068-55470068 on Build GRCh38
rs375403509 Chr.3: 55470385-55470385 on Build GRCh38
rs766254001 Chr.3: 55470284-55470284 on Build GRCh38
rs886042717 Chr.3: 55470396-55470396 on Build GRCh38
rs765101926 Chr.3: 55470366-55470366 on Build GRCh38
rs188596917 Chr.3: 55469984-55469984 on Build GRCh38
rs773617692 Chr.3: 55470072-55470072 on Build GRCh38
rs758690185 Chr.3: 55470378-55470378 on Build GRCh38
rs770199302 Chr.3: 55470331-55470331 on Build GRCh38
rs779036055 Chr.3: 55470187-55470187 on Build GRCh38
rs755247138 Chr.3: 55470285-55470285 on Build GRCh38
rs531456992 Chr.3: 55470332-55470332 on Build GRCh38
rs946840827 Chr.3: 55470273-55470273 on Build GRCh38
rs573691468 Chr.3: 55470199-55470199 on Build GRCh38
rs920456630 Chr.3: 55470053-55470053 on Build GRCh38
rs533153165 Chr.3: 55470394-55470394 on Build GRCh38
rs750243237 Chr.3: 55470041-55470041 on Build GRCh38
rs577720282 Chr.3: 55470067-55470067 on Build GRCh38
rs772896188 Chr.3: 55470346-55470346 on Build GRCh38
rs762481849 Chr.3: 55470119-55470119 on Build GRCh38
rs751478038 Chr.3: 55470013-55470013 on Build GRCh38
rs746302333 Chr.3: 55470329-55470329 on Build GRCh38
rs375700896 Chr.3: 55470249-55470249 on Build GRCh38
rs773053795 Chr.3: 55470116-55470116 on Build GRCh38
rs369701725 Chr.3: 55470298-55470298 on Build GRCh38
rs768815254 Chr.3: 55470066-55470066 on Build GRCh38
rs777938109 Chr.3: 55470307-55470307 on Build GRCh38
rs866667950 Chr.3: 55470406-55470406 on Build GRCh38
rs746392030 Chr.3: 55470411-55470411 on Build GRCh38
rs376401083 Chr.3: 55470370-55470371 on Build GRCh38
rs760960120 Chr.3: 55470077-55470077 on Build GRCh38
rs763663982 Chr.3: 55470025-55470025 on Build GRCh38
rs759194223 Chr.3: 55470357-55470357 on Build GRCh38
rs771812000 Chr.3: 55470417-55470417 on Build GRCh38
rs1050351761 Chr.3: 55470311-55470311 on Build GRCh38
rs775072555 Chr.3: 55470107-55470107 on Build GRCh38
rs372423949 Chr.3: 55470188-55470188 on Build GRCh38
rs763566950 Chr.3: 55470112-55470112 on Build GRCh38
rs752820651 Chr.3: 55470371-55470371 on Build GRCh38
rs931674113 Chr.3: 55470291-55470291 on Build GRCh38
rs369954366 Chr.3: 55470405-55470405 on Build GRCh38
rs771295790 Chr.3: 55470104-55470104 on Build GRCh38
rs1044254835 Chr.3: 55470320-55470320 on Build GRCh38
rs373282952 Chr.3: 55470259-55470259 on Build GRCh38
rs900039147 Chr.3: 55470349-55470349 on Build GRCh38
rs62620048 Chr.3: 55470054-55470054 on Build GRCh38
rs770577961 Chr.3: 55470423-55470423 on Build GRCh38
rs756401682 Chr.3: 55470042-55470042 on Build GRCh38
rs767422181 Chr.3: 55470127-55470127 on Build GRCh38
rs776714857 Chr.3: 55470356-55470356 on Build GRCh38
rs922958622 Chr.3: 55470276-55470276 on Build GRCh38
rs780501090 Chr.3: 55470413-55470413 on Build GRCh38
rs780019128 Chr.3: 55470162-55470162 on Build GRCh38
rs758357805 Chr.3: 55470306-55470306 on Build GRCh38

View Full Product Details