Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57560622-57561130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_001322176.1 NM_001322177.1 NM_177966.6

SNP Detail

SNP ID Chromosome Location
rs184376849 Chr.3: 57560924-57560924 on Build GRCh38
rs975290294 Chr.3: 57561087-57561087 on Build GRCh38
rs867369447 Chr.3: 57560963-57560963 on Build GRCh38
rs943753244 Chr.3: 57560846-57560846 on Build GRCh38
rs377626435 Chr.3: 57560955-57560955 on Build GRCh38
rs561034467 Chr.3: 57560784-57560784 on Build GRCh38
rs942585039 Chr.3: 57561042-57561042 on Build GRCh38
rs571858051 Chr.3: 57561103-57561103 on Build GRCh38
rs748894438 Chr.3: 57560660-57560660 on Build GRCh38
rs1042137277 Chr.3: 57560920-57560920 on Build GRCh38
rs939823627 Chr.3: 57560692-57560692 on Build GRCh38
rs970469575 Chr.3: 57560785-57560785 on Build GRCh38
rs572614602 Chr.3: 57561004-57561004 on Build GRCh38
rs902274316 Chr.3: 57560923-57560923 on Build GRCh38
rs932626293 Chr.3: 57560810-57560810 on Build GRCh38
rs983432684 Chr.3: 57561010-57561010 on Build GRCh38
rs549824463 Chr.3: 57560830-57560830 on Build GRCh38
rs1032515230 Chr.3: 57560931-57560931 on Build GRCh38
rs748024587 Chr.3: 57560964-57560964 on Build GRCh38
rs375531685 Chr.3: 57561035-57561035 on Build GRCh38
rs146506134 Chr.3: 57560668-57560668 on Build GRCh38
rs970726623 Chr.3: 57560981-57560981 on Build GRCh38
rs35237962 Chr.3: 57560978-57560978 on Build GRCh38
rs543059604 Chr.3: 57560738-57560738 on Build GRCh38
rs974273214 Chr.3: 57561101-57561101 on Build GRCh38
rs895383619 Chr.3: 57560954-57560954 on Build GRCh38
rs139987987 Chr.3: 57560820-57560820 on Build GRCh38
rs541272922 Chr.3: 57560716-57560716 on Build GRCh38
rs963501216 Chr.3: 57561054-57561054 on Build GRCh38
rs890826224 Chr.3: 57560825-57560825 on Build GRCh38

View Full Product Details