Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58863420-58863916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533460.2 XM_011533461.1 XM_011533463.2 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235776 c.1012C>T p.H338Y Chr.3: 58863764-58863764 on Build GRCh38
4941058 c.1075C>T p.P359S Chr.3: 58863701-58863701 on Build GRCh38
731782 c.1038G>A p.Q346Q Chr.3: 58863738-58863738 on Build GRCh38
6235777 c.1012C>T p.H338Y Chr.3: 58863764-58863764 on Build GRCh38
6214631 c.1199G>T p.G400V Chr.3: 58863577-58863577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530086324 Chr.3: 58863575-58863575 on Build GRCh38
rs760252763 Chr.3: 58863873-58863873 on Build GRCh38
rs552749948 Chr.3: 58863676-58863676 on Build GRCh38
rs200042040 Chr.3: 58863668-58863668 on Build GRCh38
rs748441249 Chr.3: 58863507-58863523 on Build GRCh38
rs947609654 Chr.3: 58863471-58863471 on Build GRCh38
rs201334935 Chr.3: 58863724-58863724 on Build GRCh38
rs946391347 Chr.3: 58863501-58863501 on Build GRCh38
rs768892837 Chr.3: 58863523-58863523 on Build GRCh38
rs757504887 Chr.3: 58863467-58863467 on Build GRCh38
rs1003209827 Chr.3: 58863564-58863564 on Build GRCh38
rs571045260 Chr.3: 58863697-58863697 on Build GRCh38
rs769497519 Chr.3: 58863685-58863685 on Build GRCh38
rs751275835 Chr.3: 58863704-58863704 on Build GRCh38
rs866283009 Chr.3: 58863599-58863599 on Build GRCh38
rs749140287 Chr.3: 58863810-58863810 on Build GRCh38
rs773838863 Chr.3: 58863735-58863735 on Build GRCh38
rs772312909 Chr.3: 58863660-58863660 on Build GRCh38
rs772866007 Chr.3: 58863866-58863866 on Build GRCh38
rs369398734 Chr.3: 58863578-58863578 on Build GRCh38
rs758435990 Chr.3: 58863779-58863779 on Build GRCh38
rs747208104 Chr.3: 58863786-58863786 on Build GRCh38
rs765890334 Chr.3: 58863771-58863771 on Build GRCh38
rs923244041 Chr.3: 58863693-58863693 on Build GRCh38
rs780475842 Chr.3: 58863497-58863497 on Build GRCh38
rs777948716 Chr.3: 58863718-58863719 on Build GRCh38
rs760806462 Chr.3: 58863594-58863597 on Build GRCh38
rs779142524 Chr.3: 58863626-58863626 on Build GRCh38
rs756476313 Chr.3: 58863613-58863613 on Build GRCh38
rs368551078 Chr.3: 58863552-58863552 on Build GRCh38
rs781495495 Chr.3: 58863461-58863461 on Build GRCh38
rs774197666 Chr.3: 58863855-58863855 on Build GRCh38
rs200717739 Chr.3: 58863572-58863572 on Build GRCh38
rs768994756 Chr.3: 58863764-58863764 on Build GRCh38
rs927476169 Chr.3: 58863814-58863814 on Build GRCh38
rs34322986 Chr.3: 58863565-58863565 on Build GRCh38
rs748398313 Chr.3: 58863541-58863541 on Build GRCh38
rs761280367 Chr.3: 58863746-58863746 on Build GRCh38
rs986873649 Chr.3: 58863734-58863734 on Build GRCh38
rs779230696 Chr.3: 58863529-58863529 on Build GRCh38
rs538087182 Chr.3: 58863698-58863698 on Build GRCh38
rs144152554 Chr.3: 58863632-58863632 on Build GRCh38
rs749351790 Chr.3: 58863508-58863508 on Build GRCh38
rs750077560 Chr.3: 58863761-58863761 on Build GRCh38
rs893494153 Chr.3: 58863472-58863472 on Build GRCh38
rs768648328 Chr.3: 58863707-58863707 on Build GRCh38
rs148755922 Chr.3: 58863576-58863576 on Build GRCh38
rs779820032 Chr.3: 58863796-58863796 on Build GRCh38
rs149551458 Chr.3: 58863569-58863569 on Build GRCh38
rs532254885 Chr.3: 58863473-58863473 on Build GRCh38
rs148860143 Chr.3: 58863649-58863649 on Build GRCh38
rs568680728 Chr.3: 58863768-58863768 on Build GRCh38
rs112633530 Chr.3: 58863748-58863748 on Build GRCh38
rs536210537 Chr.3: 58863783-58863783 on Build GRCh38
rs750778562 Chr.3: 58863580-58863580 on Build GRCh38
rs780671409 Chr.3: 58863614-58863614 on Build GRCh38
rs35778488 Chr.3: 58863864-58863864 on Build GRCh38
rs747656672 Chr.3: 58863861-58863861 on Build GRCh38
rs34631714 Chr.3: 58863617-58863617 on Build GRCh38
rs752788408 Chr.3: 58863773-58863773 on Build GRCh38
rs527798921 Chr.3: 58863672-58863672 on Build GRCh38
rs780512640 Chr.3: 58863824-58863824 on Build GRCh38
rs1016758884 Chr.3: 58863663-58863663 on Build GRCh38
rs755524466 Chr.3: 58863769-58863769 on Build GRCh38
rs112867020 Chr.3: 58863888-58863888 on Build GRCh38
rs1042024162 Chr.3: 58863502-58863502 on Build GRCh38
rs757386555 Chr.3: 58863762-58863762 on Build GRCh38
rs902277379 Chr.3: 58863563-58863563 on Build GRCh38
rs766205138 Chr.3: 58863885-58863885 on Build GRCh38
rs373396484 Chr.3: 58863625-58863625 on Build GRCh38
rs147935364 Chr.3: 58863753-58863753 on Build GRCh38
rs751232552 Chr.3: 58863458-58863458 on Build GRCh38
rs141700320 Chr.3: 58863843-58863843 on Build GRCh38
rs140879098 Chr.3: 58863638-58863638 on Build GRCh38
rs756631128 Chr.3: 58863489-58863489 on Build GRCh38
rs767136021 Chr.3: 58863579-58863579 on Build GRCh38
rs755840434 Chr.3: 58863789-58863789 on Build GRCh38
rs375445285 Chr.3: 58863546-58863546 on Build GRCh38

View Full Product Details