Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113947770-113948263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC23
Entrez Gene ID
254887
Gene Name
zinc finger DHHC-type containing 23
Gene Aliases
DHHC-23, NIDD
UniGene
Hs.21902
Gene Chromosome Location
Chr.3: 113947901-113998002 on Build GRCh38
Transcripts
NM_001320466.1 NM_001320467.1 NM_001320468.1 NM_173570.4 NR_135271.1 XM_005247269.1 XM_006713562.3 XM_011512618.2 XM_011512619.1 XM_017006084.1 XM_017006085.1 XM_017006086.1 XM_017006087.1

SNP Detail

SNP ID Chromosome Location
rs554604617 Chr.3: 113948135-113948135 on Build GRCh38
rs530374448 Chr.3: 113948178-113948178 on Build GRCh38
rs1048584430 Chr.3: 113947928-113947928 on Build GRCh38
rs574938343 Chr.3: 113948061-113948061 on Build GRCh38
rs544128864 Chr.3: 113948179-113948179 on Build GRCh38
rs553003958 Chr.3: 113947979-113947979 on Build GRCh38
rs1039772269 Chr.3: 113947947-113947947 on Build GRCh38
rs1052471185 Chr.3: 113948186-113948186 on Build GRCh38
rs947482995 Chr.3: 113948221-113948221 on Build GRCh38
rs956722256 Chr.3: 113947826-113947826 on Build GRCh38
rs1006964548 Chr.3: 113947936-113947936 on Build GRCh38
rs918863864 Chr.3: 113947914-113947914 on Build GRCh38
rs192819538 Chr.3: 113947809-113947809 on Build GRCh38
rs956679853 Chr.3: 113948011-113948011 on Build GRCh38
rs939004314 Chr.3: 113947901-113947901 on Build GRCh38
rs980481776 Chr.3: 113948069-113948069 on Build GRCh38
rs969116747 Chr.3: 113948041-113948041 on Build GRCh38
rs988957111 Chr.3: 113948208-113948208 on Build GRCh38
rs998582896 Chr.3: 113948156-113948156 on Build GRCh38
rs1022178234 Chr.3: 113948035-113948035 on Build GRCh38
rs532663605 Chr.3: 113948213-113948213 on Build GRCh38
rs956409307 Chr.3: 113948197-113948197 on Build GRCh38
rs752651082 Chr.3: 113948124-113948124 on Build GRCh38
rs571010922 Chr.3: 113948009-113948009 on Build GRCh38
rs888512090 Chr.3: 113947934-113947934 on Build GRCh38
rs947811293 Chr.3: 113947866-113947866 on Build GRCh38
rs556527853 Chr.3: 113948036-113948036 on Build GRCh38
rs930106892 Chr.3: 113947922-113947922 on Build GRCh38
rs901143072 Chr.3: 113947984-113947984 on Build GRCh38
rs879397255 Chr.3: 113948207-113948207 on Build GRCh38
rs930265900 Chr.3: 113948096-113948096 on Build GRCh38
rs965258752 Chr.3: 113947793-113947793 on Build GRCh38
rs116819004 Chr.3: 113948079-113948079 on Build GRCh38
rs960336521 Chr.3: 113948075-113948075 on Build GRCh38
rs1055808833 Chr.3: 113947883-113947883 on Build GRCh38
rs1022227397 Chr.3: 113948211-113948211 on Build GRCh38
rs534144547 Chr.3: 113947894-113947894 on Build GRCh38
rs918820751 Chr.3: 113948081-113948081 on Build GRCh38
rs867410970 Chr.3: 113947895-113947895 on Build GRCh38
rs1057494367 Chr.3: 113947902-113947902 on Build GRCh38
rs572961679 Chr.3: 113948139-113948139 on Build GRCh38
rs970418298 Chr.3: 113948146-113948146 on Build GRCh38
rs1049036408 Chr.3: 113948118-113948118 on Build GRCh38
rs544759064 Chr.3: 113948214-113948214 on Build GRCh38
rs538204217 Chr.3: 113948031-113948031 on Build GRCh38
rs895385940 Chr.3: 113947992-113947992 on Build GRCh38
rs540077519 Chr.3: 113948177-113948177 on Build GRCh38
rs927699058 Chr.3: 113948073-113948073 on Build GRCh38
rs976606380 Chr.3: 113947798-113947798 on Build GRCh38
rs998215795 Chr.3: 113948007-113948007 on Build GRCh38
rs34810933 Chr.3: 113947963-113947963 on Build GRCh38
rs982136106 Chr.3: 113948154-113948154 on Build GRCh38
rs989345092 Chr.3: 113947853-113947856 on Build GRCh38
rs914963973 Chr.3: 113947857-113947857 on Build GRCh38

View Full Product Details