Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119780403-119780913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR1I2
Entrez Gene ID
8856
Gene Name
nuclear receptor subfamily 1 group I member 2
Gene Aliases
BXR, ONR1, PAR, PAR1, PAR2, PARq, PRR, PXR, SAR, SXR
UniGene
Hs.7303
Gene Chromosome Location
Chr.3: 119780484-119818485 on Build GRCh38
Transcripts
NM_003889.3 NM_033013.2

SNP Detail

SNP ID Chromosome Location
rs989144563 Chr.3: 119780516-119780516 on Build GRCh38
rs181298440 Chr.3: 119780808-119780808 on Build GRCh38
rs1022674448 Chr.3: 119780591-119780591 on Build GRCh38
rs547253474 Chr.3: 119780746-119780746 on Build GRCh38
rs1523129 Chr.3: 119780761-119780761 on Build GRCh38
rs987542547 Chr.3: 119780673-119780673 on Build GRCh38
rs903811476 Chr.3: 119780823-119780823 on Build GRCh38
rs576423942 Chr.3: 119780628-119780628 on Build GRCh38
rs910618646 Chr.3: 119780676-119780676 on Build GRCh38
rs551499228 Chr.3: 119780782-119780782 on Build GRCh38
rs754154395 Chr.3: 119780558-119780558 on Build GRCh38
rs1000916793 Chr.3: 119780527-119780527 on Build GRCh38
rs564421096 Chr.3: 119780507-119780507 on Build GRCh38
rs188286603 Chr.3: 119780504-119780504 on Build GRCh38
rs777762950 Chr.3: 119780874-119780874 on Build GRCh38
rs959329597 Chr.3: 119780481-119780481 on Build GRCh38
rs568716234 Chr.3: 119780721-119780721 on Build GRCh38
rs55904282 Chr.3: 119780476-119780476 on Build GRCh38
rs930989070 Chr.3: 119780679-119780679 on Build GRCh38
rs943825861 Chr.3: 119780751-119780751 on Build GRCh38
rs991982977 Chr.3: 119780615-119780615 on Build GRCh38
rs549023656 Chr.3: 119780858-119780858 on Build GRCh38
rs766902650 Chr.3: 119780479-119780479 on Build GRCh38
rs956812486 Chr.3: 119780597-119780597 on Build GRCh38
rs902026271 Chr.3: 119780770-119780770 on Build GRCh38
rs114087724 Chr.3: 119780847-119780847 on Build GRCh38
rs932223963 Chr.3: 119780786-119780786 on Build GRCh38
rs538761140 Chr.3: 119780525-119780525 on Build GRCh38
rs908299301 Chr.3: 119780725-119780725 on Build GRCh38
rs185559474 Chr.3: 119780878-119780878 on Build GRCh38
rs977895863 Chr.3: 119780622-119780622 on Build GRCh38
rs892539057 Chr.3: 119780855-119780855 on Build GRCh38
rs1046759915 Chr.3: 119780686-119780686 on Build GRCh38
rs1026451540 Chr.3: 119780451-119780451 on Build GRCh38
rs1003702556 Chr.3: 119780831-119780831 on Build GRCh38
rs576073660 Chr.3: 119780513-119780513 on Build GRCh38
rs561700370 Chr.3: 119780678-119780678 on Build GRCh38
rs1523130 Chr.3: 119780660-119780660 on Build GRCh38
rs917770123 Chr.3: 119780616-119780616 on Build GRCh38
rs559532610 Chr.3: 119780759-119780759 on Build GRCh38
rs868684404 Chr.3: 119780692-119780692 on Build GRCh38
rs970469432 Chr.3: 119780455-119780455 on Build GRCh38
rs966647324 Chr.3: 119780614-119780614 on Build GRCh38
rs1000862909 Chr.3: 119780845-119780845 on Build GRCh38

View Full Product Details