Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120781621-120782157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2E1
Entrez Gene ID
2960
Gene Name
general transcription factor IIE subunit 1
Gene Aliases
FE, TF2E1, TFIIE-A
UniGene
Hs.445272
Gene Chromosome Location
Chr.3: 120742711-120783134 on Build GRCh38
Transcripts
NM_005513.2 XM_011512744.2 XM_011512745.2

SNP Detail

SNP ID Chromosome Location
rs766918508 Chr.3: 120782073-120782073 on Build GRCh38
rs1055068819 Chr.3: 120781949-120781949 on Build GRCh38
rs975300506 Chr.3: 120782106-120782106 on Build GRCh38
rs976358433 Chr.3: 120781650-120781650 on Build GRCh38
rs781528821 Chr.3: 120781997-120781998 on Build GRCh38
rs921211650 Chr.3: 120782134-120782134 on Build GRCh38
rs1018145699 Chr.3: 120781972-120781972 on Build GRCh38
rs763397203 Chr.3: 120781962-120781962 on Build GRCh38
rs138479447 Chr.3: 120781669-120781669 on Build GRCh38
rs530282988 Chr.3: 120781961-120781961 on Build GRCh38
rs547312178 Chr.3: 120781747-120781747 on Build GRCh38
rs553263765 Chr.3: 120782104-120782104 on Build GRCh38
rs189007860 Chr.3: 120781818-120781818 on Build GRCh38
rs1032126002 Chr.3: 120781766-120781766 on Build GRCh38
rs573051563 Chr.3: 120782113-120782113 on Build GRCh38
rs966228802 Chr.3: 120781975-120781975 on Build GRCh38
rs934842537 Chr.3: 120781935-120781935 on Build GRCh38
rs1017139424 Chr.3: 120782075-120782075 on Build GRCh38
rs919536186 Chr.3: 120781667-120781667 on Build GRCh38
rs1045645239 Chr.3: 120781825-120781825 on Build GRCh38
rs536606765 Chr.3: 120781965-120781965 on Build GRCh38
rs34631097 Chr.3: 120781908-120781909 on Build GRCh38
rs772192288 Chr.3: 120781706-120781710 on Build GRCh38
rs983058434 Chr.3: 120781819-120781819 on Build GRCh38
rs571024854 Chr.3: 120781909-120781909 on Build GRCh38
rs903138618 Chr.3: 120781837-120781837 on Build GRCh38
rs902419667 Chr.3: 120781732-120781732 on Build GRCh38
rs183889681 Chr.3: 120781784-120781784 on Build GRCh38
rs962585701 Chr.3: 120781835-120781835 on Build GRCh38
rs765890752 Chr.3: 120781986-120781987 on Build GRCh38
rs35166300 Chr.3: 120781662-120781662 on Build GRCh38
rs904854273 Chr.3: 120781996-120781996 on Build GRCh38
rs77067438 Chr.3: 120781657-120781657 on Build GRCh38
rs35637163 Chr.3: 120781704-120781705 on Build GRCh38
rs796277562 Chr.3: 120781715-120781715 on Build GRCh38
rs909793082 Chr.3: 120781820-120781820 on Build GRCh38
rs924713580 Chr.3: 120781916-120781916 on Build GRCh38
rs113990838 Chr.3: 120781737-120781737 on Build GRCh38
rs76802413 Chr.3: 120781731-120781731 on Build GRCh38
rs1051175930 Chr.3: 120781912-120781912 on Build GRCh38
rs1003183834 Chr.3: 120782024-120782024 on Build GRCh38
rs975329896 Chr.3: 120781863-120781863 on Build GRCh38
rs868535223 Chr.3: 120782057-120782057 on Build GRCh38
rs34391857 Chr.3: 120781760-120781760 on Build GRCh38
rs61796377 Chr.3: 120781733-120781733 on Build GRCh38
rs773889020 Chr.3: 120781715-120781716 on Build GRCh38
rs199752364 Chr.3: 120781862-120781862 on Build GRCh38
rs1007642860 Chr.3: 120781966-120781966 on Build GRCh38
rs71133517 Chr.3: 120781731-120781731 on Build GRCh38
rs1044639349 Chr.3: 120781983-120781983 on Build GRCh38
rs77037565 Chr.3: 120782098-120782098 on Build GRCh38
rs201377087 Chr.3: 120781801-120781801 on Build GRCh38
rs561583769 Chr.3: 120781719-120781719 on Build GRCh38
rs567300777 Chr.3: 120782069-120782069 on Build GRCh38
rs545243188 Chr.3: 120782120-120782120 on Build GRCh38
rs933994374 Chr.3: 120781877-120781877 on Build GRCh38
rs55697401 Chr.3: 120781872-120781872 on Build GRCh38
rs796154143 Chr.3: 120781798-120781798 on Build GRCh38
rs34267980 Chr.3: 120781671-120781671 on Build GRCh38
rs1003216486 Chr.3: 120781785-120781785 on Build GRCh38
rs531813099 Chr.3: 120781960-120781960 on Build GRCh38
rs532597703 Chr.3: 120781790-120781790 on Build GRCh38
rs982468952 Chr.3: 120781720-120781720 on Build GRCh38
rs547521865 Chr.3: 120781752-120781752 on Build GRCh38
rs556685890 Chr.3: 120781968-120781968 on Build GRCh38

View Full Product Details