Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122535439-122535951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP9
Entrez Gene ID
83666
Gene Name
poly(ADP-ribose) polymerase family member 9
Gene Aliases
ARTD9, BAL, BAL1, MGC:7868
UniGene
Hs.518200
Gene Chromosome Location
Chr.3: 122527910-122564676 on Build GRCh38
Transcripts
NM_001146102.1 NM_001146103.1 NM_001146104.1 NM_001146105.1 NM_001146106.1 NM_031458.2 XM_005247820.2 XM_011513218.1 XM_017007304.1 XM_017007305.1 XM_017007306.1

SNP Detail

SNP ID Chromosome Location
rs1053385259 Chr.3: 122535659-122535659 on Build GRCh38
rs564887117 Chr.3: 122535801-122535801 on Build GRCh38
rs560631214 Chr.3: 122535792-122535792 on Build GRCh38
rs933465594 Chr.3: 122535572-122535572 on Build GRCh38
rs1011984076 Chr.3: 122535698-122535698 on Build GRCh38
rs995247752 Chr.3: 122535834-122535834 on Build GRCh38
rs63229263 Chr.3: 122535846-122535846 on Build GRCh38
rs1025449046 Chr.3: 122535876-122535876 on Build GRCh38
rs1038299523 Chr.3: 122535487-122535487 on Build GRCh38
rs11336304 Chr.3: 122535860-122535860 on Build GRCh38
rs1001090535 Chr.3: 122535638-122535638 on Build GRCh38
rs989310517 Chr.3: 122535661-122535661 on Build GRCh38
rs1018553318 Chr.3: 122535685-122535685 on Build GRCh38
rs958045921 Chr.3: 122535587-122535587 on Build GRCh38
rs34914628 Chr.3: 122535676-122535677 on Build GRCh38
rs894809483 Chr.3: 122535668-122535668 on Build GRCh38
rs757629314 Chr.3: 122535537-122535537 on Build GRCh38
rs1012947348 Chr.3: 122535528-122535528 on Build GRCh38
rs746719817 Chr.3: 122535846-122535846 on Build GRCh38
rs142286684 Chr.3: 122535871-122535871 on Build GRCh38
rs768775975 Chr.3: 122535844-122535846 on Build GRCh38
rs184977450 Chr.3: 122535914-122535914 on Build GRCh38
rs60218019 Chr.3: 122535860-122535861 on Build GRCh38
rs754275737 Chr.3: 122535520-122535520 on Build GRCh38
rs879251104 Chr.3: 122535844-122535845 on Build GRCh38
rs16833168 Chr.3: 122535800-122535800 on Build GRCh38
rs1037866050 Chr.3: 122535921-122535921 on Build GRCh38
rs907939228 Chr.3: 122535879-122535879 on Build GRCh38
rs902776875 Chr.3: 122535598-122535598 on Build GRCh38
rs942136811 Chr.3: 122535880-122535880 on Build GRCh38
rs560515191 Chr.3: 122535530-122535530 on Build GRCh38
rs79170687 Chr.3: 122535859-122535859 on Build GRCh38
rs895384260 Chr.3: 122535494-122535494 on Build GRCh38
rs977115825 Chr.3: 122535702-122535702 on Build GRCh38
rs952748898 Chr.3: 122535913-122535913 on Build GRCh38
rs10716375 Chr.3: 122535844-122535844 on Build GRCh38
rs1024665464 Chr.3: 122535721-122535721 on Build GRCh38
rs1030680797 Chr.3: 122535585-122535585 on Build GRCh38
rs776222954 Chr.3: 122535845-122535846 on Build GRCh38
rs147971387 Chr.3: 122535843-122535843 on Build GRCh38
rs922516927 Chr.3: 122535718-122535718 on Build GRCh38
rs970611634 Chr.3: 122535868-122535868 on Build GRCh38
rs869042978 Chr.3: 122535859-122535860 on Build GRCh38
rs3817040 Chr.3: 122535922-122535922 on Build GRCh38
rs543939905 Chr.3: 122535480-122535480 on Build GRCh38
rs868784317 Chr.3: 122535810-122535810 on Build GRCh38
rs1024593385 Chr.3: 122535564-122535564 on Build GRCh38
rs76774523 Chr.3: 122535835-122535835 on Build GRCh38
rs546044555 Chr.3: 122535845-122535845 on Build GRCh38
rs1042535807 Chr.3: 122535574-122535574 on Build GRCh38
rs180794292 Chr.3: 122535567-122535567 on Build GRCh38
rs754622586 Chr.3: 122535776-122535776 on Build GRCh38
rs57077282 Chr.3: 122535570-122535570 on Build GRCh38

View Full Product Details