Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123282608-123283112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY5
Entrez Gene ID
111
Gene Name
adenylate cyclase 5
Gene Aliases
AC5, FDFM
UniGene
Hs.593292
Gene Chromosome Location
Chr.3: 123282296-123448632 on Build GRCh38
Transcripts
NM_001199642.1 NM_183357.2 XM_005247077.3 XM_005247078.1 XM_006713483.1 XM_006713484.1 XM_011512359.2 XM_011512360.2 XM_011512361.1 XM_017005638.1 XM_017005639.1 XM_017005640.1

SNP Detail

SNP ID Chromosome Location
rs537167153 Chr.3: 123282823-123282823 on Build GRCh38
rs187024714 Chr.3: 123282930-123282930 on Build GRCh38
rs6784930 Chr.3: 123282647-123282647 on Build GRCh38
rs977751487 Chr.3: 123282866-123282866 on Build GRCh38
rs560626648 Chr.3: 123283023-123283023 on Build GRCh38
rs184350309 Chr.3: 123282981-123282981 on Build GRCh38
rs920970378 Chr.3: 123282675-123282675 on Build GRCh38
rs145986442 Chr.3: 123282646-123282646 on Build GRCh38
rs547887822 Chr.3: 123282650-123282650 on Build GRCh38
rs971065892 Chr.3: 123282858-123282858 on Build GRCh38
rs895250118 Chr.3: 123283042-123283046 on Build GRCh38
rs933618215 Chr.3: 123282703-123282703 on Build GRCh38
rs934388720 Chr.3: 123282737-123282737 on Build GRCh38
rs1048197703 Chr.3: 123282735-123282735 on Build GRCh38
rs1037814668 Chr.3: 123283041-123283041 on Build GRCh38
rs532460792 Chr.3: 123283029-123283029 on Build GRCh38
rs1050851160 Chr.3: 123282718-123282718 on Build GRCh38
rs997496117 Chr.3: 123282732-123282732 on Build GRCh38
rs375691439 Chr.3: 123282733-123282733 on Build GRCh38
rs975372028 Chr.3: 123282976-123282976 on Build GRCh38
rs989225784 Chr.3: 123282931-123282931 on Build GRCh38
rs1045695540 Chr.3: 123282758-123282758 on Build GRCh38
rs934419894 Chr.3: 123283063-123283063 on Build GRCh38
rs943617784 Chr.3: 123282653-123282653 on Build GRCh38
rs573748198 Chr.3: 123282721-123282721 on Build GRCh38
rs923511723 Chr.3: 123282964-123282964 on Build GRCh38
rs759003230 Chr.3: 123282642-123282642 on Build GRCh38
rs924269601 Chr.3: 123283027-123283027 on Build GRCh38
rs962698321 Chr.3: 123282648-123282648 on Build GRCh38
rs1047720213 Chr.3: 123283005-123283006 on Build GRCh38
rs745368941 Chr.3: 123282709-123282709 on Build GRCh38
rs545028884 Chr.3: 123282808-123282808 on Build GRCh38
rs544883593 Chr.3: 123282895-123282895 on Build GRCh38
rs955029315 Chr.3: 123283002-123283002 on Build GRCh38
rs990290222 Chr.3: 123283068-123283068 on Build GRCh38
rs549652838 Chr.3: 123282649-123282649 on Build GRCh38
rs757538537 Chr.3: 123282778-123282778 on Build GRCh38
rs1039188962 Chr.3: 123282963-123282963 on Build GRCh38
rs951218910 Chr.3: 123282635-123282635 on Build GRCh38
rs531251819 Chr.3: 123282839-123282839 on Build GRCh38
rs866433407 Chr.3: 123282667-123282667 on Build GRCh38
rs184146224 Chr.3: 123282928-123282928 on Build GRCh38
rs942079116 Chr.3: 123283029-123283030 on Build GRCh38
rs1039283692 Chr.3: 123282696-123282696 on Build GRCh38
rs568114967 Chr.3: 123282767-123282767 on Build GRCh38
rs771512527 Chr.3: 123282754-123282754 on Build GRCh38
rs933568706 Chr.3: 123282969-123282969 on Build GRCh38
rs553014278 Chr.3: 123282746-123282746 on Build GRCh38
rs192717414 Chr.3: 123282932-123282932 on Build GRCh38
rs1004253139 Chr.3: 123282863-123282863 on Build GRCh38
rs902109641 Chr.3: 123282711-123282711 on Build GRCh38
rs149655052 Chr.3: 123282884-123282885 on Build GRCh38
rs914731317 Chr.3: 123282960-123282960 on Build GRCh38
rs139590302 Chr.3: 123282755-123282755 on Build GRCh38
rs887228038 Chr.3: 123282846-123282846 on Build GRCh38
rs557671604 Chr.3: 123282634-123282634 on Build GRCh38
rs1016792331 Chr.3: 123282914-123282914 on Build GRCh38
rs923607569 Chr.3: 123282877-123282877 on Build GRCh38

View Full Product Details