Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123969928-123970454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROPN1
Entrez Gene ID
54763
Gene Name
rhophilin associated tail protein 1
Gene Aliases
CT91, ODF6, RHPNAP1, ROPN1A, ropporin
UniGene
Hs.567516
Gene Chromosome Location
Chr.3: 123969006-123992170 on Build GRCh38
Transcripts
NM_001317774.1 NM_001317775.1 NM_017578.3 XM_011512933.2 XM_011512934.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4496184 c.469C>T p.P157S Chr.3: 123970145-123970145 on Build GRCh38
5774339 c.462C>G p.P154P Chr.3: 123970152-123970152 on Build GRCh38
4494679 c.437C>T p.S146L Chr.3: 123970177-123970177 on Build GRCh38
5465697 c.500A>G p.Y167C Chr.3: 123970114-123970114 on Build GRCh38
1037392 c.490C>A p.L164I Chr.3: 123970124-123970124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376450544 Chr.3: 123970149-123970149 on Build GRCh38
rs755602717 Chr.3: 123970071-123970071 on Build GRCh38
rs763224764 Chr.3: 123970134-123970134 on Build GRCh38
rs779796181 Chr.3: 123970078-123970078 on Build GRCh38
rs1010917400 Chr.3: 123970039-123970039 on Build GRCh38
rs780163393 Chr.3: 123970213-123970213 on Build GRCh38
rs767494215 Chr.3: 123970150-123970150 on Build GRCh38
rs551141816 Chr.3: 123970038-123970038 on Build GRCh38
rs371695015 Chr.3: 123969996-123969996 on Build GRCh38
rs115823829 Chr.3: 123970409-123970409 on Build GRCh38
rs765968064 Chr.3: 123970256-123970256 on Build GRCh38
rs770749257 Chr.3: 123970041-123970041 on Build GRCh38
rs928189713 Chr.3: 123970393-123970393 on Build GRCh38
rs753547120 Chr.3: 123970084-123970084 on Build GRCh38
rs778255639 Chr.3: 123970171-123970171 on Build GRCh38
rs771768627 Chr.3: 123970241-123970241 on Build GRCh38
rs112305399 Chr.3: 123969987-123969987 on Build GRCh38
rs754729357 Chr.3: 123970170-123970170 on Build GRCh38
rs778341680 Chr.3: 123970098-123970098 on Build GRCh38
rs753530728 Chr.3: 123970168-123970168 on Build GRCh38
rs370738137 Chr.3: 123970399-123970399 on Build GRCh38
rs760138564 Chr.3: 123970232-123970232 on Build GRCh38
rs770013379 Chr.3: 123969994-123969994 on Build GRCh38
rs767440392 Chr.3: 123970152-123970152 on Build GRCh38
rs749489239 Chr.3: 123970217-123970217 on Build GRCh38
rs184315106 Chr.3: 123970108-123970108 on Build GRCh38
rs532182825 Chr.3: 123970090-123970090 on Build GRCh38
rs866431486 Chr.3: 123970421-123970421 on Build GRCh38
rs774083393 Chr.3: 123970115-123970115 on Build GRCh38
rs781679746 Chr.3: 123970192-123970192 on Build GRCh38
rs948328738 Chr.3: 123970310-123970310 on Build GRCh38
rs773198184 Chr.3: 123970242-123970242 on Build GRCh38
rs1052578057 Chr.3: 123970413-123970413 on Build GRCh38
rs766055172 Chr.3: 123970161-123970161 on Build GRCh38
rs760589785 Chr.3: 123970252-123970252 on Build GRCh38
rs761866401 Chr.3: 123970051-123970051 on Build GRCh38
rs760165915 Chr.3: 123970159-123970159 on Build GRCh38
rs536607735 Chr.3: 123970262-123970262 on Build GRCh38
rs756180229 Chr.3: 123970202-123970202 on Build GRCh38
rs140868038 Chr.3: 123970156-123970156 on Build GRCh38
rs775619072 Chr.3: 123970127-123970127 on Build GRCh38
rs146441719 Chr.3: 123970151-123970151 on Build GRCh38
rs200173296 Chr.3: 123970195-123970195 on Build GRCh38
rs201935912 Chr.3: 123970013-123970013 on Build GRCh38
rs765955172 Chr.3: 123970146-123970155 on Build GRCh38
rs747918291 Chr.3: 123970230-123970230 on Build GRCh38
rs796950287 Chr.3: 123970190-123970190 on Build GRCh38
rs113100460 Chr.3: 123970179-123970179 on Build GRCh38
rs769772715 Chr.3: 123970117-123970117 on Build GRCh38
rs140446139 Chr.3: 123970116-123970116 on Build GRCh38
rs763599256 Chr.3: 123970402-123970402 on Build GRCh38
rs545145825 Chr.3: 123970366-123970366 on Build GRCh38
rs145255014 Chr.3: 123970183-123970183 on Build GRCh38
rs201154163 Chr.3: 123970055-123970055 on Build GRCh38
rs144611765 Chr.3: 123969966-123969966 on Build GRCh38
rs766913548 Chr.3: 123970054-123970054 on Build GRCh38
rs982757446 Chr.3: 123970388-123970388 on Build GRCh38
rs747453184 Chr.3: 123970101-123970101 on Build GRCh38
rs369305016 Chr.3: 123970221-123970221 on Build GRCh38
rs774586301 Chr.3: 123970226-123970226 on Build GRCh38
rs757973469 Chr.3: 123970077-123970078 on Build GRCh38
rs750341418 Chr.3: 123970155-123970155 on Build GRCh38
rs751012202 Chr.3: 123970194-123970195 on Build GRCh38
rs762674878 Chr.3: 123970016-123970016 on Build GRCh38

View Full Product Details