Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126536190-126536696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf22
Entrez Gene ID
152065
Gene Name
chromosome 3 open reading frame 22
Gene Aliases
-
UniGene
Hs.178210
Gene Chromosome Location
Chr.3: 126524439-126558965 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Gene Symbol
CHST13
Entrez Gene ID
166012
Gene Name
carbohydrate sulfotransferase 13
Gene Aliases
C4ST3
UniGene
Hs.292375
Gene Chromosome Location
Chr.3: 126524288-126543291 on Build GRCh38
Transcripts
NR_130715.1 XM_017005753.1 NM_152889.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256627 c.159G>T p.Q53H Chr.3: 126536332-126536332 on Build GRCh38
4394993 c.165C>T p.L55L Chr.3: 126536338-126536338 on Build GRCh38
279897 c.169G>T p.D57Y Chr.3: 126536342-126536342 on Build GRCh38
1693419 c.122C>T p.S41F Chr.3: 126536295-126536295 on Build GRCh38
6096338 c.137G>C p.G46A Chr.3: 126536310-126536310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371306853 Chr.3: 126536372-126536372 on Build GRCh38
rs562208555 Chr.3: 126536334-126536334 on Build GRCh38
rs765199708 Chr.3: 126536278-126536278 on Build GRCh38
rs771995779 Chr.3: 126536357-126536357 on Build GRCh38
rs989431765 Chr.3: 126536496-126536496 on Build GRCh38
rs749019948 Chr.3: 126536229-126536229 on Build GRCh38
rs946596222 Chr.3: 126536624-126536624 on Build GRCh38
rs922545632 Chr.3: 126536217-126536217 on Build GRCh38
rs372513165 Chr.3: 126536370-126536370 on Build GRCh38
rs1015629945 Chr.3: 126536426-126536426 on Build GRCh38
rs761995292 Chr.3: 126536291-126536291 on Build GRCh38
rs529448346 Chr.3: 126536421-126536421 on Build GRCh38
rs920724113 Chr.3: 126536523-126536523 on Build GRCh38
rs1056304949 Chr.3: 126536663-126536663 on Build GRCh38
rs767740229 Chr.3: 126536298-126536298 on Build GRCh38
rs763223860 Chr.3: 126536279-126536279 on Build GRCh38
rs563070610 Chr.3: 126536303-126536303 on Build GRCh38
rs1001978114 Chr.3: 126536659-126536659 on Build GRCh38
rs944654873 Chr.3: 126536260-126536260 on Build GRCh38
rs756352688 Chr.3: 126536221-126536221 on Build GRCh38
rs777752549 Chr.3: 126536412-126536412 on Build GRCh38
rs933824498 Chr.3: 126536572-126536572 on Build GRCh38
rs764808625 Chr.3: 126536655-126536655 on Build GRCh38
rs774142375 Chr.3: 126536240-126536240 on Build GRCh38
rs973578443 Chr.3: 126536522-126536522 on Build GRCh38
rs908453275 Chr.3: 126536576-126536576 on Build GRCh38
rs756407635 Chr.3: 126536304-126536304 on Build GRCh38
rs756633493 Chr.3: 126536554-126536557 on Build GRCh38
rs780415942 Chr.3: 126536305-126536305 on Build GRCh38
rs375884058 Chr.3: 126536388-126536388 on Build GRCh38
rs762236814 Chr.3: 126536387-126536387 on Build GRCh38
rs73859518 Chr.3: 126536526-126536526 on Build GRCh38
rs372803956 Chr.3: 126536277-126536277 on Build GRCh38
rs751992539 Chr.3: 126536285-126536285 on Build GRCh38
rs764723746 Chr.3: 126536283-126536283 on Build GRCh38
rs913815420 Chr.3: 126536618-126536618 on Build GRCh38
rs768192036 Chr.3: 126536278-126536278 on Build GRCh38
rs771449702 Chr.3: 126536246-126536246 on Build GRCh38
rs368537915 Chr.3: 126536255-126536255 on Build GRCh38
rs370113692 Chr.3: 126536227-126536227 on Build GRCh38
rs956742638 Chr.3: 126536445-126536445 on Build GRCh38
rs374394795 Chr.3: 126536245-126536245 on Build GRCh38
rs551179629 Chr.3: 126536423-126536423 on Build GRCh38
rs894243876 Chr.3: 126536659-126536659 on Build GRCh38
rs953128962 Chr.3: 126536524-126536524 on Build GRCh38
rs778332825 Chr.3: 126536656-126536657 on Build GRCh38
rs570310099 Chr.3: 126536601-126536601 on Build GRCh38
rs147017058 Chr.3: 126536323-126536323 on Build GRCh38
rs552065314 Chr.3: 126536558-126536558 on Build GRCh38
rs183800650 Chr.3: 126536664-126536664 on Build GRCh38
rs768876114 Chr.3: 126536377-126536377 on Build GRCh38
rs139259499 Chr.3: 126536252-126536252 on Build GRCh38
rs775837447 Chr.3: 126536264-126536264 on Build GRCh38
rs200537285 Chr.3: 126536371-126536371 on Build GRCh38
rs1047053604 Chr.3: 126536574-126536574 on Build GRCh38
rs944395254 Chr.3: 126536556-126536556 on Build GRCh38
rs768702011 Chr.3: 126536239-126536239 on Build GRCh38
rs770637595 Chr.3: 126536369-126536369 on Build GRCh38
rs748064873 Chr.3: 126536341-126536341 on Build GRCh38
rs1043558542 Chr.3: 126536634-126536634 on Build GRCh38
rs905004882 Chr.3: 126536643-126536643 on Build GRCh38
rs769988267 Chr.3: 126536381-126536381 on Build GRCh38
rs569497772 Chr.3: 126536435-126536435 on Build GRCh38
rs911633726 Chr.3: 126536543-126536543 on Build GRCh38
rs199904615 Chr.3: 126536251-126536251 on Build GRCh38
rs777280426 Chr.3: 126536359-126536359 on Build GRCh38

View Full Product Details