Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136250241-136250747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCCB
Entrez Gene ID
5096
Gene Name
propionyl-CoA carboxylase beta subunit
Gene Aliases
-
UniGene
Hs.63788
Gene Chromosome Location
Chr.3: 136250325-136330171 on Build GRCh38
Transcripts
NM_000532.4 NM_001178014.1 XM_011512873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6511559 c.118G>A p.E40K Chr.3: 136250493-136250493 on Build GRCh38
5615207 c.146G>A p.G49E Chr.3: 136250521-136250521 on Build GRCh38
4711455 c.113G>A p.R38H Chr.3: 136250488-136250488 on Build GRCh38
1581383 c.49C>A p.L17M Chr.3: 136250424-136250424 on Build GRCh38
3123284 c.119A>G p.E40G Chr.3: 136250494-136250494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771580881 Chr.3: 136250602-136250602 on Build GRCh38
rs764394169 Chr.3: 136250451-136250451 on Build GRCh38
rs966385891 Chr.3: 136250468-136250468 on Build GRCh38
rs770177479 Chr.3: 136250586-136250586 on Build GRCh38
rs532403026 Chr.3: 136250663-136250663 on Build GRCh38
rs776965589 Chr.3: 136250575-136250575 on Build GRCh38
rs747222261 Chr.3: 136250574-136250574 on Build GRCh38
rs369284492 Chr.3: 136250370-136250370 on Build GRCh38
rs1031974178 Chr.3: 136250659-136250659 on Build GRCh38
rs1020586095 Chr.3: 136250310-136250310 on Build GRCh38
rs559320564 Chr.3: 136250444-136250444 on Build GRCh38
rs916034311 Chr.3: 136250357-136250357 on Build GRCh38
rs147954363 Chr.3: 136250550-136250550 on Build GRCh38
rs745694339 Chr.3: 136250569-136250570 on Build GRCh38
rs886058016 Chr.3: 136250341-136250341 on Build GRCh38
rs767965814 Chr.3: 136250392-136250392 on Build GRCh38
rs769039164 Chr.3: 136250530-136250530 on Build GRCh38
rs780842004 Chr.3: 136250421-136250421 on Build GRCh38
rs1000707726 Chr.3: 136250635-136250635 on Build GRCh38
rs754664563 Chr.3: 136250420-136250420 on Build GRCh38
rs763499684 Chr.3: 136250593-136250593 on Build GRCh38
rs371406048 Chr.3: 136250544-136250544 on Build GRCh38
rs141137691 Chr.3: 136250457-136250457 on Build GRCh38
rs776900764 Chr.3: 136250534-136250534 on Build GRCh38
rs1031941989 Chr.3: 136250502-136250502 on Build GRCh38
rs570284303 Chr.3: 136250623-136250623 on Build GRCh38
rs959163694 Chr.3: 136250507-136250507 on Build GRCh38
rs755889266 Chr.3: 136250494-136250494 on Build GRCh38
rs745829924 Chr.3: 136250554-136250554 on Build GRCh38
rs140636870 Chr.3: 136250555-136250555 on Build GRCh38
rs778924099 Chr.3: 136250553-136250553 on Build GRCh38
rs373685372 Chr.3: 136250383-136250383 on Build GRCh38
rs771166696 Chr.3: 136250385-136250385 on Build GRCh38
rs765790211 Chr.3: 136250539-136250539 on Build GRCh38
rs759008477 Chr.3: 136250543-136250543 on Build GRCh38
rs145075817 Chr.3: 136250513-136250513 on Build GRCh38
rs749257605 Chr.3: 136250434-136250435 on Build GRCh38
rs757860152 Chr.3: 136250406-136250406 on Build GRCh38
rs372649775 Chr.3: 136250342-136250342 on Build GRCh38
rs901718142 Chr.3: 136250573-136250573 on Build GRCh38
rs748452553 Chr.3: 136250582-136250582 on Build GRCh38
rs879253813 Chr.3: 136250563-136250563 on Build GRCh38
rs377681768 Chr.3: 136250391-136250391 on Build GRCh38
rs777478881 Chr.3: 136250498-136250498 on Build GRCh38
rs398123464 Chr.3: 136250378-136250378 on Build GRCh38
rs911406903 Chr.3: 136250316-136250316 on Build GRCh38
rs546682085 Chr.3: 136250318-136250318 on Build GRCh38
rs145734157 Chr.3: 136250339-136250339 on Build GRCh38
rs189166481 Chr.3: 136250311-136250311 on Build GRCh38
rs182412270 Chr.3: 136250491-136250491 on Build GRCh38
rs565705171 Chr.3: 136250696-136250696 on Build GRCh38
rs775585333 Chr.3: 136250446-136250446 on Build GRCh38
rs959257286 Chr.3: 136250676-136250676 on Build GRCh38
rs200185747 Chr.3: 136250424-136250424 on Build GRCh38
rs147606121 Chr.3: 136250695-136250695 on Build GRCh38
rs761030422 Chr.3: 136250450-136250450 on Build GRCh38
rs754303318 Chr.3: 136250405-136250405 on Build GRCh38
rs747214250 Chr.3: 136250526-136250526 on Build GRCh38
rs749210374 Chr.3: 136250505-136250505 on Build GRCh38
rs553950866 Chr.3: 136250325-136250325 on Build GRCh38
rs772483250 Chr.3: 136250442-136250442 on Build GRCh38
rs750005289 Chr.3: 136250353-136250353 on Build GRCh38
rs771885272 Chr.3: 136250599-136250599 on Build GRCh38
rs753617448 Chr.3: 136250265-136250265 on Build GRCh38
rs773714443 Chr.3: 136250540-136250540 on Build GRCh38
rs34167145 Chr.3: 136250573-136250573 on Build GRCh38
rs398123460 Chr.3: 136250559-136250559 on Build GRCh38
rs753174948 Chr.3: 136250395-136250395 on Build GRCh38
rs780042796 Chr.3: 136250566-136250566 on Build GRCh38
rs774313248 Chr.3: 136250550-136250550 on Build GRCh38
rs375014273 Chr.3: 136250506-136250506 on Build GRCh38
rs398123461 Chr.3: 136250561-136250561 on Build GRCh38
rs968982215 Chr.3: 136250304-136250304 on Build GRCh38
rs968929973 Chr.3: 136250448-136250448 on Build GRCh38
rs751136139 Chr.3: 136250469-136250469 on Build GRCh38
rs976378178 Chr.3: 136250294-136250294 on Build GRCh38
rs1010218920 Chr.3: 136250440-136250440 on Build GRCh38
rs1039491317 Chr.3: 136250565-136250565 on Build GRCh38
rs778285068 Chr.3: 136250351-136250351 on Build GRCh38
rs941240553 Chr.3: 136250580-136250580 on Build GRCh38
rs547330460 Chr.3: 136250705-136250705 on Build GRCh38
rs777359703 Chr.3: 136250376-136250377 on Build GRCh38
rs190302995 Chr.3: 136250714-136250714 on Build GRCh38
rs571188323 Chr.3: 136250287-136250287 on Build GRCh38
rs754759438 Chr.3: 136250268-136250268 on Build GRCh38
rs561539546 Chr.3: 136250538-136250538 on Build GRCh38
rs865998519 Chr.3: 136250621-136250621 on Build GRCh38
rs764602019 Chr.3: 136250398-136250398 on Build GRCh38
rs771117263 Chr.3: 136250439-136250439 on Build GRCh38
rs377349557 Chr.3: 136250358-136250358 on Build GRCh38
rs1027737054 Chr.3: 136250270-136250270 on Build GRCh38
rs770597925 Chr.3: 136250371-136250371 on Build GRCh38
rs1044957280 Chr.3: 136250612-136250612 on Build GRCh38
rs751837162 Chr.3: 136250330-136250330 on Build GRCh38
rs775738106 Chr.3: 136250521-136250521 on Build GRCh38
rs887644809 Chr.3: 136250274-136250274 on Build GRCh38
rs376983709 Chr.3: 136250517-136250517 on Build GRCh38
rs770769499 Chr.3: 136250476-136250476 on Build GRCh38
rs909550005 Chr.3: 136250557-136250557 on Build GRCh38
rs769835356 Chr.3: 136250381-136250381 on Build GRCh38
rs748003396 Chr.3: 136250380-136250380 on Build GRCh38
rs768392066 Chr.3: 136250365-136250365 on Build GRCh38
rs763025114 Chr.3: 136250384-136250384 on Build GRCh38
rs543941015 Chr.3: 136250619-136250619 on Build GRCh38
rs759836996 Chr.3: 136250441-136250441 on Build GRCh38
rs757856928 Chr.3: 136250355-136250355 on Build GRCh38
rs777726717 Chr.3: 136250433-136250433 on Build GRCh38
rs767133576 Chr.3: 136250473-136250473 on Build GRCh38
rs199935380 Chr.3: 136250514-136250514 on Build GRCh38
rs941297817 Chr.3: 136250366-136250366 on Build GRCh38
rs527592486 Chr.3: 136250700-136250700 on Build GRCh38
rs139744249 Chr.3: 136250613-136250613 on Build GRCh38
rs769481450 Chr.3: 136250428-136250428 on Build GRCh38
rs778754792 Chr.3: 136250275-136250275 on Build GRCh38
rs765843410 Chr.3: 136250461-136250461 on Build GRCh38
rs774667212 Chr.3: 136250386-136250386 on Build GRCh38
rs749205316 Chr.3: 136250434-136250434 on Build GRCh38
rs567310996 Chr.3: 136250326-136250326 on Build GRCh38
rs773661426 Chr.3: 136250587-136250587 on Build GRCh38
rs756687004 Chr.3: 136250348-136250348 on Build GRCh38
rs757134877 Chr.3: 136250552-136250552 on Build GRCh38
rs904521735 Chr.3: 136250704-136250704 on Build GRCh38
rs752645716 Chr.3: 136250283-136250283 on Build GRCh38
rs35838767 Chr.3: 136250451-136250452 on Build GRCh38
rs398123462 Chr.3: 136250377-136250377 on Build GRCh38
rs759939456 Chr.3: 136250331-136250331 on Build GRCh38
rs1056723902 Chr.3: 136250645-136250645 on Build GRCh38
rs916083200 Chr.3: 136250515-136250515 on Build GRCh38
rs774932784 Chr.3: 136250608-136250608 on Build GRCh38
rs770951842 Chr.3: 136250523-136250524 on Build GRCh38
rs200636968 Chr.3: 136250545-136250545 on Build GRCh38
rs763949276 Chr.3: 136250548-136250548 on Build GRCh38
rs1045974346 Chr.3: 136250367-136250367 on Build GRCh38
rs751189567 Chr.3: 136250414-136250414 on Build GRCh38

View Full Product Details