Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136293638-136294133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCCB
Entrez Gene ID
5096
Gene Name
propionyl-CoA carboxylase beta subunit
Gene Aliases
-
UniGene
Hs.63788
Gene Chromosome Location
Chr.3: 136250325-136330171 on Build GRCh38
Transcripts
NM_000532.4 NM_001178014.1 XM_011512873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892900 c.662C>T p.S221F Chr.3: 136293763-136293763 on Build GRCh38
3365007 c.744G>C p.K248N Chr.3: 136293845-136293845 on Build GRCh38
6489532 c.700G>T p.V234F Chr.3: 136293801-136293801 on Build GRCh38
5016861 c.707A>G p.N236S Chr.3: 136293808-136293808 on Build GRCh38
1733217 c.673A>T p.I225F Chr.3: 136293774-136293774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761472260 Chr.3: 136293809-136293809 on Build GRCh38
rs755258738 Chr.3: 136293736-136293736 on Build GRCh38
rs774304414 Chr.3: 136293898-136293898 on Build GRCh38
rs533935803 Chr.3: 136293812-136293812 on Build GRCh38
rs796052020 Chr.3: 136293754-136293754 on Build GRCh38
rs756533167 Chr.3: 136293750-136293750 on Build GRCh38
rs374722096 Chr.3: 136293784-136293784 on Build GRCh38
rs777413288 Chr.3: 136293730-136293730 on Build GRCh38
rs1038500366 Chr.3: 136293679-136293679 on Build GRCh38
rs551261472 Chr.3: 136293695-136293695 on Build GRCh38
rs746259163 Chr.3: 136294093-136294093 on Build GRCh38
rs755054401 Chr.3: 136293831-136293831 on Build GRCh38
rs951740615 Chr.3: 136294020-136294020 on Build GRCh38
rs540270519 Chr.3: 136294089-136294089 on Build GRCh38
rs765100012 Chr.3: 136293810-136293810 on Build GRCh38
rs779581071 Chr.3: 136293847-136293847 on Build GRCh38
rs778955184 Chr.3: 136293808-136293808 on Build GRCh38
rs778769732 Chr.3: 136293818-136293819 on Build GRCh38
rs762786066 Chr.3: 136293896-136293896 on Build GRCh38
rs748444609 Chr.3: 136293748-136293748 on Build GRCh38
rs751695055 Chr.3: 136293824-136293824 on Build GRCh38
rs746448544 Chr.3: 136293848-136293848 on Build GRCh38
rs544310213 Chr.3: 136293914-136293914 on Build GRCh38
rs147032635 Chr.3: 136294075-136294075 on Build GRCh38
rs540967577 Chr.3: 136293794-136293794 on Build GRCh38
rs770797106 Chr.3: 136293897-136293897 on Build GRCh38
rs988241101 Chr.3: 136294101-136294101 on Build GRCh38
rs762986402 Chr.3: 136293820-136293820 on Build GRCh38
rs1022802671 Chr.3: 136294034-136294034 on Build GRCh38
rs1050044176 Chr.3: 136293709-136293709 on Build GRCh38
rs775211013 Chr.3: 136293773-136293773 on Build GRCh38
rs919902060 Chr.3: 136294046-136294046 on Build GRCh38
rs766526843 Chr.3: 136293726-136293726 on Build GRCh38
rs1015840807 Chr.3: 136293956-136293956 on Build GRCh38
rs993246607 Chr.3: 136293997-136293997 on Build GRCh38
rs775300517 Chr.3: 136293887-136293889 on Build GRCh38
rs186012209 Chr.3: 136294031-136294031 on Build GRCh38
rs191115697 Chr.3: 136293842-136293842 on Build GRCh38
rs766364049 Chr.3: 136293823-136293823 on Build GRCh38
rs781523031 Chr.3: 136293739-136293739 on Build GRCh38
rs777934224 Chr.3: 136293693-136293693 on Build GRCh38
rs971290235 Chr.3: 136294061-136294061 on Build GRCh38
rs768309097 Chr.3: 136293852-136293852 on Build GRCh38
rs771766167 Chr.3: 136293874-136293875 on Build GRCh38
rs778095026 Chr.3: 136293753-136293753 on Build GRCh38
rs778242891 Chr.3: 136293835-136293835 on Build GRCh38
rs757845027 Chr.3: 136293845-136293845 on Build GRCh38
rs932684358 Chr.3: 136293703-136293708 on Build GRCh38
rs1052979420 Chr.3: 136293783-136293783 on Build GRCh38
rs776370260 Chr.3: 136293858-136293858 on Build GRCh38
rs891496033 Chr.3: 136293765-136293765 on Build GRCh38
rs371685159 Chr.3: 136293903-136293903 on Build GRCh38
rs920018872 Chr.3: 136293701-136293701 on Build GRCh38
rs747881252 Chr.3: 136293859-136293859 on Build GRCh38
rs750402220 Chr.3: 136293710-136293710 on Build GRCh38
rs745811235 Chr.3: 136293857-136293859 on Build GRCh38
rs180982841 Chr.3: 136293874-136293874 on Build GRCh38
rs368081650 Chr.3: 136293732-136293732 on Build GRCh38
rs961589004 Chr.3: 136293969-136293969 on Build GRCh38
rs770586617 Chr.3: 136293954-136293954 on Build GRCh38
rs756414710 Chr.3: 136293834-136293834 on Build GRCh38
rs570594951 Chr.3: 136293933-136293933 on Build GRCh38
rs201602404 Chr.3: 136293892-136293892 on Build GRCh38
rs773124697 Chr.3: 136293891-136293891 on Build GRCh38
rs746653827 Chr.3: 136293777-136293777 on Build GRCh38
rs756802198 Chr.3: 136293790-136293790 on Build GRCh38
rs544351816 Chr.3: 136294042-136294042 on Build GRCh38
rs76321585 Chr.3: 136294108-136294108 on Build GRCh38
rs1027839148 Chr.3: 136294007-136294007 on Build GRCh38
rs541708179 Chr.3: 136293921-136293921 on Build GRCh38
rs897539246 Chr.3: 136293964-136293964 on Build GRCh38
rs373255544 Chr.3: 136294076-136294076 on Build GRCh38
rs960884807 Chr.3: 136293945-136293945 on Build GRCh38
rs752940167 Chr.3: 136293833-136293833 on Build GRCh38
rs1021206121 Chr.3: 136293804-136293804 on Build GRCh38
rs571437349 Chr.3: 136293900-136293900 on Build GRCh38
rs376645261 Chr.3: 136293761-136293761 on Build GRCh38
rs771455499 Chr.3: 136293764-136293764 on Build GRCh38

View Full Product Details