Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110223601-110224105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC4
Entrez Gene ID
3749
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
C1orf30, HKSHIIIC, KSHIIIC, KV3.4
UniGene
Hs.153521
Gene Chromosome Location
Chr.1: 110210714-110282649 on Build GRCh38
Transcripts
NM_001039574.2 NM_004978.4 NR_036437.1 XM_006710625.3 XM_006710627.3 XM_011541401.2 XM_011541402.1 XM_011541403.1 XM_011541404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617067 c.1493G>A p.R498Q Chr.1: 110223778-110223778 on Build GRCh38
5594731 c.1392C>T p.I464I Chr.1: 110223677-110223677 on Build GRCh38
4617066 c.1493G>A p.R498Q Chr.1: 110223778-110223778 on Build GRCh38
6690653 c.1499C>T p.A500V Chr.1: 110223784-110223784 on Build GRCh38
462566 c.1574C>T p.P525L Chr.1: 110223859-110223859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775939037 Chr.1: 110223710-110223710 on Build GRCh38
rs775766459 Chr.1: 110223989-110223989 on Build GRCh38
rs887985288 Chr.1: 110223857-110223857 on Build GRCh38
rs937131166 Chr.1: 110223807-110223807 on Build GRCh38
rs12411176 Chr.1: 110223844-110223844 on Build GRCh38
rs529032999 Chr.1: 110224065-110224065 on Build GRCh38
rs769642146 Chr.1: 110223888-110223888 on Build GRCh38
rs543296604 Chr.1: 110224040-110224040 on Build GRCh38
rs757822089 Chr.1: 110223677-110223677 on Build GRCh38
rs947726650 Chr.1: 110224078-110224078 on Build GRCh38
rs780984588 Chr.1: 110223782-110223782 on Build GRCh38
rs779343443 Chr.1: 110223937-110223937 on Build GRCh38
rs909824049 Chr.1: 110223966-110223966 on Build GRCh38
rs139444791 Chr.1: 110223894-110223894 on Build GRCh38
rs749727219 Chr.1: 110223784-110223784 on Build GRCh38
rs966007199 Chr.1: 110223678-110223678 on Build GRCh38
rs574096948 Chr.1: 110223861-110223861 on Build GRCh38
rs747764524 Chr.1: 110223667-110223667 on Build GRCh38
rs759701270 Chr.1: 110223887-110223887 on Build GRCh38
rs367870522 Chr.1: 110223711-110223711 on Build GRCh38
rs774462856 Chr.1: 110224035-110224035 on Build GRCh38
rs201507570 Chr.1: 110223895-110223895 on Build GRCh38
rs59123361 Chr.1: 110223832-110223832 on Build GRCh38
rs748703547 Chr.1: 110223867-110223867 on Build GRCh38
rs564083351 Chr.1: 110223939-110223939 on Build GRCh38
rs762003338 Chr.1: 110223758-110223758 on Build GRCh38
rs186028377 Chr.1: 110224057-110224057 on Build GRCh38
rs374823466 Chr.1: 110223951-110223951 on Build GRCh38
rs765279902 Chr.1: 110223772-110223772 on Build GRCh38
rs376300705 Chr.1: 110223785-110223785 on Build GRCh38
rs199853223 Chr.1: 110223890-110223890 on Build GRCh38
rs768466768 Chr.1: 110224030-110224030 on Build GRCh38
rs866305493 Chr.1: 110224082-110224082 on Build GRCh38
rs372024993 Chr.1: 110223777-110223777 on Build GRCh38
rs201939187 Chr.1: 110223665-110223665 on Build GRCh38
rs200055497 Chr.1: 110223706-110223706 on Build GRCh38
rs764896644 Chr.1: 110223827-110223827 on Build GRCh38
rs143611044 Chr.1: 110223868-110223868 on Build GRCh38
rs750070382 Chr.1: 110223860-110223860 on Build GRCh38
rs781188745 Chr.1: 110223979-110223979 on Build GRCh38
rs778305202 Chr.1: 110223950-110223950 on Build GRCh38
rs755665914 Chr.1: 110223931-110223931 on Build GRCh38
rs1044023257 Chr.1: 110223836-110223836 on Build GRCh38
rs766560714 Chr.1: 110223826-110223826 on Build GRCh38
rs1031477575 Chr.1: 110223851-110223851 on Build GRCh38
rs150643950 Chr.1: 110223862-110223862 on Build GRCh38
rs760035546 Chr.1: 110224081-110224081 on Build GRCh38
rs753798744 Chr.1: 110223631-110223631 on Build GRCh38
rs201533610 Chr.1: 110223774-110223774 on Build GRCh38
rs183575959 Chr.1: 110223831-110223831 on Build GRCh38
rs774705792 Chr.1: 110223722-110223722 on Build GRCh38
rs761450657 Chr.1: 110223901-110223901 on Build GRCh38
rs758780575 Chr.1: 110223945-110223945 on Build GRCh38
rs755978329 Chr.1: 110223623-110223623 on Build GRCh38
rs985277923 Chr.1: 110223732-110223732 on Build GRCh38
rs953966957 Chr.1: 110223714-110223714 on Build GRCh38
rs370035918 Chr.1: 110223830-110223830 on Build GRCh38
rs373322407 Chr.1: 110223897-110223897 on Build GRCh38
rs776865803 Chr.1: 110223885-110223885 on Build GRCh38
rs759363747 Chr.1: 110223825-110223825 on Build GRCh38
rs747154105 Chr.1: 110223877-110223877 on Build GRCh38
rs943730852 Chr.1: 110223834-110223834 on Build GRCh38
rs1029005555 Chr.1: 110223747-110223747 on Build GRCh38
rs1000017547 Chr.1: 110223664-110223664 on Build GRCh38
rs760839541 Chr.1: 110223767-110223767 on Build GRCh38
rs145221541 Chr.1: 110223626-110223626 on Build GRCh38
rs770979171 Chr.1: 110223884-110223884 on Build GRCh38
rs754766832 Chr.1: 110223771-110223771 on Build GRCh38
rs953414642 Chr.1: 110223942-110223942 on Build GRCh38
rs769975544 Chr.1: 110223985-110223985 on Build GRCh38
rs200706008 Chr.1: 110223824-110223824 on Build GRCh38
rs762704495 Chr.1: 110223996-110223996 on Build GRCh38
rs369670720 Chr.1: 110223846-110223846 on Build GRCh38
rs774962835 Chr.1: 110223803-110223803 on Build GRCh38
rs769351147 Chr.1: 110223715-110223715 on Build GRCh38
rs148139999 Chr.1: 110223855-110223855 on Build GRCh38
rs143546722 Chr.1: 110223759-110223759 on Build GRCh38
rs150979037 Chr.1: 110223770-110223770 on Build GRCh38
rs34364747 Chr.1: 110223991-110223992 on Build GRCh38
rs771017607 Chr.1: 110223959-110223959 on Build GRCh38
rs767096353 Chr.1: 110223920-110223920 on Build GRCh38
rs746598938 Chr.1: 110223696-110223696 on Build GRCh38
rs146151572 Chr.1: 110223818-110223818 on Build GRCh38
rs543761843 Chr.1: 110223839-110223839 on Build GRCh38
rs769233932 Chr.1: 110223841-110223841 on Build GRCh38
rs376704379 Chr.1: 110223778-110223778 on Build GRCh38
rs367839863 Chr.1: 110223980-110223980 on Build GRCh38
rs367634024 Chr.1: 110223781-110223781 on Build GRCh38
rs770301734 Chr.1: 110223699-110223699 on Build GRCh38
rs1006713654 Chr.1: 110223879-110223879 on Build GRCh38
rs962848902 Chr.1: 110223974-110223974 on Build GRCh38
rs566360632 Chr.1: 110223806-110223806 on Build GRCh38
rs768599105 Chr.1: 110223808-110223808 on Build GRCh38
rs372719409 Chr.1: 110224052-110224052 on Build GRCh38
rs140378578 Chr.1: 110223760-110223760 on Build GRCh38
rs777518449 Chr.1: 110223684-110223684 on Build GRCh38
rs372407799 Chr.1: 110223802-110223802 on Build GRCh38
rs191317326 Chr.1: 110223727-110223727 on Build GRCh38
rs778341122 Chr.1: 110223874-110223874 on Build GRCh38
rs750125320 Chr.1: 110223929-110223929 on Build GRCh38

View Full Product Details