Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110606108-110606601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA2
Entrez Gene ID
3737
Gene Name
potassium voltage-gated channel subfamily A member 2
Gene Aliases
EIEE32, HBK5, HK4, HUKIV, KV1.2, MK2, NGK1, RBK2
UniGene
Hs.248139
Gene Chromosome Location
Chr.1: 110593580-110631536 on Build GRCh38
Transcripts
NM_001204269.1 NM_004974.3 XM_011541396.2 XM_011541397.2 XM_011541398.2 XM_011541399.2 XM_011541400.2 XM_017001213.1

SNP Detail

SNP ID Chromosome Location
rs931777719 Chr.1: 110606446-110606446 on Build GRCh38
rs757520289 Chr.1: 110606497-110606497 on Build GRCh38
rs546694076 Chr.1: 110606268-110606268 on Build GRCh38
rs963491943 Chr.1: 110606130-110606130 on Build GRCh38
rs905217452 Chr.1: 110606329-110606329 on Build GRCh38
rs988324102 Chr.1: 110606169-110606169 on Build GRCh38
rs956836100 Chr.1: 110606355-110606355 on Build GRCh38
rs533938061 Chr.1: 110606233-110606233 on Build GRCh38
rs78039595 Chr.1: 110606155-110606155 on Build GRCh38
rs1056981264 Chr.1: 110606511-110606511 on Build GRCh38
rs969871740 Chr.1: 110606140-110606140 on Build GRCh38
rs768985295 Chr.1: 110606535-110606535 on Build GRCh38
rs927465573 Chr.1: 110606141-110606141 on Build GRCh38
rs184877007 Chr.1: 110606338-110606338 on Build GRCh38
rs955115387 Chr.1: 110606381-110606381 on Build GRCh38
rs114208459 Chr.1: 110606293-110606293 on Build GRCh38
rs555159051 Chr.1: 110606339-110606339 on Build GRCh38
rs919010611 Chr.1: 110606472-110606472 on Build GRCh38
rs3050009 Chr.1: 110606156-110606157 on Build GRCh38
rs569616089 Chr.1: 110606319-110606319 on Build GRCh38
rs923533711 Chr.1: 110606185-110606185 on Build GRCh38
rs936684386 Chr.1: 110606570-110606570 on Build GRCh38
rs538659868 Chr.1: 110606281-110606281 on Build GRCh38
rs376652188 Chr.1: 110606309-110606309 on Build GRCh38
rs992997577 Chr.1: 110606387-110606387 on Build GRCh38
rs929076802 Chr.1: 110606509-110606509 on Build GRCh38
rs967565007 Chr.1: 110606550-110606550 on Build GRCh38
rs895717378 Chr.1: 110606299-110606299 on Build GRCh38
rs1034396809 Chr.1: 110606525-110606525 on Build GRCh38
rs919062632 Chr.1: 110606271-110606271 on Build GRCh38
rs780149386 Chr.1: 110606246-110606246 on Build GRCh38
rs1023679005 Chr.1: 110606139-110606139 on Build GRCh38
rs1009694469 Chr.1: 110606357-110606357 on Build GRCh38
rs151146353 Chr.1: 110606155-110606156 on Build GRCh38
rs544390487 Chr.1: 110606405-110606405 on Build GRCh38
rs745341619 Chr.1: 110606185-110606186 on Build GRCh38
rs995254032 Chr.1: 110606520-110606520 on Build GRCh38
rs1053971717 Chr.1: 110606237-110606237 on Build GRCh38
rs917511103 Chr.1: 110606150-110606151 on Build GRCh38
rs369146884 Chr.1: 110606180-110606180 on Build GRCh38
rs977680937 Chr.1: 110606553-110606553 on Build GRCh38
rs936423861 Chr.1: 110606234-110606234 on Build GRCh38
rs554270186 Chr.1: 110606385-110606385 on Build GRCh38
rs375685227 Chr.1: 110606145-110606145 on Build GRCh38
rs948627428 Chr.1: 110606531-110606531 on Build GRCh38
rs957695979 Chr.1: 110606572-110606572 on Build GRCh38
rs734376 Chr.1: 110606136-110606136 on Build GRCh38
rs986422084 Chr.1: 110606390-110606390 on Build GRCh38
rs1057030854 Chr.1: 110606276-110606276 on Build GRCh38
rs1033694128 Chr.1: 110606559-110606559 on Build GRCh38
rs3860224 Chr.1: 110606367-110606367 on Build GRCh38
rs370558922 Chr.1: 110606411-110606411 on Build GRCh38
rs558573365 Chr.1: 110606414-110606414 on Build GRCh38
rs956888977 Chr.1: 110606168-110606168 on Build GRCh38
rs899135176 Chr.1: 110606512-110606512 on Build GRCh38
rs575151206 Chr.1: 110606458-110606458 on Build GRCh38
rs890376216 Chr.1: 110606295-110606295 on Build GRCh38
rs566318922 Chr.1: 110606270-110606270 on Build GRCh38
rs560752678 Chr.1: 110606160-110606160 on Build GRCh38
rs1036707311 Chr.1: 110606505-110606505 on Build GRCh38
rs867528087 Chr.1: 110606582-110606582 on Build GRCh38
rs929130557 Chr.1: 110606275-110606275 on Build GRCh38
rs1024516653 Chr.1: 110606388-110606388 on Build GRCh38
rs917120418 Chr.1: 110606528-110606528 on Build GRCh38
rs11102136 Chr.1: 110606200-110606200 on Build GRCh38
rs973113728 Chr.1: 110606420-110606420 on Build GRCh38
rs1032403242 Chr.1: 110606352-110606352 on Build GRCh38
rs905164546 Chr.1: 110606552-110606552 on Build GRCh38
rs1051543944 Chr.1: 110606448-110606448 on Build GRCh38
rs921694802 Chr.1: 110606439-110606439 on Build GRCh38
rs940600216 Chr.1: 110606481-110606481 on Build GRCh38
rs568685218 Chr.1: 110606347-110606347 on Build GRCh38
rs1004796713 Chr.1: 110606307-110606307 on Build GRCh38
rs993049954 Chr.1: 110606149-110606150 on Build GRCh38
rs900311742 Chr.1: 110606251-110606251 on Build GRCh38

View Full Product Details