Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111350125-111350635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIFO
Entrez Gene ID
128344
Gene Name
primary cilia formation
Gene Aliases
C1orf88, pitchfork
UniGene
Hs.172510
Gene Chromosome Location
Chr.1: 111324663-111353017 on Build GRCh38
Transcripts
NM_001300831.1 NM_181643.5 NR_125338.1 XM_005270472.1 XM_017000322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6120574 c.472G>C p.V158L Chr.1: 111350188-111350188 on Build GRCh38
212185 c.422C>T p.P141L Chr.1: 111350138-111350138 on Build GRCh38
674352 c.473T>A p.V158D Chr.1: 111350189-111350189 on Build GRCh38
5843057 c.451G>T p.G151* Chr.1: 111350167-111350167 on Build GRCh38
7161668 c.505G>A p.E169K Chr.1: 111350221-111350221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762666026 Chr.1: 111350169-111350169 on Build GRCh38
rs1034545298 Chr.1: 111350236-111350236 on Build GRCh38
rs369943952 Chr.1: 111350536-111350536 on Build GRCh38
rs773468106 Chr.1: 111350195-111350195 on Build GRCh38
rs914310543 Chr.1: 111350295-111350295 on Build GRCh38
rs532303863 Chr.1: 111350150-111350150 on Build GRCh38
rs765276862 Chr.1: 111350212-111350212 on Build GRCh38
rs947366626 Chr.1: 111350177-111350177 on Build GRCh38
rs887894184 Chr.1: 111350517-111350517 on Build GRCh38
rs752954545 Chr.1: 111350234-111350234 on Build GRCh38
rs189052451 Chr.1: 111350533-111350533 on Build GRCh38
rs979073067 Chr.1: 111350191-111350191 on Build GRCh38
rs761038570 Chr.1: 111350200-111350200 on Build GRCh38
rs144577712 Chr.1: 111350589-111350589 on Build GRCh38
rs978425449 Chr.1: 111350546-111350546 on Build GRCh38
rs140722327 Chr.1: 111350489-111350489 on Build GRCh38
rs569202373 Chr.1: 111350341-111350341 on Build GRCh38
rs533571339 Chr.1: 111350474-111350474 on Build GRCh38
rs560610443 Chr.1: 111350603-111350603 on Build GRCh38
rs1036389402 Chr.1: 111350547-111350547 on Build GRCh38
rs538185781 Chr.1: 111350344-111350344 on Build GRCh38
rs562750946 Chr.1: 111350201-111350201 on Build GRCh38
rs763759949 Chr.1: 111350190-111350190 on Build GRCh38
rs530070163 Chr.1: 111350578-111350578 on Build GRCh38
rs149188444 Chr.1: 111350209-111350209 on Build GRCh38
rs942545875 Chr.1: 111350214-111350214 on Build GRCh38
rs138990106 Chr.1: 111350479-111350479 on Build GRCh38
rs757356684 Chr.1: 111350232-111350232 on Build GRCh38
rs201565676 Chr.1: 111350153-111350153 on Build GRCh38
rs532542702 Chr.1: 111350604-111350604 on Build GRCh38
rs765392772 Chr.1: 111350203-111350203 on Build GRCh38
rs780911286 Chr.1: 111350346-111350346 on Build GRCh38
rs56255516 Chr.1: 111350287-111350287 on Build GRCh38
rs1009989380 Chr.1: 111350511-111350511 on Build GRCh38
rs1039607682 Chr.1: 111350472-111350472 on Build GRCh38
rs12033188 Chr.1: 111350372-111350372 on Build GRCh38
rs777995298 Chr.1: 111350235-111350235 on Build GRCh38
rs912210779 Chr.1: 111350362-111350362 on Build GRCh38
rs373100710 Chr.1: 111350261-111350261 on Build GRCh38
rs545975576 Chr.1: 111350519-111350519 on Build GRCh38
rs145133389 Chr.1: 111350303-111350303 on Build GRCh38
rs562714193 Chr.1: 111350531-111350531 on Build GRCh38
rs907191252 Chr.1: 111350584-111350584 on Build GRCh38
rs759187687 Chr.1: 111350156-111350156 on Build GRCh38
rs561984435 Chr.1: 111350558-111350558 on Build GRCh38
rs372973539 Chr.1: 111350315-111350315 on Build GRCh38
rs775334326 Chr.1: 111350168-111350168 on Build GRCh38
rs769411827 Chr.1: 111350502-111350502 on Build GRCh38
rs754339072 Chr.1: 111350207-111350207 on Build GRCh38
rs3820443 Chr.1: 111350283-111350283 on Build GRCh38
rs554848190 Chr.1: 111350363-111350363 on Build GRCh38
rs888428475 Chr.1: 111350335-111350335 on Build GRCh38
rs769418476 Chr.1: 111350158-111350158 on Build GRCh38
rs542097537 Chr.1: 111350543-111350543 on Build GRCh38
rs1014927044 Chr.1: 111350187-111350187 on Build GRCh38
rs1019986478 Chr.1: 111350516-111350516 on Build GRCh38
rs530043333 Chr.1: 111350279-111350279 on Build GRCh38
rs77570804 Chr.1: 111350247-111350247 on Build GRCh38
rs757074329 Chr.1: 111350260-111350260 on Build GRCh38

View Full Product Details