Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114677738-114678263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMPD1
Entrez Gene ID
270
Gene Name
adenosine monophosphate deaminase 1
Gene Aliases
MAD, MADA, MMDD
UniGene
Hs.89570
Gene Chromosome Location
Chr.1: 114673098-114695618 on Build GRCh38
Transcripts
NM_000036.2 NM_001172626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390709 c.1151C>A p.A384E Chr.1: 114677983-114677983 on Build GRCh38
2177369 c.1213A>G p.T405A Chr.1: 114677921-114677921 on Build GRCh38
1667844 c.1121A>C p.K374T Chr.1: 114678013-114678013 on Build GRCh38
2177368 c.1312A>G p.T438A Chr.1: 114677921-114677921 on Build GRCh38
1667843 c.1220A>C p.K407T Chr.1: 114678013-114678013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369767517 Chr.1: 114677960-114677960 on Build GRCh38
rs1030909067 Chr.1: 114677832-114677832 on Build GRCh38
rs375502580 Chr.1: 114677825-114677825 on Build GRCh38
rs1033810815 Chr.1: 114677934-114677934 on Build GRCh38
rs754437089 Chr.1: 114677950-114677950 on Build GRCh38
rs542991562 Chr.1: 114677833-114677833 on Build GRCh38
rs34245099 Chr.1: 114677795-114677798 on Build GRCh38
rs762722076 Chr.1: 114677924-114677924 on Build GRCh38
rs749039833 Chr.1: 114678004-114678004 on Build GRCh38
rs1054189037 Chr.1: 114677809-114677809 on Build GRCh38
rs769460760 Chr.1: 114677986-114677986 on Build GRCh38
rs764823637 Chr.1: 114678092-114678093 on Build GRCh38
rs191957200 Chr.1: 114678137-114678137 on Build GRCh38
rs774679347 Chr.1: 114678180-114678180 on Build GRCh38
rs765669377 Chr.1: 114678046-114678046 on Build GRCh38
rs764282319 Chr.1: 114678063-114678063 on Build GRCh38
rs960005448 Chr.1: 114678168-114678168 on Build GRCh38
rs143583425 Chr.1: 114678038-114678038 on Build GRCh38
rs560418264 Chr.1: 114677769-114677769 on Build GRCh38
rs369790858 Chr.1: 114677761-114677761 on Build GRCh38
rs571473762 Chr.1: 114677943-114677943 on Build GRCh38
rs962465663 Chr.1: 114677907-114677907 on Build GRCh38
rs966958957 Chr.1: 114678216-114678216 on Build GRCh38
rs960218778 Chr.1: 114677914-114677914 on Build GRCh38
rs899682761 Chr.1: 114677847-114677854 on Build GRCh38
rs543962875 Chr.1: 114677800-114677803 on Build GRCh38
rs200206709 Chr.1: 114677807-114677807 on Build GRCh38
rs567271778 Chr.1: 114678002-114678002 on Build GRCh38
rs774999766 Chr.1: 114677922-114677922 on Build GRCh38
rs781528554 Chr.1: 114677971-114677971 on Build GRCh38
rs375552614 Chr.1: 114678188-114678188 on Build GRCh38
rs945387821 Chr.1: 114677768-114677768 on Build GRCh38
rs35859650 Chr.1: 114677972-114677972 on Build GRCh38
rs781073590 Chr.1: 114677983-114677983 on Build GRCh38
rs970158274 Chr.1: 114677811-114677811 on Build GRCh38
rs879844274 Chr.1: 114677798-114677799 on Build GRCh38
rs779837672 Chr.1: 114677989-114677989 on Build GRCh38
rs532589702 Chr.1: 114677946-114677946 on Build GRCh38
rs369370692 Chr.1: 114678092-114678092 on Build GRCh38
rs771587586 Chr.1: 114678175-114678175 on Build GRCh38
rs773929610 Chr.1: 114677928-114677928 on Build GRCh38
rs547706415 Chr.1: 114677951-114677951 on Build GRCh38
rs751920096 Chr.1: 114677815-114677815 on Build GRCh38
rs756429163 Chr.1: 114678185-114678185 on Build GRCh38
rs555338206 Chr.1: 114677980-114677980 on Build GRCh38
rs1029528108 Chr.1: 114677886-114677886 on Build GRCh38
rs545286128 Chr.1: 114677760-114677760 on Build GRCh38
rs758582169 Chr.1: 114677970-114677970 on Build GRCh38
rs1001376319 Chr.1: 114677787-114677802 on Build GRCh38
rs903916702 Chr.1: 114677776-114677776 on Build GRCh38
rs761283118 Chr.1: 114678065-114678065 on Build GRCh38
rs143862592 Chr.1: 114678223-114678223 on Build GRCh38
rs992359951 Chr.1: 114677949-114677949 on Build GRCh38
rs527618011 Chr.1: 114677823-114677823 on Build GRCh38
rs1007698535 Chr.1: 114678127-114678127 on Build GRCh38
rs760321357 Chr.1: 114677772-114677772 on Build GRCh38
rs915800278 Chr.1: 114678176-114678176 on Build GRCh38
rs768205791 Chr.1: 114677927-114677927 on Build GRCh38
rs577919001 Chr.1: 114677757-114677757 on Build GRCh38
rs148647876 Chr.1: 114678224-114678224 on Build GRCh38
rs61179687 Chr.1: 114678139-114678139 on Build GRCh38
rs978092801 Chr.1: 114677978-114677978 on Build GRCh38
rs769666778 Chr.1: 114677920-114677920 on Build GRCh38
rs925914007 Chr.1: 114678065-114678065 on Build GRCh38
rs144014236 Chr.1: 114677913-114677913 on Build GRCh38
rs554825859 Chr.1: 114677992-114677992 on Build GRCh38
rs146437677 Chr.1: 114678022-114678022 on Build GRCh38
rs745880731 Chr.1: 114677985-114677985 on Build GRCh38
rs538867095 Chr.1: 114677819-114677819 on Build GRCh38
rs770350941 Chr.1: 114678071-114678071 on Build GRCh38
rs372451247 Chr.1: 114678037-114678037 on Build GRCh38
rs368208363 Chr.1: 114677994-114677994 on Build GRCh38
rs753835602 Chr.1: 114677803-114677803 on Build GRCh38
rs1025930916 Chr.1: 114678148-114678148 on Build GRCh38
rs959526511 Chr.1: 114677948-114677951 on Build GRCh38
rs1015693996 Chr.1: 114677904-114677904 on Build GRCh38
rs747148334 Chr.1: 114677877-114677877 on Build GRCh38
rs776335079 Chr.1: 114678052-114678052 on Build GRCh38
rs915710367 Chr.1: 114677957-114677957 on Build GRCh38
rs140176911 Chr.1: 114677902-114677902 on Build GRCh38
rs202052347 Chr.1: 114677876-114677876 on Build GRCh38
rs367641756 Chr.1: 114677874-114677874 on Build GRCh38
rs542562775 Chr.1: 114678206-114678206 on Build GRCh38
rs545166360 Chr.1: 114678131-114678131 on Build GRCh38
rs375535646 Chr.1: 114678053-114678053 on Build GRCh38
rs542940189 Chr.1: 114677917-114677917 on Build GRCh38
rs778972084 Chr.1: 114677859-114677859 on Build GRCh38
rs878996012 Chr.1: 114677800-114677815 on Build GRCh38
rs566349141 Chr.1: 114677964-114677964 on Build GRCh38
rs143303551 Chr.1: 114678023-114678023 on Build GRCh38
rs138993729 Chr.1: 114678012-114678012 on Build GRCh38
rs546734761 Chr.1: 114677808-114677811 on Build GRCh38
rs777776543 Chr.1: 114677883-114677883 on Build GRCh38
rs72687865 Chr.1: 114677799-114677799 on Build GRCh38
rs71093606 Chr.1: 114677846-114677853 on Build GRCh38
rs191152112 Chr.1: 114677903-114677903 on Build GRCh38
rs978552189 Chr.1: 114678205-114678205 on Build GRCh38
rs61401956 Chr.1: 114677850-114677853 on Build GRCh38
rs763860709 Chr.1: 114677802-114677802 on Build GRCh38

View Full Product Details