Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167720008-167720516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD10
Entrez Gene ID
11235
Gene Name
programmed cell death 10
Gene Aliases
CCM3, TFAR15
UniGene
Hs.478150
Gene Chromosome Location
Chr.3: 167683891-167735690 on Build GRCh38
Transcripts
NM_007217.3 NM_145859.1 NM_145860.1 XM_005247086.4 XM_005247087.4 XM_005247088.3 XM_006713485.3 XM_011512368.2 XM_011512369.2 XM_017005644.1 XM_017005645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4712999 c.95A>C p.E32A Chr.3: 167720063-167720063 on Build GRCh38
4713000 c.36T>G p.A12A Chr.3: 167720122-167720122 on Build GRCh38
353187 c.9_10GA>TT p.M3>? Chr.3: 167720148-167720149 on Build GRCh38
1484824 c.21G>C p.E7D Chr.3: 167720137-167720137 on Build GRCh38
5816372 c.21G>T p.E7D Chr.3: 167720137-167720137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527870507 Chr.3: 167720433-167720433 on Build GRCh38
rs779771380 Chr.3: 167720101-167720101 on Build GRCh38
rs956982137 Chr.3: 167720048-167720048 on Build GRCh38
rs1129086 Chr.3: 167720230-167720230 on Build GRCh38
rs767039857 Chr.3: 167720418-167720418 on Build GRCh38
rs940642471 Chr.3: 167720311-167720311 on Build GRCh38
rs984630391 Chr.3: 167720287-167720287 on Build GRCh38
rs1006646703 Chr.3: 167720239-167720239 on Build GRCh38
rs1051278725 Chr.3: 167720220-167720220 on Build GRCh38
rs1009835059 Chr.3: 167720454-167720454 on Build GRCh38
rs143814799 Chr.3: 167720140-167720140 on Build GRCh38
rs367727154 Chr.3: 167720171-167720171 on Build GRCh38
rs754875027 Chr.3: 167720115-167720115 on Build GRCh38
rs542736993 Chr.3: 167720098-167720098 on Build GRCh38
rs764290272 Chr.3: 167720090-167720090 on Build GRCh38
rs934279085 Chr.3: 167720475-167720475 on Build GRCh38
rs769635570 Chr.3: 167720136-167720136 on Build GRCh38
rs751602105 Chr.3: 167720052-167720053 on Build GRCh38
rs189016983 Chr.3: 167720089-167720089 on Build GRCh38
rs138275885 Chr.3: 167720100-167720100 on Build GRCh38
rs557015020 Chr.3: 167720214-167720214 on Build GRCh38
rs1036209507 Chr.3: 167720315-167720315 on Build GRCh38
rs981280582 Chr.3: 167720261-167720261 on Build GRCh38
rs932963696 Chr.3: 167720331-167720331 on Build GRCh38
rs1025964791 Chr.3: 167720294-167720294 on Build GRCh38
rs754957285 Chr.3: 167720197-167720197 on Build GRCh38
rs761169794 Chr.3: 167720323-167720323 on Build GRCh38
rs946770523 Chr.3: 167720078-167720078 on Build GRCh38
rs781637897 Chr.3: 167720230-167720232 on Build GRCh38
rs1025206066 Chr.3: 167720456-167720456 on Build GRCh38
rs376909722 Chr.3: 167720469-167720469 on Build GRCh38
rs371250285 Chr.3: 167720198-167720198 on Build GRCh38
rs548657457 Chr.3: 167720371-167720371 on Build GRCh38
rs572972046 Chr.3: 167720152-167720152 on Build GRCh38
rs751478724 Chr.3: 167720113-167720113 on Build GRCh38
rs566976122 Chr.3: 167720402-167720402 on Build GRCh38
rs552610997 Chr.3: 167720468-167720468 on Build GRCh38
rs565154432 Chr.3: 167720223-167720223 on Build GRCh38
rs780578142 Chr.3: 167720119-167720119 on Build GRCh38
rs747679505 Chr.3: 167720122-167720122 on Build GRCh38
rs146858915 Chr.3: 167720282-167720282 on Build GRCh38
rs775493345 Chr.3: 167720201-167720201 on Build GRCh38
rs140278271 Chr.3: 167720286-167720286 on Build GRCh38
rs1016576898 Chr.3: 167720271-167720271 on Build GRCh38
rs530171292 Chr.3: 167720322-167720322 on Build GRCh38
rs562649829 Chr.3: 167720304-167720304 on Build GRCh38
rs540192506 Chr.3: 167720175-167720175 on Build GRCh38
rs771446333 Chr.3: 167720172-167720172 on Build GRCh38
rs1001020204 Chr.3: 167720416-167720416 on Build GRCh38
rs752757510 Chr.3: 167720041-167720041 on Build GRCh38
rs1050607500 Chr.3: 167720345-167720345 on Build GRCh38
rs980760630 Chr.3: 167720484-167720484 on Build GRCh38

View Full Product Details