Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170084552-170085092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR160
Entrez Gene ID
26996
Gene Name
G protein-coupled receptor 160
Gene Aliases
GPCR1, GPCR150
UniGene
Hs.231320
Gene Chromosome Location
Chr.3: 170037323-170085415 on Build GRCh38
Transcripts
NM_014373.2 XM_005247346.3 XM_005247347.3 XM_006713582.3 XM_011512678.2 XM_017006161.1 XM_017006162.1 XM_017006163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4115147 c.906A>C p.K302N Chr.3: 170084878-170084878 on Build GRCh38
4115146 c.868A>G p.T290A Chr.3: 170084840-170084840 on Build GRCh38
3781110 c.762A>G p.P254P Chr.3: 170084734-170084734 on Build GRCh38
3365080 c.804G>A p.Q268Q Chr.3: 170084776-170084776 on Build GRCh38
6673879 c.708_709insA p.I239fs*28 Chr.3: 170084680-170084681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768860730 Chr.3: 170084595-170084595 on Build GRCh38
rs146297186 Chr.3: 170084813-170084813 on Build GRCh38
rs773645860 Chr.3: 170084707-170084710 on Build GRCh38
rs146985269 Chr.3: 170084790-170084790 on Build GRCh38
rs767287003 Chr.3: 170084647-170084647 on Build GRCh38
rs35969156 Chr.3: 170084908-170084908 on Build GRCh38
rs747186938 Chr.3: 170084594-170084594 on Build GRCh38
rs79112906 Chr.3: 170085053-170085053 on Build GRCh38
rs781101052 Chr.3: 170084994-170084997 on Build GRCh38
rs552972161 Chr.3: 170084846-170084846 on Build GRCh38
rs779144525 Chr.3: 170084932-170084932 on Build GRCh38
rs757277885 Chr.3: 170085022-170085024 on Build GRCh38
rs570236318 Chr.3: 170084633-170084633 on Build GRCh38
rs771901447 Chr.3: 170084761-170084761 on Build GRCh38
rs775728345 Chr.3: 170084590-170084590 on Build GRCh38
rs747932437 Chr.3: 170084969-170084969 on Build GRCh38
rs372137175 Chr.3: 170084587-170084587 on Build GRCh38
rs139558562 Chr.3: 170084828-170084828 on Build GRCh38
rs775716675 Chr.3: 170085003-170085004 on Build GRCh38
rs913131256 Chr.3: 170084775-170084775 on Build GRCh38
rs757623733 Chr.3: 170084851-170084851 on Build GRCh38
rs752373575 Chr.3: 170084983-170084985 on Build GRCh38
rs544108325 Chr.3: 170084697-170084697 on Build GRCh38
rs772699640 Chr.3: 170085005-170085005 on Build GRCh38
rs771360982 Chr.3: 170084756-170084758 on Build GRCh38
rs773179424 Chr.3: 170084749-170084749 on Build GRCh38
rs758830676 Chr.3: 170084682-170084682 on Build GRCh38
rs751971278 Chr.3: 170084722-170084722 on Build GRCh38
rs765749059 Chr.3: 170085021-170085021 on Build GRCh38
rs776765849 Chr.3: 170084966-170084966 on Build GRCh38
rs781559786 Chr.3: 170084800-170084800 on Build GRCh38
rs778858743 Chr.3: 170084628-170084628 on Build GRCh38
rs1049269309 Chr.3: 170084760-170084760 on Build GRCh38
rs773857353 Chr.3: 170085022-170085022 on Build GRCh38
rs769892382 Chr.3: 170084688-170084688 on Build GRCh38
rs534725792 Chr.3: 170084819-170084819 on Build GRCh38
rs749894318 Chr.3: 170084780-170084780 on Build GRCh38
rs997794790 Chr.3: 170085006-170085006 on Build GRCh38
rs771096280 Chr.3: 170084824-170084824 on Build GRCh38
rs749121327 Chr.3: 170084698-170084698 on Build GRCh38
rs371588925 Chr.3: 170084812-170084812 on Build GRCh38
rs201476169 Chr.3: 170084942-170084942 on Build GRCh38
rs1001508653 Chr.3: 170084843-170084843 on Build GRCh38
rs762337181 Chr.3: 170084863-170084863 on Build GRCh38
rs990373050 Chr.3: 170085058-170085058 on Build GRCh38
rs760528134 Chr.3: 170084769-170084769 on Build GRCh38
rs749317743 Chr.3: 170084821-170084821 on Build GRCh38
rs759114760 Chr.3: 170085025-170085025 on Build GRCh38
rs761859494 Chr.3: 170084619-170084619 on Build GRCh38
rs756990605 Chr.3: 170084726-170084726 on Build GRCh38
rs750880925 Chr.3: 170084675-170084675 on Build GRCh38
rs769057032 Chr.3: 170084721-170084721 on Build GRCh38
rs760162382 Chr.3: 170084990-170084992 on Build GRCh38
rs377308505 Chr.3: 170084815-170084815 on Build GRCh38
rs781475418 Chr.3: 170084852-170084852 on Build GRCh38
rs968096166 Chr.3: 170084958-170084958 on Build GRCh38
rs776570089 Chr.3: 170084835-170084835 on Build GRCh38
rs752100174 Chr.3: 170084791-170084791 on Build GRCh38
rs774710652 Chr.3: 170084868-170084872 on Build GRCh38
rs745603395 Chr.3: 170084630-170084631 on Build GRCh38
rs760706542 Chr.3: 170084830-170084830 on Build GRCh38
rs376639895 Chr.3: 170084672-170084672 on Build GRCh38
rs746873485 Chr.3: 170084933-170084933 on Build GRCh38
rs764007123 Chr.3: 170084770-170084770 on Build GRCh38
rs369808304 Chr.3: 170085038-170085038 on Build GRCh38
rs764782116 Chr.3: 170084840-170084840 on Build GRCh38
rs752442376 Chr.3: 170084650-170084650 on Build GRCh38
rs1054333575 Chr.3: 170084748-170084748 on Build GRCh38
rs764735904 Chr.3: 170084777-170084777 on Build GRCh38
rs546087919 Chr.3: 170084912-170084912 on Build GRCh38
rs775559956 Chr.3: 170084768-170084768 on Build GRCh38
rs757984039 Chr.3: 170084787-170084787 on Build GRCh38
rs777838754 Chr.3: 170084918-170084918 on Build GRCh38
rs774305246 Chr.3: 170084639-170084639 on Build GRCh38
rs749569388 Chr.3: 170084919-170084919 on Build GRCh38
rs142366644 Chr.3: 170084792-170084792 on Build GRCh38
rs138076750 Chr.3: 170085043-170085043 on Build GRCh38
rs756561779 Chr.3: 170084917-170084917 on Build GRCh38
rs142162073 Chr.3: 170085052-170085052 on Build GRCh38
rs758963706 Chr.3: 170084580-170084580 on Build GRCh38
rs529567697 Chr.3: 170085000-170085001 on Build GRCh38
rs767804991 Chr.3: 170084913-170084913 on Build GRCh38
rs776896213 Chr.3: 170084600-170084600 on Build GRCh38
rs748401347 Chr.3: 170084733-170084733 on Build GRCh38
rs768510633 Chr.3: 170084833-170084833 on Build GRCh38
rs533322464 Chr.3: 170084668-170084668 on Build GRCh38
rs746134920 Chr.3: 170085049-170085049 on Build GRCh38
rs747320666 Chr.3: 170084702-170084702 on Build GRCh38
rs148749842 Chr.3: 170084776-170084776 on Build GRCh38
rs972803595 Chr.3: 170084638-170084638 on Build GRCh38
rs748213865 Chr.3: 170084680-170084681 on Build GRCh38
rs759416350 Chr.3: 170084641-170084641 on Build GRCh38
rs199980016 Chr.3: 170084782-170084782 on Build GRCh38
rs564340262 Chr.3: 170084632-170084636 on Build GRCh38
rs759842450 Chr.3: 170084874-170084875 on Build GRCh38
rs765945136 Chr.3: 170084874-170084874 on Build GRCh38
rs760617683 Chr.3: 170084651-170084651 on Build GRCh38
rs762689539 Chr.3: 170085018-170085018 on Build GRCh38
rs756406779 Chr.3: 170085041-170085041 on Build GRCh38
rs753254990 Chr.3: 170085039-170085039 on Build GRCh38
rs767199943 Chr.3: 170084937-170084938 on Build GRCh38
rs781521940 Chr.3: 170084662-170084663 on Build GRCh38
rs761765017 Chr.3: 170084837-170084837 on Build GRCh38
rs753058492 Chr.3: 170084914-170084914 on Build GRCh38
rs1055766952 Chr.3: 170084883-170084883 on Build GRCh38
rs759452986 Chr.3: 170084757-170084757 on Build GRCh38
rs769816175 Chr.3: 170084740-170084740 on Build GRCh38
rs144269646 Chr.3: 170084658-170084658 on Build GRCh38
rs756783792 Chr.3: 170084663-170084663 on Build GRCh38
rs757231798 Chr.3: 170084921-170084921 on Build GRCh38
rs149617592 Chr.3: 170084657-170084657 on Build GRCh38
rs763512509 Chr.3: 170084708-170084709 on Build GRCh38
rs746801516 Chr.3: 170084602-170084603 on Build GRCh38
rs548241662 Chr.3: 170085010-170085010 on Build GRCh38
rs745775541 Chr.3: 170085017-170085017 on Build GRCh38

View Full Product Details