Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183919844-183920336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC5
Entrez Gene ID
10057
Gene Name
ATP binding cassette subfamily C member 5
Gene Aliases
ABC33, EST277145, MOAT-C, MOATC, MRP5, SMRP, pABC11
UniGene
Hs.368563
Gene Chromosome Location
Chr.3: 183919934-184018010 on Build GRCh38
Transcripts
NM_001320032.1 NM_005688.3 XM_005247058.4 XM_005247059.4 XM_011512314.1 XM_017005492.1

SNP Detail

SNP ID Chromosome Location
rs35543804 Chr.3: 183920083-183920083 on Build GRCh38
rs143330011 Chr.3: 183920110-183920110 on Build GRCh38
rs1035092260 Chr.3: 183920272-183920272 on Build GRCh38
rs887868094 Chr.3: 183920236-183920236 on Build GRCh38
rs3805114 Chr.3: 183919934-183919934 on Build GRCh38
rs73044694 Chr.3: 183920278-183920278 on Build GRCh38
rs111657478 Chr.3: 183920242-183920245 on Build GRCh38
rs150303978 Chr.3: 183920242-183920243 on Build GRCh38
rs924167876 Chr.3: 183920100-183920100 on Build GRCh38
rs150079849 Chr.3: 183920263-183920263 on Build GRCh38
rs942594930 Chr.3: 183919865-183919865 on Build GRCh38
rs896513145 Chr.3: 183919971-183919971 on Build GRCh38
rs537922268 Chr.3: 183920093-183920093 on Build GRCh38
rs191957272 Chr.3: 183920095-183920095 on Build GRCh38
rs562 Chr.3: 183920057-183920057 on Build GRCh38
rs796764222 Chr.3: 183920171-183920171 on Build GRCh38
rs995598478 Chr.3: 183920066-183920066 on Build GRCh38
rs546356591 Chr.3: 183919985-183919985 on Build GRCh38
rs532761265 Chr.3: 183920187-183920187 on Build GRCh38
rs369518015 Chr.3: 183920196-183920196 on Build GRCh38
rs755741842 Chr.3: 183920188-183920188 on Build GRCh38
rs963719536 Chr.3: 183920129-183920129 on Build GRCh38
rs781082097 Chr.3: 183920059-183920059 on Build GRCh38
rs376499573 Chr.3: 183919928-183919928 on Build GRCh38
rs914488933 Chr.3: 183919902-183919902 on Build GRCh38
rs369545875 Chr.3: 183919906-183919906 on Build GRCh38
rs561594029 Chr.3: 183920055-183920055 on Build GRCh38
rs1050493592 Chr.3: 183920209-183920209 on Build GRCh38
rs572834774 Chr.3: 183919964-183919964 on Build GRCh38
rs540027454 Chr.3: 183920023-183920023 on Build GRCh38
rs976475347 Chr.3: 183920160-183920160 on Build GRCh38
rs184559648 Chr.3: 183920210-183920210 on Build GRCh38
rs993808031 Chr.3: 183919878-183919878 on Build GRCh38
rs549703085 Chr.3: 183920267-183920267 on Build GRCh38
rs528040857 Chr.3: 183920266-183920266 on Build GRCh38
rs533084284 Chr.3: 183919927-183919927 on Build GRCh38
rs556712854 Chr.3: 183919943-183919943 on Build GRCh38
rs995754963 Chr.3: 183920304-183920304 on Build GRCh38
rs752260148 Chr.3: 183920243-183920243 on Build GRCh38
rs960725492 Chr.3: 183919989-183919989 on Build GRCh38
rs924460652 Chr.3: 183920241-183920241 on Build GRCh38
rs1025750832 Chr.3: 183920311-183920311 on Build GRCh38
rs967784854 Chr.3: 183920293-183920293 on Build GRCh38
rs767239318 Chr.3: 183920254-183920257 on Build GRCh38
rs897055146 Chr.3: 183920264-183920264 on Build GRCh38
rs4148596 Chr.3: 183920242-183920243 on Build GRCh38
rs776394998 Chr.3: 183920244-183920245 on Build GRCh38
rs1039654823 Chr.3: 183919885-183919885 on Build GRCh38
rs990621076 Chr.3: 183920012-183920012 on Build GRCh38
rs538449659 Chr.3: 183920276-183920276 on Build GRCh38
rs1025803127 Chr.3: 183920076-183920076 on Build GRCh38
rs114133046 Chr.3: 183920273-183920273 on Build GRCh38

View Full Product Details