Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185282859-185283364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHHADH
Entrez Gene ID
1962
Gene Name
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
Gene Aliases
ECHD, FRTS3, L-PBE, LBFP, LBP, PBFE
UniGene
Hs.429879
Gene Chromosome Location
Chr.3: 185190624-185282886 on Build GRCh38
Transcripts
XM_011512517.1 XM_017005839.1 NM_001242314.1 NM_001242317.1 NR_038322.1 XM_017007456.1 XM_017007458.1

Gene Symbol
MAP3K13
Entrez Gene ID
9175
Gene Name
mitogen-activated protein kinase kinase kinase 13
Gene Aliases
LZK, MEKK13, MLK
UniGene
Hs.591306
Gene Chromosome Location
Chr.3: 185282938-185489094 on Build GRCh38
Transcripts
XM_011512517.1 XM_017005839.1 NM_001242314.1 NM_001242317.1 NR_038322.1 XM_017007456.1 XM_017007458.1

SNP Detail

SNP ID Chromosome Location
rs4073440 Chr.3: 185283295-185283295 on Build GRCh38
rs949708640 Chr.3: 185282916-185282916 on Build GRCh38
rs1044047083 Chr.3: 185282926-185282926 on Build GRCh38
rs530025058 Chr.3: 185282919-185282919 on Build GRCh38
rs778832107 Chr.3: 185282939-185282939 on Build GRCh38
rs759725543 Chr.3: 185283303-185283303 on Build GRCh38
rs1050844449 Chr.3: 185283162-185283162 on Build GRCh38
rs76072522 Chr.3: 185283200-185283200 on Build GRCh38
rs999320828 Chr.3: 185283203-185283203 on Build GRCh38
rs917005632 Chr.3: 185283056-185283056 on Build GRCh38
rs548939152 Chr.3: 185283247-185283247 on Build GRCh38
rs995535652 Chr.3: 185283246-185283246 on Build GRCh38
rs982906442 Chr.3: 185283334-185283335 on Build GRCh38
rs965345827 Chr.3: 185283036-185283036 on Build GRCh38
rs909679506 Chr.3: 185283093-185283093 on Build GRCh38
rs771085733 Chr.3: 185283310-185283310 on Build GRCh38
rs553761515 Chr.3: 185283179-185283179 on Build GRCh38
rs926968606 Chr.3: 185283121-185283121 on Build GRCh38
rs983585873 Chr.3: 185283055-185283055 on Build GRCh38
rs575053758 Chr.3: 185282976-185282976 on Build GRCh38
rs971167006 Chr.3: 185283312-185283312 on Build GRCh38
rs374498027 Chr.3: 185283001-185283001 on Build GRCh38
rs144968974 Chr.3: 185283267-185283267 on Build GRCh38
rs1013122551 Chr.3: 185283288-185283288 on Build GRCh38
rs747054428 Chr.3: 185283178-185283178 on Build GRCh38
rs995086490 Chr.3: 185283044-185283044 on Build GRCh38
rs1054378535 Chr.3: 185283136-185283136 on Build GRCh38
rs1009647381 Chr.3: 185282992-185282992 on Build GRCh38
rs754882704 Chr.3: 185282888-185282888 on Build GRCh38
rs893258211 Chr.3: 185283238-185283238 on Build GRCh38
rs968984616 Chr.3: 185283002-185283002 on Build GRCh38
rs114221354 Chr.3: 185283199-185283199 on Build GRCh38
rs957661693 Chr.3: 185282994-185282994 on Build GRCh38
rs900481715 Chr.3: 185283214-185283214 on Build GRCh38
rs772096748 Chr.3: 185283022-185283022 on Build GRCh38
rs371067312 Chr.3: 185282968-185282968 on Build GRCh38
rs367784500 Chr.3: 185283025-185283025 on Build GRCh38
rs1042442574 Chr.3: 185283005-185283005 on Build GRCh38
rs541049057 Chr.3: 185282897-185282897 on Build GRCh38
rs979777956 Chr.3: 185283090-185283090 on Build GRCh38
rs982655877 Chr.3: 185283054-185283054 on Build GRCh38
rs565780441 Chr.3: 185283184-185283184 on Build GRCh38
rs991672310 Chr.3: 185282997-185282997 on Build GRCh38
rs571736076 Chr.3: 185283125-185283125 on Build GRCh38
rs949180017 Chr.3: 185282900-185282900 on Build GRCh38
rs77429300 Chr.3: 185283201-185283201 on Build GRCh38
rs77769097 Chr.3: 185283327-185283327 on Build GRCh38
rs753415307 Chr.3: 185282886-185282886 on Build GRCh38
rs983179930 Chr.3: 185282911-185282911 on Build GRCh38
rs935907311 Chr.3: 185282964-185282964 on Build GRCh38
rs971113018 Chr.3: 185283068-185283068 on Build GRCh38
rs530768898 Chr.3: 185282991-185282991 on Build GRCh38
rs559638200 Chr.3: 185282910-185282910 on Build GRCh38
rs979860542 Chr.3: 185283315-185283315 on Build GRCh38
rs950909547 Chr.3: 185283052-185283052 on Build GRCh38
rs1005073043 Chr.3: 185283291-185283291 on Build GRCh38
rs945889846 Chr.3: 185283154-185283154 on Build GRCh38
rs1028260464 Chr.3: 185283051-185283051 on Build GRCh38
rs913092251 Chr.3: 185283146-185283146 on Build GRCh38
rs929821186 Chr.3: 185282893-185282893 on Build GRCh38
rs547905903 Chr.3: 185283128-185283128 on Build GRCh38
rs973995602 Chr.3: 185282887-185282887 on Build GRCh38
rs941267290 Chr.3: 185283120-185283120 on Build GRCh38
rs777722233 Chr.3: 185283037-185283037 on Build GRCh38
rs886652500 Chr.3: 185283259-185283259 on Build GRCh38
rs554868647 Chr.3: 185283206-185283206 on Build GRCh38
rs999020405 Chr.3: 185282978-185282978 on Build GRCh38
rs929825424 Chr.3: 185283050-185283051 on Build GRCh38
rs111951887 Chr.3: 185282971-185282971 on Build GRCh38
rs1027914744 Chr.3: 185283258-185283258 on Build GRCh38
rs531261715 Chr.3: 185282915-185282915 on Build GRCh38
rs957054387 Chr.3: 185283328-185283328 on Build GRCh38
rs888353862 Chr.3: 185282901-185282901 on Build GRCh38
rs969058437 Chr.3: 185283294-185283294 on Build GRCh38
rs748122538 Chr.3: 185283010-185283010 on Build GRCh38
rs549973113 Chr.3: 185283008-185283008 on Build GRCh38

View Full Product Details