Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 195725979-195726497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC20
Entrez Gene ID
200958
Gene Name
mucin 20, cell surface associated
Gene Aliases
MUC-20
UniGene
Hs.308992
Gene Chromosome Location
Chr.3: 195720882-195733553 on Build GRCh38
Transcripts
NM_001282506.1 NM_001291833.1 NM_020790.1 NM_152673.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5534941 c.1406C>T p.P469L Chr.3: 195726009-195726009 on Build GRCh38
1149707 c.1314G>C p.K438N Chr.3: 195726484-195726484 on Build GRCh38
5455177 c.1749C>A p.N583K Chr.3: 195726352-195726352 on Build GRCh38
339807 c.1862C>T p.T621M Chr.3: 195726465-195726465 on Build GRCh38
4597266 c.1209-1T>C p.? Chr.3: 195726378-195726378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778636928 Chr.3: 195726232-195726232 on Build GRCh38
rs757903437 Chr.3: 195726128-195726128 on Build GRCh38
rs779386000 Chr.3: 195726040-195726040 on Build GRCh38
rs3828406 Chr.3: 195726386-195726386 on Build GRCh38
rs533915346 Chr.3: 195726396-195726396 on Build GRCh38
rs376180538 Chr.3: 195726448-195726448 on Build GRCh38
rs750811450 Chr.3: 195726140-195726140 on Build GRCh38
rs746427196 Chr.3: 195726369-195726369 on Build GRCh38
rs4093817 Chr.3: 195726089-195726089 on Build GRCh38
rs373203016 Chr.3: 195726375-195726375 on Build GRCh38
rs762600322 Chr.3: 195726117-195726117 on Build GRCh38
rs372141463 Chr.3: 195726466-195726466 on Build GRCh38
rs767806344 Chr.3: 195726119-195726119 on Build GRCh38
rs780767289 Chr.3: 195726207-195726207 on Build GRCh38
rs750403178 Chr.3: 195726143-195726143 on Build GRCh38
rs765298515 Chr.3: 195726330-195726330 on Build GRCh38
rs562232731 Chr.3: 195726106-195726106 on Build GRCh38
rs757452990 Chr.3: 195726403-195726403 on Build GRCh38
rs771608778 Chr.3: 195726045-195726045 on Build GRCh38
rs745885664 Chr.3: 195726406-195726406 on Build GRCh38
rs778934211 Chr.3: 195726291-195726291 on Build GRCh38
rs781623120 Chr.3: 195726470-195726470 on Build GRCh38
rs763494488 Chr.3: 195726188-195726188 on Build GRCh38
rs200951705 Chr.3: 195726465-195726465 on Build GRCh38
rs779514570 Chr.3: 195726101-195726101 on Build GRCh38
rs9866681 Chr.3: 195726116-195726116 on Build GRCh38
rs548603585 Chr.3: 195726150-195726150 on Build GRCh38
rs780748804 Chr.3: 195726146-195726146 on Build GRCh38
rs765795314 Chr.3: 195726179-195726179 on Build GRCh38
rs565508901 Chr.3: 195726058-195726058 on Build GRCh38
rs3828407 Chr.3: 195726378-195726378 on Build GRCh38
rs2550233 Chr.3: 195726193-195726193 on Build GRCh38
rs760315791 Chr.3: 195726324-195726324 on Build GRCh38
rs765973754 Chr.3: 195726165-195726166 on Build GRCh38
rs1018694131 Chr.3: 195726203-195726203 on Build GRCh38
rs745678866 Chr.3: 195726296-195726296 on Build GRCh38
rs756302465 Chr.3: 195726344-195726344 on Build GRCh38
rs373456247 Chr.3: 195726340-195726340 on Build GRCh38
rs770050031 Chr.3: 195726307-195726307 on Build GRCh38
rs202189746 Chr.3: 195726333-195726333 on Build GRCh38
rs751331604 Chr.3: 195726390-195726390 on Build GRCh38
rs747365863 Chr.3: 195726416-195726416 on Build GRCh38
rs772584537 Chr.3: 195726408-195726408 on Build GRCh38
rs761044751 Chr.3: 195726374-195726374 on Build GRCh38
rs368952817 Chr.3: 195726175-195726178 on Build GRCh38
rs754232475 Chr.3: 195726209-195726210 on Build GRCh38
rs755773681 Chr.3: 195726282-195726282 on Build GRCh38
rs767191940 Chr.3: 195726275-195726275 on Build GRCh38
rs754760300 Chr.3: 195726395-195726395 on Build GRCh38
rs771361744 Chr.3: 195726312-195726312 on Build GRCh38
rs9758494 Chr.3: 195726439-195726439 on Build GRCh38
rs764315332 Chr.3: 195726122-195726122 on Build GRCh38
rs563189586 Chr.3: 195726124-195726124 on Build GRCh38
rs558389727 Chr.3: 195726426-195726426 on Build GRCh38
rs770884143 Chr.3: 195726227-195726227 on Build GRCh38
rs774713064 Chr.3: 195726263-195726263 on Build GRCh38
rs192564648 Chr.3: 195726237-195726237 on Build GRCh38
rs3828408 Chr.3: 195726372-195726372 on Build GRCh38
rs746435828 Chr.3: 195726053-195726053 on Build GRCh38
rs776852644 Chr.3: 195726175-195726175 on Build GRCh38
rs371697936 Chr.3: 195726329-195726329 on Build GRCh38
rs766843643 Chr.3: 195726259-195726259 on Build GRCh38
rs760722486 Chr.3: 195725999-195725999 on Build GRCh38
rs752228945 Chr.3: 195726208-195726208 on Build GRCh38
rs761584464 Chr.3: 195726421-195726421 on Build GRCh38
rs748143745 Chr.3: 195726113-195726113 on Build GRCh38
rs759367297 Chr.3: 195726444-195726444 on Build GRCh38
rs760001706 Chr.3: 195726320-195726320 on Build GRCh38
rs757813638 Chr.3: 195726096-195726096 on Build GRCh38
rs750854832 Chr.3: 195726184-195726184 on Build GRCh38
rs3828411 Chr.3: 195726120-195726120 on Build GRCh38
rs752390520 Chr.3: 195726280-195726280 on Build GRCh38
rs201786008 Chr.3: 195726474-195726474 on Build GRCh38
rs752718179 Chr.3: 195726020-195726020 on Build GRCh38
rs779036709 Chr.3: 195726405-195726405 on Build GRCh38
rs140946665 Chr.3: 195726373-195726373 on Build GRCh38
rs748439238 Chr.3: 195726305-195726305 on Build GRCh38
rs774796725 Chr.3: 195726319-195726319 on Build GRCh38
rs776837792 Chr.3: 195726420-195726420 on Build GRCh38
rs781393465 Chr.3: 195726018-195726018 on Build GRCh38
rs754135416 Chr.3: 195726397-195726397 on Build GRCh38
rs751821376 Chr.3: 195726014-195726014 on Build GRCh38
rs758639943 Chr.3: 195726141-195726141 on Build GRCh38
rs201067977 Chr.3: 195726273-195726273 on Build GRCh38
rs752839315 Chr.3: 195726088-195726088 on Build GRCh38
rs750527278 Chr.3: 195726461-195726461 on Build GRCh38
rs202171251 Chr.3: 195726171-195726171 on Build GRCh38
rs776942524 Chr.3: 195726241-195726241 on Build GRCh38
rs757733993 Chr.3: 195726322-195726324 on Build GRCh38
rs765139750 Chr.3: 195726119-195726120 on Build GRCh38
rs770676738 Chr.3: 195726153-195726153 on Build GRCh38
rs372274630 Chr.3: 195726256-195726256 on Build GRCh38
rs560563342 Chr.3: 195726151-195726151 on Build GRCh38
rs777310652 Chr.3: 195726216-195726216 on Build GRCh38
rs758072325 Chr.3: 195726100-195726100 on Build GRCh38
rs144288174 Chr.3: 195726147-195726147 on Build GRCh38
rs373472255 Chr.3: 195726217-195726217 on Build GRCh38
rs370118314 Chr.3: 195726438-195726438 on Build GRCh38
rs766619706 Chr.3: 195726013-195726013 on Build GRCh38
rs368105388 Chr.3: 195726283-195726283 on Build GRCh38
rs754432854 Chr.3: 195726094-195726094 on Build GRCh38
rs779556780 Chr.3: 195726444-195726447 on Build GRCh38
rs58305151 Chr.3: 195726341-195726341 on Build GRCh38
rs767914017 Chr.3: 195726322-195726322 on Build GRCh38
rs749355582 Chr.3: 195726349-195726349 on Build GRCh38
rs768993180 Chr.3: 195726170-195726170 on Build GRCh38
rs376059301 Chr.3: 195726164-195726164 on Build GRCh38
rs780564191 Chr.3: 195726415-195726415 on Build GRCh38
rs761693996 Chr.3: 195726061-195726062 on Build GRCh38
rs554198408 Chr.3: 195726001-195726001 on Build GRCh38
rs746212743 Chr.3: 195726099-195726099 on Build GRCh38
rs866545718 Chr.3: 195726239-195726239 on Build GRCh38
rs376246962 Chr.3: 195726055-195726055 on Build GRCh38
rs776308281 Chr.3: 195726377-195726377 on Build GRCh38
rs2688542 Chr.3: 195726080-195726080 on Build GRCh38
rs779786660 Chr.3: 195726142-195726142 on Build GRCh38
rs200060340 Chr.3: 195726385-195726385 on Build GRCh38
rs779475073 Chr.3: 195726098-195726098 on Build GRCh38
rs373026727 Chr.3: 195726459-195726459 on Build GRCh38
rs368331468 Chr.3: 195726255-195726255 on Build GRCh38
rs762502872 Chr.3: 195726437-195726437 on Build GRCh38
rs544862173 Chr.3: 195726109-195726109 on Build GRCh38
rs73203946 Chr.3: 195726458-195726458 on Build GRCh38
rs749329559 Chr.3: 195726311-195726311 on Build GRCh38
rs201867992 Chr.3: 195726056-195726056 on Build GRCh38
rs749851441 Chr.3: 195726246-195726246 on Build GRCh38
rs753381078 Chr.3: 195726287-195726287 on Build GRCh38
rs3828405 Chr.3: 195726417-195726417 on Build GRCh38
rs754631977 Chr.3: 195726201-195726201 on Build GRCh38
rs745580271 Chr.3: 195726042-195726042 on Build GRCh38
rs763437748 Chr.3: 195726257-195726257 on Build GRCh38
rs755584993 Chr.3: 195726215-195726215 on Build GRCh38
rs758226396 Chr.3: 195726297-195726297 on Build GRCh38
rs776444759 Chr.3: 195726022-195726022 on Build GRCh38
rs779963793 Chr.3: 195726298-195726298 on Build GRCh38
rs201777974 Chr.3: 195726090-195726090 on Build GRCh38
rs372880164 Chr.3: 195726087-195726087 on Build GRCh38
rs772871124 Chr.3: 195726071-195726071 on Build GRCh38
rs572650436 Chr.3: 195726002-195726002 on Build GRCh38
rs769720088 Chr.3: 195726115-195726115 on Build GRCh38
rs200009986 Chr.3: 195726382-195726382 on Build GRCh38
rs536346831 Chr.3: 195726172-195726173 on Build GRCh38
rs577444973 Chr.3: 195726108-195726108 on Build GRCh38
rs755134488 Chr.3: 195726016-195726016 on Build GRCh38
rs764724566 Chr.3: 195726092-195726092 on Build GRCh38
rs540461603 Chr.3: 195726037-195726037 on Build GRCh38
rs761924665 Chr.3: 195726176-195726176 on Build GRCh38
rs200670251 Chr.3: 195726038-195726038 on Build GRCh38
rs780700418 Chr.3: 195726177-195726182 on Build GRCh38
rs751917792 Chr.3: 195726197-195726197 on Build GRCh38
rs762014480 Chr.3: 195726243-195726243 on Build GRCh38
rs9870815 Chr.3: 195726334-195726334 on Build GRCh38
rs966145111 Chr.3: 195726249-195726249 on Build GRCh38
rs3204353 Chr.3: 195726425-195726425 on Build GRCh38
rs753951718 Chr.3: 195726123-195726123 on Build GRCh38
rs3828410 Chr.3: 195726143-195726143 on Build GRCh38
rs746551150 Chr.3: 195726103-195726103 on Build GRCh38
rs775823249 Chr.3: 195726323-195726323 on Build GRCh38
rs568816906 Chr.3: 195726365-195726365 on Build GRCh38
rs200319871 Chr.3: 195726383-195726383 on Build GRCh38
rs371526551 Chr.3: 195726233-195726233 on Build GRCh38
rs756652109 Chr.3: 195726026-195726026 on Build GRCh38
rs757144011 Chr.3: 195726290-195726290 on Build GRCh38
rs772861528 Chr.3: 195726431-195726431 on Build GRCh38
rs771079983 Chr.3: 195726364-195726364 on Build GRCh38
rs754613494 Chr.3: 195726145-195726145 on Build GRCh38
rs775130401 Chr.3: 195726050-195726050 on Build GRCh38
rs530074504 Chr.3: 195726137-195726137 on Build GRCh38
rs765066194 Chr.3: 195726067-195726067 on Build GRCh38

View Full Product Details