Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1985995-1986490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR943
Entrez Gene ID
100126332
Gene Name
microRNA 943
Gene Aliases
MIRN943, hsa-mir-943
UniGene
Gene Chromosome Location
Chr.4: 1986384-1986477 on Build GRCh38
Transcripts
NR_030641.1 NM_005663.4 XM_017008589.1

Gene Symbol
NELFA
Entrez Gene ID
7469
Gene Name
negative elongation factor complex member A
Gene Aliases
NELF-A, P/OKcl.15, WHSC2
UniGene
Hs.21771
Gene Chromosome Location
Chr.4: 1982714-2009972 on Build GRCh38
Transcripts
NR_030641.1 NM_005663.4 XM_017008589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661033 c.660G>T p.Q220H Chr.4: 1986377-1986377 on Build GRCh38
4741219 c.686C>T p.P229L Chr.4: 1986384-1986384 on Build GRCh38
7280482 c.818G>A p.R273Q Chr.4: 1986131-1986131 on Build GRCh38
1232756 c.725C>T p.P242L Chr.4: 1986312-1986312 on Build GRCh38
4846301 c.806G>A p.R269Q Chr.4: 1986143-1986143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540803015 Chr.4: 1986227-1986227 on Build GRCh38
rs2234567 Chr.4: 1986013-1986013 on Build GRCh38
rs747693345 Chr.4: 1986349-1986349 on Build GRCh38
rs780766143 Chr.4: 1986347-1986347 on Build GRCh38
rs368408285 Chr.4: 1986106-1986106 on Build GRCh38
rs373280394 Chr.4: 1986216-1986216 on Build GRCh38
rs371676108 Chr.4: 1986108-1986108 on Build GRCh38
rs751930593 Chr.4: 1986212-1986212 on Build GRCh38
rs373613034 Chr.4: 1986264-1986264 on Build GRCh38
rs142924150 Chr.4: 1986389-1986389 on Build GRCh38
rs368905227 Chr.4: 1986449-1986449 on Build GRCh38
rs760311104 Chr.4: 1986081-1986081 on Build GRCh38
rs144909996 Chr.4: 1986151-1986151 on Build GRCh38
rs746328582 Chr.4: 1986357-1986357 on Build GRCh38
rs3034718 Chr.4: 1986461-1986462 on Build GRCh38
rs764871539 Chr.4: 1986417-1986420 on Build GRCh38
rs150291014 Chr.4: 1986122-1986122 on Build GRCh38
rs1048525339 Chr.4: 1986070-1986070 on Build GRCh38
rs201953811 Chr.4: 1986457-1986458 on Build GRCh38
rs766475819 Chr.4: 1986412-1986412 on Build GRCh38
rs938200010 Chr.4: 1986032-1986032 on Build GRCh38
rs761358582 Chr.4: 1986219-1986220 on Build GRCh38
rs374310144 Chr.4: 1986312-1986312 on Build GRCh38
rs765825469 Chr.4: 1986082-1986082 on Build GRCh38
rs761891212 Chr.4: 1986455-1986455 on Build GRCh38
rs775104766 Chr.4: 1986079-1986079 on Build GRCh38
rs965035471 Chr.4: 1986090-1986090 on Build GRCh38
rs757184445 Chr.4: 1986231-1986231 on Build GRCh38
rs1034890783 Chr.4: 1986398-1986398 on Build GRCh38
rs369149559 Chr.4: 1986095-1986095 on Build GRCh38
rs186568343 Chr.4: 1986417-1986417 on Build GRCh38
rs942872825 Chr.4: 1986261-1986261 on Build GRCh38
rs1037097269 Chr.4: 1986055-1986055 on Build GRCh38
rs536560942 Chr.4: 1986450-1986450 on Build GRCh38
rs200567269 Chr.4: 1986217-1986217 on Build GRCh38
rs1041258652 Chr.4: 1986305-1986305 on Build GRCh38
rs768212928 Chr.4: 1986191-1986191 on Build GRCh38
rs764259371 Chr.4: 1986310-1986310 on Build GRCh38
rs368356799 Chr.4: 1986411-1986411 on Build GRCh38
rs901434143 Chr.4: 1986313-1986313 on Build GRCh38
rs372580372 Chr.4: 1986413-1986413 on Build GRCh38
rs748834676 Chr.4: 1986439-1986439 on Build GRCh38
rs371533378 Chr.4: 1986431-1986431 on Build GRCh38
rs761072900 Chr.4: 1986375-1986375 on Build GRCh38
rs747700472 Chr.4: 1986428-1986428 on Build GRCh38
rs867042160 Chr.4: 1986464-1986464 on Build GRCh38
rs1000971550 Chr.4: 1986074-1986074 on Build GRCh38
rs754655693 Chr.4: 1986346-1986346 on Build GRCh38
rs549364702 Chr.4: 1986361-1986361 on Build GRCh38
rs938687232 Chr.4: 1986047-1986047 on Build GRCh38
rs374079729 Chr.4: 1986384-1986384 on Build GRCh38
rs574001743 Chr.4: 1986088-1986088 on Build GRCh38
rs1053782912 Chr.4: 1986272-1986272 on Build GRCh38
rs1077020 Chr.4: 1986466-1986466 on Build GRCh38
rs775048821 Chr.4: 1986193-1986193 on Build GRCh38
rs749053117 Chr.4: 1986029-1986029 on Build GRCh38
rs779957545 Chr.4: 1986321-1986321 on Build GRCh38
rs995631451 Chr.4: 1986059-1986059 on Build GRCh38
rs749188582 Chr.4: 1986131-1986131 on Build GRCh38
rs1025161769 Chr.4: 1986409-1986409 on Build GRCh38
rs376625058 Chr.4: 1986436-1986436 on Build GRCh38
rs962249137 Chr.4: 1986405-1986405 on Build GRCh38
rs374954650 Chr.4: 1986091-1986091 on Build GRCh38
rs1052710847 Chr.4: 1986460-1986460 on Build GRCh38
rs779998574 Chr.4: 1986238-1986238 on Build GRCh38
rs752852498 Chr.4: 1986225-1986225 on Build GRCh38
rs527649560 Chr.4: 1986244-1986244 on Build GRCh38
rs916410813 Chr.4: 1986144-1986144 on Build GRCh38
rs766835829 Chr.4: 1986207-1986207 on Build GRCh38
rs760256257 Chr.4: 1986190-1986190 on Build GRCh38
rs918208825 Chr.4: 1986036-1986036 on Build GRCh38
rs753927968 Chr.4: 1986311-1986311 on Build GRCh38
rs770876912 Chr.4: 1986030-1986030 on Build GRCh38
rs200107238 Chr.4: 1986392-1986392 on Build GRCh38
rs149004004 Chr.4: 1986166-1986166 on Build GRCh38
rs750317727 Chr.4: 1986316-1986316 on Build GRCh38
rs767815116 Chr.4: 1986271-1986271 on Build GRCh38
rs897246964 Chr.4: 1986058-1986058 on Build GRCh38
rs375025131 Chr.4: 1986132-1986132 on Build GRCh38
rs376220630 Chr.4: 1986240-1986240 on Build GRCh38
rs529227132 Chr.4: 1986353-1986353 on Build GRCh38
rs1046381223 Chr.4: 1986033-1986033 on Build GRCh38
rs541375712 Chr.4: 1986097-1986097 on Build GRCh38
rs369907817 Chr.4: 1986232-1986232 on Build GRCh38
rs760843788 Chr.4: 1986300-1986300 on Build GRCh38
rs750550938 Chr.4: 1986402-1986402 on Build GRCh38
rs746599876 Chr.4: 1986239-1986239 on Build GRCh38
rs949615993 Chr.4: 1986052-1986052 on Build GRCh38
rs999616765 Chr.4: 1986093-1986093 on Build GRCh38
rs372992255 Chr.4: 1986233-1986233 on Build GRCh38
rs751035859 Chr.4: 1986086-1986086 on Build GRCh38
rs773406107 Chr.4: 1986461-1986461 on Build GRCh38
rs984129819 Chr.4: 1986422-1986422 on Build GRCh38
rs373731879 Chr.4: 1986453-1986453 on Build GRCh38
rs778725431 Chr.4: 1986137-1986137 on Build GRCh38
rs369468339 Chr.4: 1986205-1986205 on Build GRCh38
rs2234565 Chr.4: 1986376-1986376 on Build GRCh38
rs377602638 Chr.4: 1986270-1986270 on Build GRCh38
rs145918918 Chr.4: 1986356-1986356 on Build GRCh38
rs377031993 Chr.4: 1986263-1986263 on Build GRCh38
rs756256163 Chr.4: 1986098-1986098 on Build GRCh38
rs571972940 Chr.4: 1986468-1986468 on Build GRCh38
rs776785724 Chr.4: 1986383-1986383 on Build GRCh38
rs775798532 Chr.4: 1986365-1986365 on Build GRCh38
rs746842666 Chr.4: 1986337-1986337 on Build GRCh38
rs772059482 Chr.4: 1986236-1986236 on Build GRCh38
rs745686118 Chr.4: 1986143-1986143 on Build GRCh38
rs373364109 Chr.4: 1986096-1986096 on Build GRCh38
rs537747868 Chr.4: 1986037-1986037 on Build GRCh38
rs777778901 Chr.4: 1986228-1986228 on Build GRCh38
rs767305085 Chr.4: 1986065-1986065 on Build GRCh38
rs2234566 Chr.4: 1986087-1986087 on Build GRCh38
rs201974342 Chr.4: 1986443-1986443 on Build GRCh38
rs770096895 Chr.4: 1986456-1986456 on Build GRCh38
rs78020018 Chr.4: 1986258-1986258 on Build GRCh38
rs916441943 Chr.4: 1986016-1986016 on Build GRCh38
rs537205291 Chr.4: 1986426-1986426 on Build GRCh38
rs758421252 Chr.4: 1986319-1986319 on Build GRCh38
rs772541698 Chr.4: 1986442-1986442 on Build GRCh38
rs370942710 Chr.4: 1986291-1986291 on Build GRCh38

View Full Product Details