Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116768224-116768738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2
Entrez Gene ID
914
Gene Name
CD2 molecule
Gene Aliases
LFA-2, SRBC, T11
UniGene
Hs.523500
Gene Chromosome Location
Chr.1: 116754464-116769229 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4127560 c.957A>G p.Q319Q Chr.1: 116768684-116768684 on Build GRCh38
7360221 c.955C>A p.Q319K Chr.1: 116768682-116768682 on Build GRCh38
5809409 c.850C>A p.P284T Chr.1: 116768577-116768577 on Build GRCh38
4430615 c.974G>A p.G325D Chr.1: 116768701-116768701 on Build GRCh38
5377336 c.805C>T p.P269S Chr.1: 116768532-116768532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770748992 Chr.1: 116768422-116768422 on Build GRCh38
rs773596778 Chr.1: 116768587-116768587 on Build GRCh38
rs778439027 Chr.1: 116768660-116768660 on Build GRCh38
rs377222693 Chr.1: 116768613-116768613 on Build GRCh38
rs760958788 Chr.1: 116768509-116768509 on Build GRCh38
rs201567293 Chr.1: 116768503-116768503 on Build GRCh38
rs772447791 Chr.1: 116768620-116768620 on Build GRCh38
rs267597947 Chr.1: 116768691-116768691 on Build GRCh38
rs888930708 Chr.1: 116768353-116768353 on Build GRCh38
rs747590344 Chr.1: 116768661-116768661 on Build GRCh38
rs750326100 Chr.1: 116768690-116768690 on Build GRCh38
rs1055585474 Chr.1: 116768460-116768460 on Build GRCh38
rs769448924 Chr.1: 116768442-116768442 on Build GRCh38
rs748882774 Chr.1: 116768635-116768635 on Build GRCh38
rs745490266 Chr.1: 116768440-116768440 on Build GRCh38
rs780725107 Chr.1: 116768424-116768424 on Build GRCh38
rs777077219 Chr.1: 116768388-116768388 on Build GRCh38
rs780987566 Chr.1: 116768547-116768547 on Build GRCh38
rs771873581 Chr.1: 116768322-116768322 on Build GRCh38
rs954810232 Chr.1: 116768398-116768398 on Build GRCh38
rs761203896 Chr.1: 116768591-116768591 on Build GRCh38
rs759864783 Chr.1: 116768600-116768600 on Build GRCh38
rs777102347 Chr.1: 116768593-116768593 on Build GRCh38
rs767546325 Chr.1: 116768689-116768689 on Build GRCh38
rs765512102 Chr.1: 116768603-116768603 on Build GRCh38
rs543762110 Chr.1: 116768379-116768379 on Build GRCh38
rs151304988 Chr.1: 116768709-116768709 on Build GRCh38
rs778399584 Chr.1: 116768710-116768710 on Build GRCh38
rs999871668 Chr.1: 116768546-116768546 on Build GRCh38
rs755975526 Chr.1: 116768695-116768695 on Build GRCh38
rs568137617 Chr.1: 116768543-116768543 on Build GRCh38
rs763036877 Chr.1: 116768611-116768611 on Build GRCh38
rs1000163147 Chr.1: 116768346-116768346 on Build GRCh38
rs745624462 Chr.1: 116768549-116768549 on Build GRCh38
rs754039794 Chr.1: 116768514-116768514 on Build GRCh38
rs576400985 Chr.1: 116768319-116768319 on Build GRCh38
rs1028856945 Chr.1: 116768331-116768331 on Build GRCh38
rs764171109 Chr.1: 116768612-116768612 on Build GRCh38
rs864622018 Chr.1: 116768701-116768701 on Build GRCh38
rs766698238 Chr.1: 116768512-116768512 on Build GRCh38
rs770420450 Chr.1: 116768451-116768451 on Build GRCh38
rs748864256 Chr.1: 116768582-116768582 on Build GRCh38
rs779845333 Chr.1: 116768577-116768577 on Build GRCh38
rs961921996 Chr.1: 116768393-116768393 on Build GRCh38
rs773828485 Chr.1: 116768461-116768461 on Build GRCh38
rs1019935192 Chr.1: 116768313-116768313 on Build GRCh38
rs777001150 Chr.1: 116768670-116768670 on Build GRCh38
rs763125368 Chr.1: 116768675-116768675 on Build GRCh38
rs766235998 Chr.1: 116768704-116768704 on Build GRCh38
rs759707410 Chr.1: 116768515-116768515 on Build GRCh38
rs777632148 Chr.1: 116768529-116768529 on Build GRCh38
rs775184596 Chr.1: 116768447-116768447 on Build GRCh38
rs145678592 Chr.1: 116768614-116768614 on Build GRCh38
rs543369186 Chr.1: 116768275-116768275 on Build GRCh38
rs377723619 Chr.1: 116768476-116768476 on Build GRCh38
rs866683032 Chr.1: 116768383-116768383 on Build GRCh38
rs775679910 Chr.1: 116768604-116768604 on Build GRCh38
rs746850439 Chr.1: 116768415-116768415 on Build GRCh38
rs922009355 Chr.1: 116768394-116768394 on Build GRCh38
rs958412838 Chr.1: 116768391-116768391 on Build GRCh38
rs699738 Chr.1: 116768525-116768525 on Build GRCh38
rs1008857122 Chr.1: 116768382-116768382 on Build GRCh38
rs937171505 Chr.1: 116768450-116768450 on Build GRCh38
rs973466125 Chr.1: 116768680-116768680 on Build GRCh38
rs35591188 Chr.1: 116768672-116768672 on Build GRCh38
rs527463622 Chr.1: 116768583-116768583 on Build GRCh38
rs754565683 Chr.1: 116768644-116768644 on Build GRCh38
rs10489634 Chr.1: 116768392-116768392 on Build GRCh38
rs150567561 Chr.1: 116768535-116768535 on Build GRCh38
rs369716425 Chr.1: 116768494-116768494 on Build GRCh38
rs761791386 Chr.1: 116768688-116768688 on Build GRCh38
rs771511394 Chr.1: 116768662-116768662 on Build GRCh38
rs769607172 Chr.1: 116768567-116768567 on Build GRCh38
rs199588776 Chr.1: 116768627-116768627 on Build GRCh38
rs966968810 Chr.1: 116768327-116768327 on Build GRCh38
rs764196122 Chr.1: 116768681-116768681 on Build GRCh38
rs758471774 Chr.1: 116768528-116768528 on Build GRCh38
rs750307503 Chr.1: 116768621-116768621 on Build GRCh38
rs532610592 Chr.1: 116768466-116768466 on Build GRCh38
rs370570850 Chr.1: 116768674-116768674 on Build GRCh38
rs373098399 Chr.1: 116768511-116768511 on Build GRCh38
rs771222747 Chr.1: 116768592-116768592 on Build GRCh38
rs987456909 Chr.1: 116768400-116768400 on Build GRCh38
rs753565810 Chr.1: 116768705-116768705 on Build GRCh38
rs774525354 Chr.1: 116768682-116768682 on Build GRCh38
rs779863841 Chr.1: 116768634-116768634 on Build GRCh38
rs1037996061 Chr.1: 116768436-116768436 on Build GRCh38
rs200585938 Chr.1: 116768497-116768497 on Build GRCh38
rs1032607030 Chr.1: 116768376-116768376 on Build GRCh38
rs940722868 Chr.1: 116768416-116768416 on Build GRCh38
rs559951468 Chr.1: 116768553-116768553 on Build GRCh38
rs746292283 Chr.1: 116768671-116768671 on Build GRCh38
rs371859358 Chr.1: 116768312-116768312 on Build GRCh38
rs1013865519 Chr.1: 116768286-116768286 on Build GRCh38

View Full Product Details