Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37443195-37443692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD2
Entrez Gene ID
57495
Gene Name
NACHT and WD repeat domain containing 2
Gene Aliases
KIAA1239
UniGene
Hs.4280
Gene Chromosome Location
Chr.4: 37245068-37449465 on Build GRCh38
Transcripts
NM_001144990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5943743 c.1626delC p.L543fs*14 Chr.4: 37443614-37443614 on Build GRCh38
4825681 c.1356A>T p.R452S Chr.4: 37443344-37443344 on Build GRCh38
4818826 c.1340C>A p.P447Q Chr.4: 37443328-37443328 on Build GRCh38
6529206 c.1518G>C p.Q506H Chr.4: 37443506-37443506 on Build GRCh38
6865674 c.1330G>T p.E444* Chr.4: 37443318-37443318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535613170 Chr.4: 37443424-37443424 on Build GRCh38
rs890395770 Chr.4: 37443639-37443639 on Build GRCh38
rs777002295 Chr.4: 37443371-37443371 on Build GRCh38
rs373302413 Chr.4: 37443554-37443554 on Build GRCh38
rs748145773 Chr.4: 37443623-37443623 on Build GRCh38
rs996853716 Chr.4: 37443305-37443305 on Build GRCh38
rs867395346 Chr.4: 37443490-37443490 on Build GRCh38
rs763111526 Chr.4: 37443473-37443473 on Build GRCh38
rs867836939 Chr.4: 37443499-37443499 on Build GRCh38
rs1022429491 Chr.4: 37443347-37443347 on Build GRCh38
rs756877795 Chr.4: 37443598-37443598 on Build GRCh38
rs751884123 Chr.4: 37443250-37443250 on Build GRCh38
rs1008787026 Chr.4: 37443650-37443650 on Build GRCh38
rs780033189 Chr.4: 37443278-37443278 on Build GRCh38
rs892139373 Chr.4: 37443664-37443664 on Build GRCh38
rs778856174 Chr.4: 37443622-37443622 on Build GRCh38
rs751103000 Chr.4: 37443527-37443527 on Build GRCh38
rs747628178 Chr.4: 37443335-37443335 on Build GRCh38
rs777307279 Chr.4: 37443659-37443659 on Build GRCh38
rs757902890 Chr.4: 37443638-37443638 on Build GRCh38
rs370592141 Chr.4: 37443228-37443228 on Build GRCh38
rs980935501 Chr.4: 37443358-37443358 on Build GRCh38
rs11944914 Chr.4: 37443464-37443464 on Build GRCh38
rs376203240 Chr.4: 37443588-37443588 on Build GRCh38
rs1050204781 Chr.4: 37443620-37443620 on Build GRCh38
rs765596515 Chr.4: 37443356-37443356 on Build GRCh38
rs749161360 Chr.4: 37443286-37443286 on Build GRCh38
rs375358436 Chr.4: 37443423-37443423 on Build GRCh38
rs996907171 Chr.4: 37443608-37443608 on Build GRCh38
rs757604933 Chr.4: 37443559-37443559 on Build GRCh38
rs142523381 Chr.4: 37443522-37443522 on Build GRCh38
rs1045959275 Chr.4: 37443245-37443245 on Build GRCh38
rs771588035 Chr.4: 37443338-37443338 on Build GRCh38
rs893111904 Chr.4: 37443606-37443606 on Build GRCh38
rs916045721 Chr.4: 37443219-37443219 on Build GRCh38
rs767657395 Chr.4: 37443569-37443569 on Build GRCh38
rs753024880 Chr.4: 37443461-37443461 on Build GRCh38
rs778751181 Chr.4: 37443324-37443324 on Build GRCh38
rs868385964 Chr.4: 37443663-37443663 on Build GRCh38
rs755965472 Chr.4: 37443266-37443266 on Build GRCh38
rs941323274 Chr.4: 37443530-37443530 on Build GRCh38
rs971718582 Chr.4: 37443382-37443382 on Build GRCh38
rs148039237 Chr.4: 37443339-37443339 on Build GRCh38
rs1052753854 Chr.4: 37443653-37443653 on Build GRCh38
rs998805702 Chr.4: 37443285-37443285 on Build GRCh38
rs759862811 Chr.4: 37443268-37443268 on Build GRCh38
rs934874792 Chr.4: 37443593-37443593 on Build GRCh38
rs539623310 Chr.4: 37443301-37443301 on Build GRCh38
rs1010670089 Chr.4: 37443340-37443340 on Build GRCh38
rs950360711 Chr.4: 37443232-37443232 on Build GRCh38
rs1029258179 Chr.4: 37443308-37443308 on Build GRCh38
rs573305620 Chr.4: 37443343-37443343 on Build GRCh38
rs751798672 Chr.4: 37443528-37443528 on Build GRCh38
rs951782182 Chr.4: 37443457-37443457 on Build GRCh38
rs372373854 Chr.4: 37443355-37443355 on Build GRCh38

View Full Product Details