Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42402121-42402630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHISA3
Entrez Gene ID
152573
Gene Name
shisa family member 3
Gene Aliases
hShisa3
UniGene
Hs.370904
Gene Chromosome Location
Chr.4: 42397839-42402487 on Build GRCh38
Transcripts
NM_001080505.1

SNP Detail

SNP ID Chromosome Location
rs572433790 Chr.4: 42402207-42402207 on Build GRCh38
rs73235556 Chr.4: 42402499-42402499 on Build GRCh38
rs956532814 Chr.4: 42402415-42402415 on Build GRCh38
rs935275293 Chr.4: 42402150-42402150 on Build GRCh38
rs1052550138 Chr.4: 42402605-42402605 on Build GRCh38
rs960966057 Chr.4: 42402291-42402291 on Build GRCh38
rs1023623529 Chr.4: 42402304-42402306 on Build GRCh38
rs984463196 Chr.4: 42402221-42402221 on Build GRCh38
rs760616042 Chr.4: 42402548-42402548 on Build GRCh38
rs888884916 Chr.4: 42402245-42402245 on Build GRCh38
rs538989596 Chr.4: 42402174-42402174 on Build GRCh38
rs574254920 Chr.4: 42402292-42402292 on Build GRCh38
rs763174428 Chr.4: 42402151-42402151 on Build GRCh38
rs545916512 Chr.4: 42402440-42402440 on Build GRCh38
rs778143397 Chr.4: 42402287-42402290 on Build GRCh38
rs376155968 Chr.4: 42402513-42402513 on Build GRCh38
rs192778473 Chr.4: 42402507-42402507 on Build GRCh38
rs948289192 Chr.4: 42402273-42402273 on Build GRCh38
rs188728229 Chr.4: 42402286-42402286 on Build GRCh38
rs766120175 Chr.4: 42402557-42402557 on Build GRCh38
rs970149069 Chr.4: 42402569-42402569 on Build GRCh38
rs540924297 Chr.4: 42402248-42402248 on Build GRCh38
rs143022504 Chr.4: 42402535-42402535 on Build GRCh38
rs923197746 Chr.4: 42402436-42402436 on Build GRCh38
rs1011447321 Chr.4: 42402441-42402441 on Build GRCh38
rs772765578 Chr.4: 42402330-42402332 on Build GRCh38
rs558664538 Chr.4: 42402199-42402199 on Build GRCh38
rs73235555 Chr.4: 42402371-42402371 on Build GRCh38
rs1043693610 Chr.4: 42402469-42402469 on Build GRCh38
rs915599340 Chr.4: 42402264-42402264 on Build GRCh38
rs563371665 Chr.4: 42402368-42402368 on Build GRCh38
rs951825180 Chr.4: 42402549-42402549 on Build GRCh38
rs969380359 Chr.4: 42402586-42402586 on Build GRCh38
rs528138038 Chr.4: 42402480-42402480 on Build GRCh38
rs984948434 Chr.4: 42402556-42402556 on Build GRCh38
rs971783880 Chr.4: 42402171-42402171 on Build GRCh38
rs965881226 Chr.4: 42402167-42402167 on Build GRCh38
rs11939228 Chr.4: 42402563-42402563 on Build GRCh38
rs930414648 Chr.4: 42402187-42402187 on Build GRCh38
rs750157456 Chr.4: 42402527-42402527 on Build GRCh38
rs570264847 Chr.4: 42402579-42402579 on Build GRCh38
rs925317576 Chr.4: 42402402-42402402 on Build GRCh38
rs932992925 Chr.4: 42402175-42402175 on Build GRCh38
rs969494881 Chr.4: 42402353-42402353 on Build GRCh38
rs559711362 Chr.4: 42402299-42402302 on Build GRCh38
rs906688829 Chr.4: 42402287-42402287 on Build GRCh38
rs928208823 Chr.4: 42402580-42402580 on Build GRCh38
rs184858884 Chr.4: 42402161-42402161 on Build GRCh38
rs939639863 Chr.4: 42402593-42402593 on Build GRCh38
rs530270248 Chr.4: 42402524-42402524 on Build GRCh38
rs1005921497 Chr.4: 42402247-42402247 on Build GRCh38
rs987961544 Chr.4: 42402419-42402419 on Build GRCh38
rs993317789 Chr.4: 42402542-42402542 on Build GRCh38
rs896759667 Chr.4: 42402560-42402560 on Build GRCh38
rs76964688 Chr.4: 42402372-42402372 on Build GRCh38
rs1050068343 Chr.4: 42402242-42402242 on Build GRCh38
rs774518954 Chr.4: 42402309-42402309 on Build GRCh38
rs1050540760 Chr.4: 42402465-42402465 on Build GRCh38
rs559062406 Chr.4: 42402466-42402466 on Build GRCh38
rs543219915 Chr.4: 42402295-42402295 on Build GRCh38

View Full Product Details