Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52060338-52060829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA18
Entrez Gene ID
132671
Gene Name
spermatogenesis associated 18
Gene Aliases
Mieap, SPETEX1
UniGene
Hs.527090
Gene Chromosome Location
Chr.4: 52051331-52097305 on Build GRCh38
Transcripts
NM_001297608.1 NM_145263.3 NR_123728.1 XM_005265716.1 XM_005265717.2 XM_005265718.1 XM_011534369.1 XM_011534370.1 XM_011534372.1 XM_017007731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125052 c.206A>T p.D69V Chr.4: 52060794-52060794 on Build GRCh38
6100324 c.211G>T p.V71L Chr.4: 52060799-52060799 on Build GRCh38
1055929 c.152A>G p.Q51R Chr.4: 52060483-52060483 on Build GRCh38
3604410 c.166G>A p.G56R Chr.4: 52060497-52060497 on Build GRCh38
1055927 c.129C>A p.L43L Chr.4: 52060460-52060460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995159794 Chr.4: 52060659-52060660 on Build GRCh38
rs764662108 Chr.4: 52060797-52060797 on Build GRCh38
rs942512943 Chr.4: 52060660-52060660 on Build GRCh38
rs374661277 Chr.4: 52060504-52060504 on Build GRCh38
rs748102742 Chr.4: 52060538-52060538 on Build GRCh38
rs370337585 Chr.4: 52060744-52060744 on Build GRCh38
rs769882469 Chr.4: 52060525-52060525 on Build GRCh38
rs1013028449 Chr.4: 52060661-52060661 on Build GRCh38
rs763337012 Chr.4: 52060663-52060663 on Build GRCh38
rs1027522612 Chr.4: 52060660-52060660 on Build GRCh38
rs368693724 Chr.4: 52060512-52060512 on Build GRCh38
rs771866295 Chr.4: 52060541-52060541 on Build GRCh38
rs199772940 Chr.4: 52060458-52060458 on Build GRCh38
rs911651669 Chr.4: 52060803-52060803 on Build GRCh38
rs138025019 Chr.4: 52060478-52060478 on Build GRCh38
rs567063333 Chr.4: 52060788-52060788 on Build GRCh38
rs766070707 Chr.4: 52060554-52060554 on Build GRCh38
rs200997138 Chr.4: 52060787-52060787 on Build GRCh38
rs746094784 Chr.4: 52060754-52060754 on Build GRCh38
rs146976675 Chr.4: 52060450-52060450 on Build GRCh38
rs374357974 Chr.4: 52060398-52060398 on Build GRCh38
rs371430744 Chr.4: 52060765-52060765 on Build GRCh38
rs759373861 Chr.4: 52060746-52060747 on Build GRCh38
rs753244156 Chr.4: 52060470-52060470 on Build GRCh38
rs918503758 Chr.4: 52060389-52060389 on Build GRCh38
rs1038394739 Chr.4: 52060387-52060387 on Build GRCh38
rs772922983 Chr.4: 52060548-52060548 on Build GRCh38
rs1000938453 Chr.4: 52060430-52060430 on Build GRCh38
rs760261292 Chr.4: 52060438-52060438 on Build GRCh38
rs892012106 Chr.4: 52060461-52060461 on Build GRCh38
rs776788321 Chr.4: 52060412-52060412 on Build GRCh38
rs774413394 Chr.4: 52060478-52060480 on Build GRCh38
rs191477749 Chr.4: 52060359-52060359 on Build GRCh38
rs1008575545 Chr.4: 52060614-52060614 on Build GRCh38
rs770599522 Chr.4: 52060394-52060396 on Build GRCh38
rs573027367 Chr.4: 52060411-52060411 on Build GRCh38
rs567884182 Chr.4: 52060667-52060667 on Build GRCh38
rs759035761 Chr.4: 52060561-52060561 on Build GRCh38
rs747225021 Chr.4: 52060399-52060399 on Build GRCh38
rs781306576 Chr.4: 52060751-52060751 on Build GRCh38
rs772534419 Chr.4: 52060394-52060394 on Build GRCh38
rs753239827 Chr.4: 52060448-52060448 on Build GRCh38
rs113204633 Chr.4: 52060637-52060637 on Build GRCh38
rs139975898 Chr.4: 52060755-52060755 on Build GRCh38
rs750757528 Chr.4: 52060597-52060597 on Build GRCh38
rs370633778 Chr.4: 52060498-52060498 on Build GRCh38
rs773113022 Chr.4: 52060784-52060784 on Build GRCh38
rs936585924 Chr.4: 52060584-52060595 on Build GRCh38
rs903594390 Chr.4: 52060669-52060669 on Build GRCh38
rs753161765 Chr.4: 52060570-52060570 on Build GRCh38
rs375823281 Chr.4: 52060772-52060772 on Build GRCh38
rs576685620 Chr.4: 52060764-52060764 on Build GRCh38
rs866930605 Chr.4: 52060401-52060401 on Build GRCh38
rs764587218 Chr.4: 52060733-52060733 on Build GRCh38
rs141339947 Chr.4: 52060427-52060427 on Build GRCh38
rs764794740 Chr.4: 52060568-52060568 on Build GRCh38
rs762976290 Chr.4: 52060415-52060415 on Build GRCh38
rs558596998 Chr.4: 52060676-52060676 on Build GRCh38
rs200396040 Chr.4: 52060372-52060372 on Build GRCh38
rs758911276 Chr.4: 52060576-52060576 on Build GRCh38
rs924902747 Chr.4: 52060484-52060484 on Build GRCh38
rs1039849838 Chr.4: 52060426-52060426 on Build GRCh38
rs761549752 Chr.4: 52060422-52060422 on Build GRCh38
rs199804628 Chr.4: 52060659-52060659 on Build GRCh38
rs936440305 Chr.4: 52060690-52060690 on Build GRCh38
rs534567049 Chr.4: 52060800-52060800 on Build GRCh38
rs892148684 Chr.4: 52060711-52060711 on Build GRCh38
rs897741658 Chr.4: 52060640-52060640 on Build GRCh38
rs374603453 Chr.4: 52060466-52060466 on Build GRCh38
rs1052010533 Chr.4: 52060706-52060706 on Build GRCh38
rs200441243 Chr.4: 52060419-52060419 on Build GRCh38
rs767294350 Chr.4: 52060423-52060423 on Build GRCh38
rs370448764 Chr.4: 52060382-52060382 on Build GRCh38
rs776316460 Chr.4: 52060560-52060560 on Build GRCh38
rs759174259 Chr.4: 52060793-52060793 on Build GRCh38
rs866361341 Chr.4: 52060699-52060699 on Build GRCh38
rs977458068 Chr.4: 52060714-52060714 on Build GRCh38
rs146015590 Chr.4: 52060500-52060500 on Build GRCh38
rs183444588 Chr.4: 52060799-52060799 on Build GRCh38
rs760344887 Chr.4: 52060551-52060551 on Build GRCh38
rs994685733 Chr.4: 52060406-52060406 on Build GRCh38
rs755021887 Chr.4: 52060792-52060792 on Build GRCh38
rs766532432 Chr.4: 52060742-52060742 on Build GRCh38
rs551177428 Chr.4: 52060650-52060650 on Build GRCh38
rs183316171 Chr.4: 52060371-52060371 on Build GRCh38
rs781352630 Chr.4: 52060508-52060508 on Build GRCh38
rs1054557439 Chr.4: 52060494-52060494 on Build GRCh38
rs754490890 Chr.4: 52060457-52060457 on Build GRCh38
rs929855665 Chr.4: 52060578-52060578 on Build GRCh38
rs774218221 Chr.4: 52060526-52060526 on Build GRCh38
rs187371217 Chr.4: 52060440-52060440 on Build GRCh38
rs201717083 Chr.4: 52060491-52060491 on Build GRCh38
rs757670669 Chr.4: 52060746-52060746 on Build GRCh38
rs55653366 Chr.4: 52060418-52060418 on Build GRCh38

View Full Product Details