Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52751206-52751697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVMER34-1
Entrez Gene ID
100288413
Gene Name
endogenous retrovirus group MER34 member 1
Gene Aliases
-
UniGene
Hs.363087
Gene Chromosome Location
Chr.4: 52743517-52751640 on Build GRCh38
Transcripts
NM_001242690.1 NM_024534.5

SNP Detail

SNP ID Chromosome Location
rs934234873 Chr.4: 52751352-52751352 on Build GRCh38
rs1024169510 Chr.4: 52751468-52751468 on Build GRCh38
rs1016751289 Chr.4: 52751549-52751549 on Build GRCh38
rs192754782 Chr.4: 52751592-52751592 on Build GRCh38
rs914036920 Chr.4: 52751404-52751404 on Build GRCh38
rs904262208 Chr.4: 52751429-52751429 on Build GRCh38
rs985660038 Chr.4: 52751262-52751262 on Build GRCh38
rs1021293202 Chr.4: 52751597-52751597 on Build GRCh38
rs567280196 Chr.4: 52751562-52751562 on Build GRCh38
rs1003982885 Chr.4: 52751511-52751511 on Build GRCh38
rs945518141 Chr.4: 52751411-52751411 on Build GRCh38
rs891501712 Chr.4: 52751507-52751507 on Build GRCh38
rs558987128 Chr.4: 52751659-52751659 on Build GRCh38
rs951226796 Chr.4: 52751255-52751255 on Build GRCh38
rs550636382 Chr.4: 52751492-52751492 on Build GRCh38
rs935726764 Chr.4: 52751647-52751647 on Build GRCh38
rs921277430 Chr.4: 52751309-52751309 on Build GRCh38
rs1018913616 Chr.4: 52751632-52751632 on Build GRCh38
rs968998770 Chr.4: 52751616-52751616 on Build GRCh38
rs1016971428 Chr.4: 52751263-52751263 on Build GRCh38
rs113816783 Chr.4: 52751265-52751265 on Build GRCh38
rs138716124 Chr.4: 52751583-52751583 on Build GRCh38
rs975426065 Chr.4: 52751293-52751293 on Build GRCh38
rs963142749 Chr.4: 52751275-52751275 on Build GRCh38
rs373300771 Chr.4: 52751669-52751669 on Build GRCh38
rs925463834 Chr.4: 52751643-52751643 on Build GRCh38
rs538490753 Chr.4: 52751633-52751633 on Build GRCh38
rs941947614 Chr.4: 52751431-52751431 on Build GRCh38
rs1014607954 Chr.4: 52751244-52751244 on Build GRCh38
rs1035465410 Chr.4: 52751473-52751473 on Build GRCh38
rs914152918 Chr.4: 52751623-52751623 on Build GRCh38
rs1028282968 Chr.4: 52751588-52751588 on Build GRCh38
rs911392117 Chr.4: 52751280-52751280 on Build GRCh38
rs987459454 Chr.4: 52751386-52751386 on Build GRCh38
rs944144447 Chr.4: 52751314-52751314 on Build GRCh38
rs918686401 Chr.4: 52751674-52751674 on Build GRCh38
rs966982250 Chr.4: 52751629-52751629 on Build GRCh38
rs887065551 Chr.4: 52751502-52751502 on Build GRCh38
rs986831187 Chr.4: 52751611-52751611 on Build GRCh38
rs573022025 Chr.4: 52751455-52751455 on Build GRCh38
rs866570265 Chr.4: 52751671-52751671 on Build GRCh38
rs1014138878 Chr.4: 52751463-52751463 on Build GRCh38
rs935764991 Chr.4: 52751439-52751439 on Build GRCh38
rs773858326 Chr.4: 52751635-52751635 on Build GRCh38
rs1043943372 Chr.4: 52751428-52751428 on Build GRCh38
rs566624031 Chr.4: 52751612-52751612 on Build GRCh38
rs530523410 Chr.4: 52751440-52751440 on Build GRCh38
rs918642406 Chr.4: 52751373-52751373 on Build GRCh38
rs551443890 Chr.4: 52751251-52751251 on Build GRCh38
rs929980620 Chr.4: 52751379-52751379 on Build GRCh38
rs116370051 Chr.4: 52751449-52751449 on Build GRCh38
rs564861318 Chr.4: 52751256-52751256 on Build GRCh38
rs955563023 Chr.4: 52751604-52751604 on Build GRCh38

View Full Product Details