Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56396455-56397000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPAT
Entrez Gene ID
5471
Gene Name
phosphoribosyl pyrophosphate amidotransferase
Gene Aliases
ATASE, GPAT, PRAT
UniGene
Hs.331420
Gene Chromosome Location
Chr.4: 56393362-56435636 on Build GRCh38
Transcripts
NM_002703.4 XM_011534387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6535337 c.1357G>C p.G453R Chr.4: 56396619-56396619 on Build GRCh38
3993732 c.1253C>T p.A418V Chr.4: 56396723-56396723 on Build GRCh38
6447052 c.1356A>C p.L452L Chr.4: 56396620-56396620 on Build GRCh38
6399040 c.1237G>T p.V413L Chr.4: 56396739-56396739 on Build GRCh38
1430308 c.1298C>A p.P433H Chr.4: 56396678-56396678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765223715 Chr.4: 56396791-56396791 on Build GRCh38
rs1001307050 Chr.4: 56396531-56396531 on Build GRCh38
rs777842143 Chr.4: 56396578-56396578 on Build GRCh38
rs373657410 Chr.4: 56396566-56396566 on Build GRCh38
rs974837731 Chr.4: 56396883-56396883 on Build GRCh38
rs368755475 Chr.4: 56396784-56396784 on Build GRCh38
rs766194038 Chr.4: 56396778-56396778 on Build GRCh38
rs763309864 Chr.4: 56396612-56396615 on Build GRCh38
rs770028075 Chr.4: 56396739-56396739 on Build GRCh38
rs751646898 Chr.4: 56396589-56396589 on Build GRCh38
rs763355375 Chr.4: 56396915-56396915 on Build GRCh38
rs749814871 Chr.4: 56396670-56396670 on Build GRCh38
rs932540971 Chr.4: 56396695-56396695 on Build GRCh38
rs905679848 Chr.4: 56396507-56396507 on Build GRCh38
rs111521601 Chr.4: 56396958-56396958 on Build GRCh38
rs776255491 Chr.4: 56396588-56396591 on Build GRCh38
rs1023106352 Chr.4: 56396884-56396884 on Build GRCh38
rs540376105 Chr.4: 56396934-56396934 on Build GRCh38
rs201250405 Chr.4: 56396592-56396592 on Build GRCh38
rs760470295 Chr.4: 56396777-56396777 on Build GRCh38
rs993814729 Chr.4: 56396682-56396682 on Build GRCh38
rs191712823 Chr.4: 56396925-56396925 on Build GRCh38
rs753710607 Chr.4: 56396779-56396779 on Build GRCh38
rs187837106 Chr.4: 56396788-56396788 on Build GRCh38
rs1035983039 Chr.4: 56396878-56396878 on Build GRCh38
rs780663607 Chr.4: 56396637-56396637 on Build GRCh38
rs952511589 Chr.4: 56396884-56396887 on Build GRCh38
rs541677937 Chr.4: 56396579-56396579 on Build GRCh38
rs566943962 Chr.4: 56396481-56396481 on Build GRCh38
rs773988670 Chr.4: 56396701-56396701 on Build GRCh38
rs1016416973 Chr.4: 56396845-56396845 on Build GRCh38
rs746217808 Chr.4: 56396600-56396600 on Build GRCh38
rs768225373 Chr.4: 56396676-56396676 on Build GRCh38
rs766722911 Chr.4: 56396742-56396743 on Build GRCh38
rs376043196 Chr.4: 56396609-56396609 on Build GRCh38
rs770073845 Chr.4: 56396635-56396635 on Build GRCh38
rs781308955 Chr.4: 56396594-56396594 on Build GRCh38
rs183766324 Chr.4: 56396558-56396558 on Build GRCh38
rs774584249 Chr.4: 56396591-56396591 on Build GRCh38
rs950972727 Chr.4: 56396820-56396820 on Build GRCh38
rs761300467 Chr.4: 56396726-56396726 on Build GRCh38
rs939922576 Chr.4: 56396572-56396572 on Build GRCh38
rs964831248 Chr.4: 56396865-56396865 on Build GRCh38
rs774738569 Chr.4: 56396789-56396793 on Build GRCh38
rs550118130 Chr.4: 56396831-56396831 on Build GRCh38
rs868271645 Chr.4: 56396785-56396785 on Build GRCh38
rs923372902 Chr.4: 56396922-56396922 on Build GRCh38
rs553701803 Chr.4: 56396883-56396884 on Build GRCh38
rs903211341 Chr.4: 56396810-56396810 on Build GRCh38
rs889215199 Chr.4: 56396761-56396761 on Build GRCh38
rs772789380 Chr.4: 56396748-56396748 on Build GRCh38
rs988790550 Chr.4: 56396947-56396947 on Build GRCh38

View Full Product Details